Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 104856550-104857039 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POU3F3
Entrez Gene ID
5455
Gene Name
POU class 3 homeobox 3
Gene Aliases
BRN1, OTF8, brain-1, oct-8
UniGene
Hs.248158
Gene Chromosome Location
Chr.2: 104854068-104858574 on Build GRCh38
Transcripts
NM_006236.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1398566 c.1202C>T p.A401V Chr.2: 104856712-104856712 on Build GRCh38
3708786 c.1422C>A p.P474P Chr.2: 104856932-104856932 on Build GRCh38
5394060 c.1162G>A p.D388N Chr.2: 104856672-104856672 on Build GRCh38
1398568 c.1418C>T p.T473M Chr.2: 104856928-104856928 on Build GRCh38
378304 c.1247G>C p.S416T Chr.2: 104856757-104856757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900039938 Chr.2: 104856666-104856666 on Build GRCh38
rs758371926 Chr.2: 104856935-104856935 on Build GRCh38
rs770227812 Chr.2: 104857001-104857001 on Build GRCh38
rs773772879 Chr.2: 104856696-104856696 on Build GRCh38
rs777289770 Chr.2: 104856998-104856998 on Build GRCh38
rs769403407 Chr.2: 104856821-104856821 on Build GRCh38
rs371621473 Chr.2: 104856572-104856572 on Build GRCh38
rs199565525 Chr.2: 104856683-104856683 on Build GRCh38
rs113920817 Chr.2: 104857021-104857021 on Build GRCh38
rs766869344 Chr.2: 104856711-104856711 on Build GRCh38
rs751477427 Chr.2: 104856938-104856938 on Build GRCh38
rs576061892 Chr.2: 104856969-104856969 on Build GRCh38
rs867035121 Chr.2: 104856729-104856729 on Build GRCh38
rs761309325 Chr.2: 104856846-104856846 on Build GRCh38
rs977576430 Chr.2: 104856650-104856650 on Build GRCh38
rs768241893 Chr.2: 104856835-104856835 on Build GRCh38
rs775054761 Chr.2: 104856825-104856825 on Build GRCh38
rs756997559 Chr.2: 104856779-104856779 on Build GRCh38
rs377188135 Chr.2: 104856785-104856785 on Build GRCh38
rs755059340 Chr.2: 104856596-104856596 on Build GRCh38
rs147315122 Chr.2: 104856857-104856857 on Build GRCh38
rs143105467 Chr.2: 104856710-104856710 on Build GRCh38
rs776229188 Chr.2: 104856668-104856668 on Build GRCh38
rs759817331 Chr.2: 104856932-104856932 on Build GRCh38
rs372764445 Chr.2: 104856731-104856731 on Build GRCh38
rs770719599 Chr.2: 104856667-104856667 on Build GRCh38
rs748084015 Chr.2: 104856623-104856623 on Build GRCh38
rs764256978 Chr.2: 104856936-104856936 on Build GRCh38
rs919194877 Chr.2: 104857008-104857008 on Build GRCh38
rs778904576 Chr.2: 104856617-104856617 on Build GRCh38
rs960304746 Chr.2: 104856957-104856957 on Build GRCh38
rs779530854 Chr.2: 104856968-104856968 on Build GRCh38
rs775527775 Chr.2: 104857010-104857010 on Build GRCh38
rs867307766 Chr.2: 104856883-104856883 on Build GRCh38
rs754144331 Chr.2: 104856590-104856590 on Build GRCh38
rs140419524 Chr.2: 104856689-104856689 on Build GRCh38
rs777608264 Chr.2: 104856644-104856644 on Build GRCh38
rs377514724 Chr.2: 104856940-104856940 on Build GRCh38
rs751709007 Chr.2: 104857018-104857018 on Build GRCh38
rs768025847 Chr.2: 104856974-104856974 on Build GRCh38
rs777201991 Chr.2: 104856911-104856911 on Build GRCh38
rs746751751 Chr.2: 104856664-104856664 on Build GRCh38
rs780354913 Chr.2: 104856587-104856587 on Build GRCh38
rs1035661352 Chr.2: 104856837-104856837 on Build GRCh38
rs140129722 Chr.2: 104856716-104856716 on Build GRCh38
rs774903968 Chr.2: 104856685-104856685 on Build GRCh38
rs374340255 Chr.2: 104856995-104856995 on Build GRCh38
rs950008916 Chr.2: 104856960-104856960 on Build GRCh38
rs758331254 Chr.2: 104856626-104856626 on Build GRCh38
rs767968798 Chr.2: 104856693-104856693 on Build GRCh38
rs915893139 Chr.2: 104856920-104856920 on Build GRCh38
rs745493142 Chr.2: 104856819-104856819 on Build GRCh38
rs137877846 Chr.2: 104856725-104856725 on Build GRCh38
rs146486201 Chr.2: 104856697-104856697 on Build GRCh38
rs745546456 Chr.2: 104856671-104856671 on Build GRCh38
rs748677714 Chr.2: 104856832-104856832 on Build GRCh38
rs778602880 Chr.2: 104856981-104856981 on Build GRCh38
rs756558999 Chr.2: 104856581-104856581 on Build GRCh38
rs759751110 Chr.2: 104857000-104857000 on Build GRCh38
rs755952227 Chr.2: 104856964-104856964 on Build GRCh38
rs758516234 Chr.2: 104856740-104856740 on Build GRCh38
rs777507294 Chr.2: 104856749-104856749 on Build GRCh38
rs746982147 Chr.2: 104856778-104856778 on Build GRCh38
rs202016096 Chr.2: 104856983-104856983 on Build GRCh38
rs750280176 Chr.2: 104856950-104856950 on Build GRCh38
rs780965214 Chr.2: 104856947-104856947 on Build GRCh38

View Full Product Details