Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 134859028-134859541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACMSD
Entrez Gene ID
130013
Gene Name
aminocarboxymuconate semialdehyde decarboxylase
Gene Aliases
-
UniGene
Hs.655728
Gene Chromosome Location
Chr.2: 134838319-134902906 on Build GRCh38
Transcripts
NM_001307983.1 NM_138326.2 XM_005263586.4 XM_005263588.4 XM_005263589.4 XM_011510592.2 XM_017003325.1 XM_017003326.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4520049 c.103G>A p.G35R Chr.2: 134859261-134859261 on Build GRCh38
1684865 c.199G>A p.G67R Chr.2: 134859357-134859357 on Build GRCh38
1399474 c.148C>T p.R50* Chr.2: 134859306-134859306 on Build GRCh38
1006681 c.175C>T p.R59C Chr.2: 134859333-134859333 on Build GRCh38
4531229 c.176G>C p.R59P Chr.2: 134859334-134859334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042489294 Chr.2: 134859135-134859135 on Build GRCh38
rs903040306 Chr.2: 134859198-134859198 on Build GRCh38
rs750319385 Chr.2: 134859378-134859378 on Build GRCh38
rs576035999 Chr.2: 134859466-134859466 on Build GRCh38
rs573731825 Chr.2: 134859408-134859408 on Build GRCh38
rs867248338 Chr.2: 134859238-134859238 on Build GRCh38
rs542462880 Chr.2: 134859443-134859443 on Build GRCh38
rs897981429 Chr.2: 134859240-134859240 on Build GRCh38
rs267598885 Chr.2: 134859333-134859333 on Build GRCh38
rs771610050 Chr.2: 134859327-134859327 on Build GRCh38
rs993720012 Chr.2: 134859256-134859256 on Build GRCh38
rs778651133 Chr.2: 134859293-134859293 on Build GRCh38
rs776825019 Chr.2: 134859228-134859228 on Build GRCh38
rs966700440 Chr.2: 134859471-134859471 on Build GRCh38
rs768271616 Chr.2: 134859314-134859314 on Build GRCh38
rs765646424 Chr.2: 134859242-134859242 on Build GRCh38
rs1446523 Chr.2: 134859051-134859051 on Build GRCh38
rs935184916 Chr.2: 134859059-134859059 on Build GRCh38
rs750385746 Chr.2: 134859297-134859297 on Build GRCh38
rs894408891 Chr.2: 134859239-134859239 on Build GRCh38
rs12053395 Chr.2: 134859065-134859065 on Build GRCh38
rs1010850952 Chr.2: 134859110-134859110 on Build GRCh38
rs757853847 Chr.2: 134859301-134859301 on Build GRCh38
rs773545820 Chr.2: 134859325-134859325 on Build GRCh38
rs368944201 Chr.2: 134859409-134859409 on Build GRCh38
rs943589288 Chr.2: 134859058-134859058 on Build GRCh38
rs762282908 Chr.2: 134859233-134859233 on Build GRCh38
rs905222427 Chr.2: 134859157-134859157 on Build GRCh38
rs952243793 Chr.2: 134859365-134859365 on Build GRCh38
rs768969011 Chr.2: 134859210-134859210 on Build GRCh38
rs751052499 Chr.2: 134859400-134859400 on Build GRCh38
rs539329795 Chr.2: 134859275-134859275 on Build GRCh38
rs181857322 Chr.2: 134859306-134859306 on Build GRCh38
rs866389689 Chr.2: 134859357-134859357 on Build GRCh38
rs145720089 Chr.2: 134859307-134859307 on Build GRCh38
rs751796286 Chr.2: 134859269-134859269 on Build GRCh38
rs115858143 Chr.2: 134859127-134859127 on Build GRCh38
rs757143841 Chr.2: 134859292-134859292 on Build GRCh38
rs752970262 Chr.2: 134859361-134859361 on Build GRCh38
rs765101824 Chr.2: 134859370-134859370 on Build GRCh38
rs780012221 Chr.2: 134859386-134859386 on Build GRCh38
rs117363436 Chr.2: 134859321-134859321 on Build GRCh38
rs978198795 Chr.2: 134859479-134859479 on Build GRCh38
rs761455897 Chr.2: 134859362-134859362 on Build GRCh38
rs1003711521 Chr.2: 134859160-134859160 on Build GRCh38
rs767630747 Chr.2: 134859336-134859336 on Build GRCh38
rs568825076 Chr.2: 134859262-134859262 on Build GRCh38
rs547064186 Chr.2: 134859108-134859108 on Build GRCh38
rs755224773 Chr.2: 134859271-134859271 on Build GRCh38
rs1006402991 Chr.2: 134859397-134859397 on Build GRCh38
rs766592428 Chr.2: 134859265-134859265 on Build GRCh38
rs935835224 Chr.2: 134859205-134859205 on Build GRCh38
rs748454862 Chr.2: 134859320-134859320 on Build GRCh38
rs945452709 Chr.2: 134859133-134859133 on Build GRCh38
rs529060575 Chr.2: 134859245-134859245 on Build GRCh38
rs199676397 Chr.2: 134859255-134859255 on Build GRCh38
rs1052948209 Chr.2: 134859062-134859062 on Build GRCh38
rs555448669 Chr.2: 134859326-134859326 on Build GRCh38
rs964994725 Chr.2: 134859422-134859422 on Build GRCh38
rs780952596 Chr.2: 134859316-134859316 on Build GRCh38
rs375652048 Chr.2: 134859176-134859176 on Build GRCh38
rs144211190 Chr.2: 134859334-134859334 on Build GRCh38
rs758623592 Chr.2: 134859231-134859231 on Build GRCh38
rs753563049 Chr.2: 134859285-134859285 on Build GRCh38
rs780639586 Chr.2: 134859402-134859402 on Build GRCh38
rs977038800 Chr.2: 134859456-134859456 on Build GRCh38
rs528625564 Chr.2: 134859100-134859100 on Build GRCh38
rs186924885 Chr.2: 134859470-134859470 on Build GRCh38
rs557574825 Chr.2: 134859450-134859450 on Build GRCh38
rs1013209332 Chr.2: 134859276-134859276 on Build GRCh38

View Full Product Details