Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 150471500-150472030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RND3
Entrez Gene ID
390
Gene Name
Rho family GTPase 3
Gene Aliases
ARHE, Rho8, RhoE, memB
UniGene
Hs.6838
Gene Chromosome Location
Chr.2: 150468193-150487695 on Build GRCh38
Transcripts
NM_001254738.1 NM_005168.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5904730 c.364C>T p.Q122* Chr.2: 150471746-150471746 on Build GRCh38
145874 c.464C>T p.T155M Chr.2: 150471646-150471646 on Build GRCh38
6087795 c.415C>A p.L139M Chr.2: 150471695-150471695 on Build GRCh38
5711841 c.374G>T p.C125F Chr.2: 150471736-150471736 on Build GRCh38
4491269 c.377C>T p.P126L Chr.2: 150471733-150471733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768674719 Chr.2: 150471785-150471785 on Build GRCh38
rs190820528 Chr.2: 150471847-150471847 on Build GRCh38
rs948863820 Chr.2: 150471523-150471523 on Build GRCh38
rs772537086 Chr.2: 150471778-150471778 on Build GRCh38
rs924716586 Chr.2: 150471791-150471791 on Build GRCh38
rs768764713 Chr.2: 150471675-150471675 on Build GRCh38
rs531576853 Chr.2: 150471967-150471967 on Build GRCh38
rs145448904 Chr.2: 150471646-150471646 on Build GRCh38
rs766103665 Chr.2: 150471691-150471691 on Build GRCh38
rs373095716 Chr.2: 150471591-150471591 on Build GRCh38
rs917467699 Chr.2: 150471844-150471844 on Build GRCh38
rs781616489 Chr.2: 150471525-150471525 on Build GRCh38
rs780684839 Chr.2: 150471614-150471614 on Build GRCh38
rs747287295 Chr.2: 150471783-150471783 on Build GRCh38
rs751164683 Chr.2: 150471609-150471609 on Build GRCh38
rs775841994 Chr.2: 150471671-150471671 on Build GRCh38
rs993598633 Chr.2: 150471654-150471654 on Build GRCh38
rs777453229 Chr.2: 150471736-150471736 on Build GRCh38
rs780110795 Chr.2: 150471782-150471782 on Build GRCh38
rs752141865 Chr.2: 150471615-150471615 on Build GRCh38
rs1033868173 Chr.2: 150471745-150471745 on Build GRCh38
rs746037801 Chr.2: 150471658-150471658 on Build GRCh38
rs941556024 Chr.2: 150471953-150471953 on Build GRCh38
rs943187556 Chr.2: 150471536-150471536 on Build GRCh38
rs11338446 Chr.2: 150471771-150471771 on Build GRCh38
rs766193845 Chr.2: 150471580-150471580 on Build GRCh38
rs779437802 Chr.2: 150471651-150471651 on Build GRCh38
rs755543766 Chr.2: 150471733-150471733 on Build GRCh38
rs762809702 Chr.2: 150471805-150471805 on Build GRCh38
rs75090836 Chr.2: 150471529-150471529 on Build GRCh38
rs959552606 Chr.2: 150471754-150471754 on Build GRCh38
rs762900010 Chr.2: 150471690-150471690 on Build GRCh38
rs778398680 Chr.2: 150471644-150471644 on Build GRCh38
rs779577936 Chr.2: 150471770-150471771 on Build GRCh38
rs75449135 Chr.2: 150471781-150471782 on Build GRCh38
rs943239750 Chr.2: 150471939-150471941 on Build GRCh38
rs142625334 Chr.2: 150471719-150471719 on Build GRCh38
rs904919745 Chr.2: 150471543-150471543 on Build GRCh38
rs376289784 Chr.2: 150471606-150471606 on Build GRCh38
rs748092273 Chr.2: 150471558-150471558 on Build GRCh38
rs976316372 Chr.2: 150471970-150471970 on Build GRCh38
rs770848291 Chr.2: 150471806-150471806 on Build GRCh38
rs76104238 Chr.2: 150471772-150471772 on Build GRCh38
rs769673918 Chr.2: 150471717-150471717 on Build GRCh38
rs754599817 Chr.2: 150471610-150471610 on Build GRCh38
rs548656400 Chr.2: 150471998-150471998 on Build GRCh38
rs772181502 Chr.2: 150471660-150471660 on Build GRCh38
rs1017439234 Chr.2: 150471755-150471755 on Build GRCh38
rs749847768 Chr.2: 150471645-150471645 on Build GRCh38
rs1048150963 Chr.2: 150471676-150471676 on Build GRCh38
rs754342785 Chr.2: 150471747-150471747 on Build GRCh38
rs760650944 Chr.2: 150471672-150471672 on Build GRCh38
rs185474781 Chr.2: 150471804-150471804 on Build GRCh38
rs758153844 Chr.2: 150471771-150471773 on Build GRCh38
rs1040296287 Chr.2: 150471540-150471540 on Build GRCh38
rs1007361820 Chr.2: 150471748-150471748 on Build GRCh38
rs540244240 Chr.2: 150471685-150471685 on Build GRCh38
rs150247069 Chr.2: 150471861-150471861 on Build GRCh38
rs765153824 Chr.2: 150471771-150471772 on Build GRCh38
rs397986259 Chr.2: 150471782-150471782 on Build GRCh38
rs751315149 Chr.2: 150471700-150471700 on Build GRCh38
rs759440612 Chr.2: 150471711-150471711 on Build GRCh38
rs757228331 Chr.2: 150471770-150471770 on Build GRCh38
rs910051949 Chr.2: 150471884-150471884 on Build GRCh38
rs200714598 Chr.2: 150471770-150471770 on Build GRCh38
rs201814244 Chr.2: 150471765-150471765 on Build GRCh38
rs910288236 Chr.2: 150471906-150471906 on Build GRCh38
rs186861791 Chr.2: 150471535-150471535 on Build GRCh38
rs755704322 Chr.2: 150471622-150471622 on Build GRCh38
rs202144473 Chr.2: 150471783-150471783 on Build GRCh38

View Full Product Details