Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 159804182-159804698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LY75
Entrez Gene ID
4065
Gene Name
lymphocyte antigen 75
Gene Aliases
CD205, CLEC13B, DEC-205, GP200-MR6, LY-75
UniGene
Hs.153563
Gene Chromosome Location
Chr.2: 159803355-159904756 on Build GRCh38
Transcripts
NM_002349.3 NM_001198759.1 NM_001198760.1

Gene Symbol
LY75-CD302
Entrez Gene ID
100526664
Gene Name
LY75-CD302 readthrough
Gene Aliases
-
UniGene
Hs.153563
Gene Chromosome Location
Chr.2: 159768628-159904756 on Build GRCh38
Transcripts
NM_002349.3 NM_001198759.1 NM_001198760.1

SNP Detail

SNP ID Chromosome Location
rs889157961 Chr.2: 159804447-159804447 on Build GRCh38
rs1019187198 Chr.2: 159804224-159804224 on Build GRCh38
rs920658920 Chr.2: 159804415-159804415 on Build GRCh38
rs143375040 Chr.2: 159804285-159804285 on Build GRCh38
rs903111517 Chr.2: 159804497-159804497 on Build GRCh38
rs570617629 Chr.2: 159804241-159804241 on Build GRCh38
rs752737672 Chr.2: 159804379-159804379 on Build GRCh38
rs1009227746 Chr.2: 159804455-159804455 on Build GRCh38
rs930721633 Chr.2: 159804650-159804650 on Build GRCh38
rs920792254 Chr.2: 159804624-159804624 on Build GRCh38
rs779874799 Chr.2: 159804641-159804641 on Build GRCh38
rs745651484 Chr.2: 159804307-159804307 on Build GRCh38
rs943346143 Chr.2: 159804446-159804446 on Build GRCh38
rs1016537903 Chr.2: 159804499-159804499 on Build GRCh38
rs998675534 Chr.2: 159804498-159804498 on Build GRCh38
rs981524694 Chr.2: 159804594-159804594 on Build GRCh38
rs957094407 Chr.2: 159804529-159804529 on Build GRCh38
rs977665415 Chr.2: 159804266-159804266 on Build GRCh38
rs112613414 Chr.2: 159804300-159804300 on Build GRCh38
rs930752789 Chr.2: 159804418-159804418 on Build GRCh38
rs554627358 Chr.2: 159804505-159804505 on Build GRCh38
rs910608411 Chr.2: 159804612-159804612 on Build GRCh38
rs755275924 Chr.2: 159804655-159804655 on Build GRCh38
rs1053714512 Chr.2: 159804239-159804239 on Build GRCh38
rs1012703577 Chr.2: 159804535-159804535 on Build GRCh38
rs4510173 Chr.2: 159804596-159804596 on Build GRCh38
rs1032733869 Chr.2: 159804281-159804281 on Build GRCh38
rs1022634404 Chr.2: 159804570-159804570 on Build GRCh38
rs1040256165 Chr.2: 159804472-159804472 on Build GRCh38
rs1008845464 Chr.2: 159804205-159804205 on Build GRCh38
rs536388269 Chr.2: 159804473-159804473 on Build GRCh38
rs567312684 Chr.2: 159804240-159804240 on Build GRCh38
rs919457839 Chr.2: 159804587-159804587 on Build GRCh38
rs972334430 Chr.2: 159804526-159804526 on Build GRCh38
rs983728981 Chr.2: 159804478-159804478 on Build GRCh38
rs940837355 Chr.2: 159804372-159804372 on Build GRCh38
rs962629750 Chr.2: 159804609-159804609 on Build GRCh38
rs1036503979 Chr.2: 159804412-159804412 on Build GRCh38
rs971047237 Chr.2: 159804591-159804591 on Build GRCh38
rs963997380 Chr.2: 159804503-159804503 on Build GRCh38
rs538920364 Chr.2: 159804371-159804371 on Build GRCh38
rs566139170 Chr.2: 159804413-159804413 on Build GRCh38
rs928211122 Chr.2: 159804313-159804313 on Build GRCh38
rs538228986 Chr.2: 159804286-159804286 on Build GRCh38
rs554346254 Chr.2: 159804373-159804373 on Build GRCh38
rs1050372707 Chr.2: 159804444-159804444 on Build GRCh38
rs972190112 Chr.2: 159804616-159804616 on Build GRCh38
rs970035196 Chr.2: 159804306-159804306 on Build GRCh38

View Full Product Details