Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 178124008-178124506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM45
Entrez Gene ID
129831
Gene Name
RNA binding motif protein 45
Gene Aliases
DRB1, RB-1
UniGene
Hs.377257
Gene Chromosome Location
Chr.2: 178112409-178129656 on Build GRCh38
Transcripts
NM_152945.3 XM_005246287.4 XM_017003320.1 XM_017003321.1 XM_017003322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6354807 c.1129T>C p.S377P Chr.2: 178124187-178124187 on Build GRCh38
4765496 c.1135_1136insA p.A382fs*4 Chr.2: 178124193-178124194 on Build GRCh38
717583 c.1128A>G p.P376P Chr.2: 178124186-178124186 on Build GRCh38
717582 c.1160C>A p.P387H Chr.2: 178124218-178124218 on Build GRCh38
1482216 c.1102C>G p.Q368E Chr.2: 178124160-178124160 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987960307 Chr.2: 178124204-178124204 on Build GRCh38
rs780371794 Chr.2: 178124247-178124247 on Build GRCh38
rs556541850 Chr.2: 178124440-178124440 on Build GRCh38
rs755827939 Chr.2: 178124199-178124199 on Build GRCh38
rs773489040 Chr.2: 178124282-178124282 on Build GRCh38
rs751549526 Chr.2: 178124177-178124177 on Build GRCh38
rs771047013 Chr.2: 178124193-178124193 on Build GRCh38
rs940370889 Chr.2: 178124079-178124079 on Build GRCh38
rs768742152 Chr.2: 178124110-178124110 on Build GRCh38
rs759522256 Chr.2: 178124310-178124311 on Build GRCh38
rs547932892 Chr.2: 178124054-178124054 on Build GRCh38
rs144362886 Chr.2: 178124074-178124074 on Build GRCh38
rs746164355 Chr.2: 178124219-178124219 on Build GRCh38
rs958260762 Chr.2: 178124333-178124333 on Build GRCh38
rs777542667 Chr.2: 178124202-178124202 on Build GRCh38
rs752243855 Chr.2: 178124317-178124317 on Build GRCh38
rs776738237 Chr.2: 178124111-178124111 on Build GRCh38
rs112984792 Chr.2: 178124082-178124082 on Build GRCh38
rs774336801 Chr.2: 178124049-178124049 on Build GRCh38
rs761990727 Chr.2: 178124127-178124127 on Build GRCh38
rs974890245 Chr.2: 178124358-178124358 on Build GRCh38
rs1048666474 Chr.2: 178124370-178124370 on Build GRCh38
rs746313820 Chr.2: 178124086-178124086 on Build GRCh38
rs745941279 Chr.2: 178124233-178124235 on Build GRCh38
rs772446807 Chr.2: 178124098-178124098 on Build GRCh38
rs772867292 Chr.2: 178124131-178124131 on Build GRCh38
rs139199245 Chr.2: 178124271-178124271 on Build GRCh38
rs956560479 Chr.2: 178124174-178124174 on Build GRCh38
rs940220120 Chr.2: 178124383-178124383 on Build GRCh38
rs975333902 Chr.2: 178124051-178124051 on Build GRCh38
rs759519558 Chr.2: 178124311-178124311 on Build GRCh38
rs933494258 Chr.2: 178124396-178124396 on Build GRCh38
rs369190430 Chr.2: 178124109-178124109 on Build GRCh38
rs908789310 Chr.2: 178124372-178124372 on Build GRCh38
rs553675752 Chr.2: 178124129-178124129 on Build GRCh38
rs774576367 Chr.2: 178124222-178124225 on Build GRCh38
rs767114774 Chr.2: 178124180-178124180 on Build GRCh38
rs776066762 Chr.2: 178124102-178124102 on Build GRCh38
rs766163957 Chr.2: 178124175-178124175 on Build GRCh38
rs868144892 Chr.2: 178124113-178124113 on Build GRCh38
rs373228142 Chr.2: 178124226-178124226 on Build GRCh38
rs1020213510 Chr.2: 178124304-178124304 on Build GRCh38
rs376552238 Chr.2: 178124289-178124289 on Build GRCh38
rs567927198 Chr.2: 178124075-178124075 on Build GRCh38
rs745958092 Chr.2: 178124067-178124067 on Build GRCh38
rs201379634 Chr.2: 178124178-178124178 on Build GRCh38
rs749563266 Chr.2: 178124192-178124193 on Build GRCh38
rs901270462 Chr.2: 178124463-178124463 on Build GRCh38
rs747488007 Chr.2: 178124264-178124264 on Build GRCh38
rs557463661 Chr.2: 178124041-178124041 on Build GRCh38
rs762818888 Chr.2: 178124142-178124142 on Build GRCh38
rs539318102 Chr.2: 178124423-178124423 on Build GRCh38
rs1041198642 Chr.2: 178124438-178124438 on Build GRCh38
rs752298945 Chr.2: 178124190-178124190 on Build GRCh38
rs539666856 Chr.2: 178124305-178124305 on Build GRCh38
rs548948475 Chr.2: 178124299-178124300 on Build GRCh38
rs762618263 Chr.2: 178124288-178124288 on Build GRCh38
rs898931602 Chr.2: 178124137-178124137 on Build GRCh38
rs772239481 Chr.2: 178124256-178124257 on Build GRCh38
rs762530036 Chr.2: 178124315-178124315 on Build GRCh38
rs1032320635 Chr.2: 178124331-178124331 on Build GRCh38
rs199858309 Chr.2: 178124270-178124270 on Build GRCh38
rs769848281 Chr.2: 178124280-178124280 on Build GRCh38
rs184953512 Chr.2: 178124078-178124078 on Build GRCh38
rs370369491 Chr.2: 178124303-178124303 on Build GRCh38

View Full Product Details