Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 181975126-181975640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R1C
Entrez Gene ID
151242
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 1C
Gene Aliases
IPP5
UniGene
Hs.10941
Gene Chromosome Location
Chr.2: 181954241-182131382 on Build GRCh38
Transcripts
NR_048567.1

SNP Detail

SNP ID Chromosome Location
rs897619438 Chr.2: 181975430-181975430 on Build GRCh38
rs1014807901 Chr.2: 181975408-181975408 on Build GRCh38
rs911261399 Chr.2: 181975164-181975164 on Build GRCh38
rs955478403 Chr.2: 181975512-181975512 on Build GRCh38
rs1045857099 Chr.2: 181975252-181975252 on Build GRCh38
rs549684496 Chr.2: 181975448-181975448 on Build GRCh38
rs1026148501 Chr.2: 181975216-181975216 on Build GRCh38
rs34940050 Chr.2: 181975226-181975226 on Build GRCh38
rs186704206 Chr.2: 181975447-181975447 on Build GRCh38
rs979288475 Chr.2: 181975377-181975382 on Build GRCh38
rs906951992 Chr.2: 181975307-181975307 on Build GRCh38
rs987863579 Chr.2: 181975405-181975405 on Build GRCh38
rs963865254 Chr.2: 181975310-181975310 on Build GRCh38
rs878954135 Chr.2: 181975463-181975463 on Build GRCh38
rs563295503 Chr.2: 181975596-181975596 on Build GRCh38
rs1012418121 Chr.2: 181975222-181975222 on Build GRCh38
rs575690222 Chr.2: 181975273-181975273 on Build GRCh38
rs747410402 Chr.2: 181975356-181975356 on Build GRCh38
rs913377849 Chr.2: 181975238-181975238 on Build GRCh38
rs937523743 Chr.2: 181975546-181975546 on Build GRCh38
rs1035099319 Chr.2: 181975187-181975187 on Build GRCh38
rs774783188 Chr.2: 181975459-181975471 on Build GRCh38
rs898790647 Chr.2: 181975599-181975599 on Build GRCh38
rs561287282 Chr.2: 181975556-181975556 on Build GRCh38
rs946246981 Chr.2: 181975247-181975247 on Build GRCh38
rs1006076566 Chr.2: 181975397-181975397 on Build GRCh38
rs985573458 Chr.2: 181975520-181975520 on Build GRCh38
rs545165691 Chr.2: 181975200-181975200 on Build GRCh38
rs980931228 Chr.2: 181975460-181975463 on Build GRCh38
rs1008736739 Chr.2: 181975232-181975232 on Build GRCh38
rs895216274 Chr.2: 181975191-181975191 on Build GRCh38
rs373754472 Chr.2: 181975358-181975358 on Build GRCh38
rs937124960 Chr.2: 181975324-181975324 on Build GRCh38
rs1055379889 Chr.2: 181975361-181975361 on Build GRCh38
rs56201324 Chr.2: 181975387-181975387 on Build GRCh38
rs990426687 Chr.2: 181975203-181975203 on Build GRCh38
rs77148218 Chr.2: 181975197-181975197 on Build GRCh38
rs898487210 Chr.2: 181975165-181975165 on Build GRCh38
rs1032248823 Chr.2: 181975383-181975383 on Build GRCh38
rs547279884 Chr.2: 181975601-181975601 on Build GRCh38
rs112558560 Chr.2: 181975476-181975476 on Build GRCh38

View Full Product Details