Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200467198-200467698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATS2L
Entrez Gene ID
26010
Gene Name
spermatogenesis associated serine rich 2 like
Gene Aliases
DNAPTP6, SGNP
UniGene
Hs.120323
Gene Chromosome Location
Chr.2: 200305881-200482263 on Build GRCh38
Transcripts
NM_001100422.1 NM_001100423.1 NM_001100424.1 NM_001282735.1 NM_001282743.1 NM_001282744.1 NM_015535.2 XM_005246456.1 XM_005246457.1 XM_005246458.1 XM_005246459.1 XM_011510938.1 XM_011510939.1 XM_017003783.1 XM_017003784.1 XM_017003785.1 XM_017003786.1 XM_017003787.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4090070 c.1036G>A p.A346T Chr.2: 200467388-200467388 on Build GRCh38
3407467 c.856C>T p.L286L Chr.2: 200467298-200467298 on Build GRCh38
4399498 c.1006A>G p.M336V Chr.2: 200467358-200467358 on Build GRCh38
6882120 c.863C>T p.A288V Chr.2: 200467305-200467305 on Build GRCh38
7143428 c.897C>A p.L299L Chr.2: 200467339-200467339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371126605 Chr.2: 200467277-200467277 on Build GRCh38
rs767903768 Chr.2: 200467259-200467259 on Build GRCh38
rs529807905 Chr.2: 200467451-200467451 on Build GRCh38
rs370788582 Chr.2: 200467540-200467540 on Build GRCh38
rs772962639 Chr.2: 200467544-200467544 on Build GRCh38
rs748395189 Chr.2: 200467429-200467429 on Build GRCh38
rs749118359 Chr.2: 200467414-200467414 on Build GRCh38
rs1050058589 Chr.2: 200467563-200467563 on Build GRCh38
rs925723355 Chr.2: 200467518-200467518 on Build GRCh38
rs373528852 Chr.2: 200467244-200467244 on Build GRCh38
rs537882026 Chr.2: 200467658-200467658 on Build GRCh38
rs751287230 Chr.2: 200467339-200467339 on Build GRCh38
rs920513701 Chr.2: 200467433-200467433 on Build GRCh38
rs570458979 Chr.2: 200467652-200467652 on Build GRCh38
rs950084206 Chr.2: 200467521-200467521 on Build GRCh38
rs910807459 Chr.2: 200467627-200467627 on Build GRCh38
rs768822901 Chr.2: 200467252-200467252 on Build GRCh38
rs868490503 Chr.2: 200467350-200467350 on Build GRCh38
rs1014982092 Chr.2: 200467348-200467348 on Build GRCh38
rs749945400 Chr.2: 200467276-200467276 on Build GRCh38
rs770782571 Chr.2: 200467647-200467647 on Build GRCh38
rs772262549 Chr.2: 200467431-200467431 on Build GRCh38
rs774259053 Chr.2: 200467427-200467427 on Build GRCh38
rs781180903 Chr.2: 200467351-200467351 on Build GRCh38
rs375135835 Chr.2: 200467378-200467378 on Build GRCh38
rs757912810 Chr.2: 200467330-200467330 on Build GRCh38
rs768789972 Chr.2: 200467422-200467422 on Build GRCh38
rs548367116 Chr.2: 200467490-200467490 on Build GRCh38
rs958565042 Chr.2: 200467457-200467457 on Build GRCh38
rs778340247 Chr.2: 200467304-200467304 on Build GRCh38
rs777089153 Chr.2: 200467337-200467337 on Build GRCh38
rs935668630 Chr.2: 200467525-200467525 on Build GRCh38
rs936008992 Chr.2: 200467574-200467574 on Build GRCh38
rs552533138 Chr.2: 200467638-200467638 on Build GRCh38
rs774651807 Chr.2: 200467254-200467254 on Build GRCh38
rs973692701 Chr.2: 200467424-200467424 on Build GRCh38
rs762738067 Chr.2: 200467575-200467575 on Build GRCh38
rs76313836 Chr.2: 200467556-200467556 on Build GRCh38
rs944917087 Chr.2: 200467643-200467643 on Build GRCh38
rs763086707 Chr.2: 200467251-200467251 on Build GRCh38
rs761960052 Chr.2: 200467255-200467255 on Build GRCh38
rs769788385 Chr.2: 200467243-200467243 on Build GRCh38
rs769684155 Chr.2: 200467402-200467402 on Build GRCh38
rs765666270 Chr.2: 200467340-200467340 on Build GRCh38
rs368507049 Chr.2: 200467407-200467407 on Build GRCh38
rs954450926 Chr.2: 200467302-200467302 on Build GRCh38
rs566625381 Chr.2: 200467516-200467516 on Build GRCh38
rs750739522 Chr.2: 200467262-200467262 on Build GRCh38
rs1041736282 Chr.2: 200467535-200467535 on Build GRCh38
rs900076746 Chr.2: 200467598-200467598 on Build GRCh38
rs766880246 Chr.2: 200467287-200467287 on Build GRCh38

View Full Product Details