Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 216043793-216044312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PECR
Entrez Gene ID
55825
Gene Name
peroxisomal trans-2-enoyl-CoA reductase
Gene Aliases
DCRRP, HPDHASE, HSA250303, PVIARL, SDR29C1, TERP
UniGene
Hs.281680
Gene Chromosome Location
Chr.2: 216029088-216081823 on Build GRCh38
Transcripts
NM_018441.5 XM_011511478.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
149060 c.774G>A p.S258S Chr.2: 216043956-216043956 on Build GRCh38
476889 c.761T>C p.I254T Chr.2: 216043969-216043969 on Build GRCh38
1614338 c.826+4A>T p.? Chr.2: 216043900-216043900 on Build GRCh38
2717523 c.812C>T p.S271L Chr.2: 216043918-216043918 on Build GRCh38
6743842 c.778G>T p.D260Y Chr.2: 216043952-216043952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773331067 Chr.2: 216043936-216043936 on Build GRCh38
rs935502809 Chr.2: 216043851-216043851 on Build GRCh38
rs773276921 Chr.2: 216044035-216044035 on Build GRCh38
rs779542206 Chr.2: 216044023-216044023 on Build GRCh38
rs767838655 Chr.2: 216043859-216043859 on Build GRCh38
rs753708257 Chr.2: 216043866-216043866 on Build GRCh38
rs774925577 Chr.2: 216044159-216044159 on Build GRCh38
rs750071185 Chr.2: 216043989-216043989 on Build GRCh38
rs150385654 Chr.2: 216043907-216043907 on Build GRCh38
rs111932305 Chr.2: 216044062-216044062 on Build GRCh38
rs749554065 Chr.2: 216044112-216044112 on Build GRCh38
rs915476465 Chr.2: 216044071-216044071 on Build GRCh38
rs954507188 Chr.2: 216043821-216043824 on Build GRCh38
rs939374340 Chr.2: 216044145-216044145 on Build GRCh38
rs766998977 Chr.2: 216043987-216043987 on Build GRCh38
rs762137328 Chr.2: 216043857-216043857 on Build GRCh38
rs773214437 Chr.2: 216044287-216044287 on Build GRCh38
rs757671221 Chr.2: 216043888-216043888 on Build GRCh38
rs954537708 Chr.2: 216044038-216044038 on Build GRCh38
rs186297072 Chr.2: 216044199-216044199 on Build GRCh38
rs768809172 Chr.2: 216044164-216044164 on Build GRCh38
rs137883002 Chr.2: 216043934-216043934 on Build GRCh38
rs930493916 Chr.2: 216044250-216044250 on Build GRCh38
rs981167967 Chr.2: 216044105-216044105 on Build GRCh38
rs757816694 Chr.2: 216043926-216043927 on Build GRCh38
rs764017941 Chr.2: 216043967-216043967 on Build GRCh38
rs374888148 Chr.2: 216043924-216043924 on Build GRCh38
rs899062537 Chr.2: 216044211-216044211 on Build GRCh38
rs758268817 Chr.2: 216043905-216043905 on Build GRCh38
rs139977790 Chr.2: 216043994-216043994 on Build GRCh38
rs762938738 Chr.2: 216043957-216043957 on Build GRCh38
rs888912579 Chr.2: 216044266-216044266 on Build GRCh38
rs772075274 Chr.2: 216044034-216044034 on Build GRCh38
rs927959218 Chr.2: 216044106-216044106 on Build GRCh38
rs190770435 Chr.2: 216043937-216043937 on Build GRCh38
rs1028685376 Chr.2: 216044036-216044036 on Build GRCh38
rs566885499 Chr.2: 216044248-216044248 on Build GRCh38
rs142404140 Chr.2: 216044073-216044073 on Build GRCh38
rs943075578 Chr.2: 216044285-216044285 on Build GRCh38
rs372529683 Chr.2: 216043917-216043917 on Build GRCh38
rs746208719 Chr.2: 216044072-216044072 on Build GRCh38
rs901485306 Chr.2: 216044288-216044288 on Build GRCh38
rs546380026 Chr.2: 216044243-216044243 on Build GRCh38
rs755608449 Chr.2: 216044020-216044020 on Build GRCh38
rs915502367 Chr.2: 216043858-216043858 on Build GRCh38
rs115170819 Chr.2: 216044055-216044055 on Build GRCh38
rs181515885 Chr.2: 216044163-216044163 on Build GRCh38
rs747212508 Chr.2: 216044040-216044040 on Build GRCh38
rs755724496 Chr.2: 216043992-216043992 on Build GRCh38
rs147938259 Chr.2: 216043929-216043929 on Build GRCh38
rs1037539217 Chr.2: 216044204-216044204 on Build GRCh38
rs2303842 Chr.2: 216043956-216043956 on Build GRCh38
rs780360950 Chr.2: 216044085-216044085 on Build GRCh38
rs749655382 Chr.2: 216044000-216044000 on Build GRCh38
rs533394186 Chr.2: 216043927-216043927 on Build GRCh38
rs996076627 Chr.2: 216044220-216044220 on Build GRCh38
rs751105875 Chr.2: 216043977-216043977 on Build GRCh38
rs764947491 Chr.2: 216043951-216043951 on Build GRCh38
rs987623706 Chr.2: 216043821-216043821 on Build GRCh38
rs748619069 Chr.2: 216044031-216044031 on Build GRCh38
rs1991533 Chr.2: 216044187-216044187 on Build GRCh38
rs751058679 Chr.2: 216043915-216043915 on Build GRCh38
rs924071199 Chr.2: 216043828-216043828 on Build GRCh38
rs939262899 Chr.2: 216044202-216044202 on Build GRCh38
rs372956838 Chr.2: 216043981-216043981 on Build GRCh38
rs562985451 Chr.2: 216043922-216043922 on Build GRCh38
rs771176000 Chr.2: 216044050-216044050 on Build GRCh38
rs1049362326 Chr.2: 216044257-216044257 on Build GRCh38
rs762709210 Chr.2: 216044061-216044061 on Build GRCh38
rs971952560 Chr.2: 216044228-216044228 on Build GRCh38
rs369272870 Chr.2: 216043930-216043930 on Build GRCh38
rs956794168 Chr.2: 216043830-216043830 on Build GRCh38
rs538519057 Chr.2: 216044135-216044135 on Build GRCh38
rs1022406371 Chr.2: 216044137-216044137 on Build GRCh38
rs890087075 Chr.2: 216044280-216044280 on Build GRCh38
rs769228139 Chr.2: 216043925-216043925 on Build GRCh38
rs759230810 Chr.2: 216043874-216043874 on Build GRCh38
rs772568764 Chr.2: 216044041-216044041 on Build GRCh38
rs2163015 Chr.2: 216044200-216044200 on Build GRCh38
rs149798403 Chr.2: 216043918-216043918 on Build GRCh38
rs368826601 Chr.2: 216043879-216043879 on Build GRCh38
rs200792059 Chr.2: 216043960-216043960 on Build GRCh38
rs989491863 Chr.2: 216043834-216043834 on Build GRCh38

View Full Product Details