Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232542341-232542850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNG
Entrez Gene ID
1146
Gene Name
cholinergic receptor nicotinic gamma subunit
Gene Aliases
ACHRG
UniGene
Hs.248101
Gene Chromosome Location
Chr.2: 232539727-232546328 on Build GRCh38
Transcripts
NM_005199.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
720235 c.571G>A p.E191K Chr.2: 232542487-232542487 on Build GRCh38
4552237 c.553G>A p.E185K Chr.2: 232542469-232542469 on Build GRCh38
1406240 c.561C>T p.G187G Chr.2: 232542477-232542477 on Build GRCh38
5509885 c.538C>A p.L180M Chr.2: 232542454-232542454 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778912321 Chr.2: 232542466-232542466 on Build GRCh38
rs999942430 Chr.2: 232542565-232542565 on Build GRCh38
rs758978775 Chr.2: 232542541-232542541 on Build GRCh38
rs138653046 Chr.2: 232542509-232542509 on Build GRCh38
rs143063340 Chr.2: 232542709-232542709 on Build GRCh38
rs753280990 Chr.2: 232542473-232542473 on Build GRCh38
rs916939669 Chr.2: 232542616-232542616 on Build GRCh38
rs776414812 Chr.2: 232542389-232542389 on Build GRCh38
rs775512922 Chr.2: 232542401-232542401 on Build GRCh38
rs989888612 Chr.2: 232542687-232542687 on Build GRCh38
rs766650311 Chr.2: 232542426-232542426 on Build GRCh38
rs895171760 Chr.2: 232542752-232542752 on Build GRCh38
rs779393244 Chr.2: 232542379-232542379 on Build GRCh38
rs765469759 Chr.2: 232542557-232542557 on Build GRCh38
rs189660100 Chr.2: 232542487-232542487 on Build GRCh38
rs980067806 Chr.2: 232542666-232542666 on Build GRCh38
rs561484280 Chr.2: 232542818-232542818 on Build GRCh38
rs907928384 Chr.2: 232542692-232542692 on Build GRCh38
rs773839306 Chr.2: 232542643-232542643 on Build GRCh38
rs930513815 Chr.2: 232542678-232542678 on Build GRCh38
rs975726404 Chr.2: 232542662-232542662 on Build GRCh38
rs771103168 Chr.2: 232542704-232542704 on Build GRCh38
rs988736576 Chr.2: 232542366-232542366 on Build GRCh38
rs6761667 Chr.2: 232542701-232542701 on Build GRCh38
rs1032794708 Chr.2: 232542589-232542589 on Build GRCh38
rs927127794 Chr.2: 232542667-232542667 on Build GRCh38
rs1048891113 Chr.2: 232542685-232542685 on Build GRCh38
rs1029560657 Chr.2: 232542539-232542539 on Build GRCh38
rs112102541 Chr.2: 232542695-232542697 on Build GRCh38
rs780996760 Chr.2: 232542500-232542500 on Build GRCh38
rs773411025 Chr.2: 232542387-232542387 on Build GRCh38
rs766193925 Chr.2: 232542449-232542449 on Build GRCh38
rs765868304 Chr.2: 232542433-232542436 on Build GRCh38
rs988843687 Chr.2: 232542612-232542612 on Build GRCh38
rs1024372201 Chr.2: 232542821-232542821 on Build GRCh38
rs201132844 Chr.2: 232542535-232542535 on Build GRCh38
rs747371344 Chr.2: 232542388-232542388 on Build GRCh38
rs192576889 Chr.2: 232542373-232542373 on Build GRCh38
rs764775180 Chr.2: 232542400-232542400 on Build GRCh38
rs1012392490 Chr.2: 232542553-232542553 on Build GRCh38
rs1003960580 Chr.2: 232542446-232542447 on Build GRCh38
rs748543001 Chr.2: 232542381-232542381 on Build GRCh38
rs905733612 Chr.2: 232542393-232542393 on Build GRCh38
rs2853462 Chr.2: 232542410-232542410 on Build GRCh38
rs565093373 Chr.2: 232542793-232542793 on Build GRCh38
rs755060349 Chr.2: 232542465-232542465 on Build GRCh38
rs901875177 Chr.2: 232542720-232542720 on Build GRCh38
rs965699821 Chr.2: 232542637-232542637 on Build GRCh38
rs1012323697 Chr.2: 232542774-232542774 on Build GRCh38
rs758872639 Chr.2: 232542486-232542486 on Build GRCh38
rs535274168 Chr.2: 232542608-232542608 on Build GRCh38
rs772506348 Chr.2: 232542384-232542384 on Build GRCh38
rs1031178915 Chr.2: 232542808-232542808 on Build GRCh38
rs774901684 Chr.2: 232542534-232542534 on Build GRCh38
rs774341652 Chr.2: 232542536-232542536 on Build GRCh38
rs184423691 Chr.2: 232542411-232542411 on Build GRCh38
rs761596188 Chr.2: 232542540-232542540 on Build GRCh38
rs562312580 Chr.2: 232542733-232542733 on Build GRCh38
rs752769617 Chr.2: 232542564-232542564 on Build GRCh38
rs983711786 Chr.2: 232542590-232542590 on Build GRCh38
rs2697782 Chr.2: 232542806-232542806 on Build GRCh38
rs780226198 Chr.2: 232542372-232542372 on Build GRCh38
rs376319807 Chr.2: 232542394-232542394 on Build GRCh38
rs145938094 Chr.2: 232542368-232542368 on Build GRCh38
rs1027773442 Chr.2: 232542791-232542791 on Build GRCh38
rs762753205 Chr.2: 232542404-232542404 on Build GRCh38
rs772569741 Chr.2: 232542607-232542607 on Build GRCh38
rs753990044 Chr.2: 232542459-232542459 on Build GRCh38
rs764059549 Chr.2: 232542409-232542409 on Build GRCh38
rs996815009 Chr.2: 232542722-232542722 on Build GRCh38
rs763507518 Chr.2: 232542383-232542384 on Build GRCh38
rs773842433 Chr.2: 232542468-232542468 on Build GRCh38
rs201865286 Chr.2: 232542424-232542424 on Build GRCh38
rs879370376 Chr.2: 232542725-232542725 on Build GRCh38
rs138875718 Chr.2: 232542635-232542635 on Build GRCh38
rs755042001 Chr.2: 232542377-232542377 on Build GRCh38
rs769453177 Chr.2: 232542512-232542512 on Build GRCh38
rs771417982 Chr.2: 232542489-232542489 on Build GRCh38
rs754079475 Chr.2: 232542548-232542548 on Build GRCh38
rs767238885 Chr.2: 232542543-232542543 on Build GRCh38
rs368873210 Chr.2: 232542556-232542556 on Build GRCh38
rs955780881 Chr.2: 232542599-232542599 on Build GRCh38
rs955751639 Chr.2: 232542817-232542817 on Build GRCh38
rs914399838 Chr.2: 232542690-232542690 on Build GRCh38
rs576014872 Chr.2: 232542773-232542773 on Build GRCh38
rs1005462691 Chr.2: 232542806-232542806 on Build GRCh38
rs543051076 Chr.2: 232542374-232542374 on Build GRCh38

View Full Product Details