Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 237574949-237575458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB17
Entrez Gene ID
64284
Gene Name
RAB17, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.44278
Gene Chromosome Location
Chr.2: 237574322-237591126 on Build GRCh38
Transcripts
NM_022449.3 NR_033308.1 XM_006712689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
327507 c.599A>G p.N200S Chr.2: 237575059-237575059 on Build GRCh38
5502644 c.563G>T p.G188V Chr.2: 237575095-237575095 on Build GRCh38
5728139 c.608C>T p.P203L Chr.2: 237575050-237575050 on Build GRCh38
7216056 c.630C>T p.C210C Chr.2: 237575028-237575028 on Build GRCh38
1018388 c.537G>A p.E179E Chr.2: 237575121-237575121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775097225 Chr.2: 237575027-237575027 on Build GRCh38
rs967811925 Chr.2: 237575061-237575061 on Build GRCh38
rs755087792 Chr.2: 237575048-237575048 on Build GRCh38
rs777336889 Chr.2: 237575179-237575179 on Build GRCh38
rs983681704 Chr.2: 237575157-237575157 on Build GRCh38
rs778876923 Chr.2: 237575000-237575000 on Build GRCh38
rs749607650 Chr.2: 237575077-237575078 on Build GRCh38
rs773609004 Chr.2: 237575042-237575042 on Build GRCh38
rs751454231 Chr.2: 237575067-237575067 on Build GRCh38
rs780701778 Chr.2: 237575073-237575073 on Build GRCh38
rs957270992 Chr.2: 237575295-237575295 on Build GRCh38
rs566785323 Chr.2: 237575406-237575406 on Build GRCh38
rs776052242 Chr.2: 237575014-237575014 on Build GRCh38
rs928954955 Chr.2: 237575329-237575329 on Build GRCh38
rs111491353 Chr.2: 237575231-237575231 on Build GRCh38
rs998269623 Chr.2: 237575204-237575204 on Build GRCh38
rs750353942 Chr.2: 237575125-237575125 on Build GRCh38
rs771660639 Chr.2: 237575424-237575424 on Build GRCh38
rs765832427 Chr.2: 237575403-237575403 on Build GRCh38
rs199916925 Chr.2: 237575074-237575074 on Build GRCh38
rs372061703 Chr.2: 237575378-237575378 on Build GRCh38
rs112742374 Chr.2: 237575117-237575117 on Build GRCh38
rs777246989 Chr.2: 237575103-237575103 on Build GRCh38
rs138383903 Chr.2: 237575394-237575394 on Build GRCh38
rs574237585 Chr.2: 237575191-237575191 on Build GRCh38
rs751697747 Chr.2: 237575377-237575377 on Build GRCh38
rs370434872 Chr.2: 237575342-237575342 on Build GRCh38
rs550958511 Chr.2: 237575076-237575076 on Build GRCh38
rs907938573 Chr.2: 237574997-237574997 on Build GRCh38
rs1011906748 Chr.2: 237575325-237575325 on Build GRCh38
rs368761463 Chr.2: 237575049-237575049 on Build GRCh38
rs554595249 Chr.2: 237575182-237575182 on Build GRCh38
rs561367060 Chr.2: 237575320-237575320 on Build GRCh38
rs1004243366 Chr.2: 237575202-237575202 on Build GRCh38
rs779540815 Chr.2: 237575094-237575095 on Build GRCh38
rs984858415 Chr.2: 237575333-237575333 on Build GRCh38
rs530959758 Chr.2: 237575292-237575292 on Build GRCh38
rs763825689 Chr.2: 237575114-237575114 on Build GRCh38
rs946276473 Chr.2: 237575255-237575255 on Build GRCh38
rs182147656 Chr.2: 237575083-237575083 on Build GRCh38
rs148349183 Chr.2: 237575106-237575106 on Build GRCh38
rs1051941508 Chr.2: 237575032-237575032 on Build GRCh38
rs766512859 Chr.2: 237574987-237574987 on Build GRCh38
rs371209656 Chr.2: 237575069-237575069 on Build GRCh38
rs771618290 Chr.2: 237575090-237575090 on Build GRCh38
rs764424565 Chr.2: 237575372-237575372 on Build GRCh38
rs1032018525 Chr.2: 237575092-237575092 on Build GRCh38
rs200357322 Chr.2: 237575096-237575096 on Build GRCh38
rs760855976 Chr.2: 237575330-237575330 on Build GRCh38
rs1004762980 Chr.2: 237575340-237575340 on Build GRCh38
rs369098612 Chr.2: 237575012-237575012 on Build GRCh38
rs374905720 Chr.2: 237575105-237575105 on Build GRCh38
rs757711254 Chr.2: 237575418-237575418 on Build GRCh38
rs376729614 Chr.2: 237575407-237575407 on Build GRCh38
rs575419413 Chr.2: 237575122-237575122 on Build GRCh38
rs904654685 Chr.2: 237575299-237575299 on Build GRCh38
rs116394440 Chr.2: 237575430-237575430 on Build GRCh38
rs542845053 Chr.2: 237574985-237574985 on Build GRCh38
rs190873355 Chr.2: 237575203-237575203 on Build GRCh38
rs375159510 Chr.2: 237575046-237575046 on Build GRCh38
rs919002129 Chr.2: 237575321-237575321 on Build GRCh38
rs758182252 Chr.2: 237575009-237575009 on Build GRCh38
rs751318867 Chr.2: 237575116-237575116 on Build GRCh38
rs957157911 Chr.2: 237575079-237575079 on Build GRCh38
rs970616252 Chr.2: 237575132-237575132 on Build GRCh38
rs769259195 Chr.2: 237575082-237575082 on Build GRCh38
rs747788987 Chr.2: 237575416-237575416 on Build GRCh38
rs41270747 Chr.2: 237575425-237575425 on Build GRCh38
rs767284036 Chr.2: 237575382-237575382 on Build GRCh38
rs745846541 Chr.2: 237575211-237575211 on Build GRCh38
rs140150815 Chr.2: 237575357-237575357 on Build GRCh38
rs537396490 Chr.2: 237575145-237575145 on Build GRCh38
rs568969245 Chr.2: 237575431-237575431 on Build GRCh38
rs908923235 Chr.2: 237575355-237575355 on Build GRCh38
rs778411928 Chr.2: 237575159-237575159 on Build GRCh38
rs59945834 Chr.2: 237575028-237575028 on Build GRCh38
rs756008721 Chr.2: 237575400-237575400 on Build GRCh38
rs2280289 Chr.2: 237575086-237575086 on Build GRCh38
rs777565176 Chr.2: 237575065-237575065 on Build GRCh38
rs755214475 Chr.2: 237574989-237574989 on Build GRCh38
rs746347621 Chr.2: 237575377-237575377 on Build GRCh38
rs59607721 Chr.2: 237575258-237575258 on Build GRCh38
rs931965956 Chr.2: 237575214-237575214 on Build GRCh38
rs1047773446 Chr.2: 237574980-237574980 on Build GRCh38
rs771569398 Chr.2: 237575078-237575078 on Build GRCh38
rs775838317 Chr.2: 237575433-237575433 on Build GRCh38
rs559642549 Chr.2: 237575213-237575213 on Build GRCh38
rs745550540 Chr.2: 237575024-237575024 on Build GRCh38
rs763106085 Chr.2: 237575371-237575371 on Build GRCh38
rs34311889 Chr.2: 237575108-237575108 on Build GRCh38
rs747419341 Chr.2: 237575170-237575170 on Build GRCh38
rs991426433 Chr.2: 237575298-237575298 on Build GRCh38
rs888709176 Chr.2: 237575187-237575187 on Build GRCh38
rs747944104 Chr.2: 237575008-237575008 on Build GRCh38
rs755676860 Chr.2: 237575139-237575139 on Build GRCh38
rs35969188 Chr.2: 237575097-237575098 on Build GRCh38
rs754122038 Chr.2: 237574988-237574988 on Build GRCh38
rs200880823 Chr.2: 237575084-237575084 on Build GRCh38
rs778694655 Chr.2: 237575415-237575415 on Build GRCh38
rs768309703 Chr.2: 237575155-237575155 on Build GRCh38
rs777638509 Chr.2: 237575010-237575010 on Build GRCh38
rs1039847720 Chr.2: 237575196-237575196 on Build GRCh38
rs201004899 Chr.2: 237575047-237575047 on Build GRCh38
rs146107983 Chr.2: 237575307-237575307 on Build GRCh38
rs769998175 Chr.2: 237575349-237575349 on Build GRCh38
rs779553066 Chr.2: 237575140-237575140 on Build GRCh38
rs544000040 Chr.2: 237575237-237575237 on Build GRCh38
rs780814920 Chr.2: 237575150-237575150 on Build GRCh38
rs529923135 Chr.2: 237575332-237575332 on Build GRCh38
rs72990107 Chr.2: 237575011-237575011 on Build GRCh38
rs768277188 Chr.2: 237575306-237575306 on Build GRCh38
rs778306697 Chr.2: 237575042-237575046 on Build GRCh38
rs571426654 Chr.2: 237575034-237575034 on Build GRCh38
rs771418054 Chr.2: 237575171-237575171 on Build GRCh38

View Full Product Details