Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 67020278-67020782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35D1
Entrez Gene ID
23169
Gene Name
solute carrier family 35 member D1
Gene Aliases
UGTREL7
UniGene
Hs.213642
Gene Chromosome Location
Chr.1: 66999332-67054397 on Build GRCh38
Transcripts
NM_015139.2 XM_006710478.1 XM_011541070.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4652788 c.831G>A p.T277T Chr.1: 67020414-67020414 on Build GRCh38
464895 c.846T>G p.A282A Chr.1: 67020399-67020399 on Build GRCh38
5569661 c.831G>T p.T277T Chr.1: 67020414-67020414 on Build GRCh38
6781693 c.830C>T p.T277M Chr.1: 67020415-67020415 on Build GRCh38
1474167 c.843T>G p.S281S Chr.1: 67020402-67020402 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003051723 Chr.1: 67020301-67020301 on Build GRCh38
rs767721209 Chr.1: 67020417-67020417 on Build GRCh38
rs143745141 Chr.1: 67020394-67020394 on Build GRCh38
rs1007572201 Chr.1: 67020679-67020679 on Build GRCh38
rs143218310 Chr.1: 67020431-67020431 on Build GRCh38
rs751569615 Chr.1: 67020415-67020415 on Build GRCh38
rs746241766 Chr.1: 67020499-67020499 on Build GRCh38
rs752637008 Chr.1: 67020433-67020433 on Build GRCh38
rs1056928065 Chr.1: 67020586-67020586 on Build GRCh38
rs759969801 Chr.1: 67020385-67020385 on Build GRCh38
rs780937106 Chr.1: 67020483-67020483 on Build GRCh38
rs565764487 Chr.1: 67020670-67020670 on Build GRCh38
rs148053630 Chr.1: 67020448-67020448 on Build GRCh38
rs983323095 Chr.1: 67020487-67020487 on Build GRCh38
rs777206121 Chr.1: 67020366-67020366 on Build GRCh38
rs771540177 Chr.1: 67020331-67020331 on Build GRCh38
rs746517683 Chr.1: 67020480-67020480 on Build GRCh38
rs944390641 Chr.1: 67020502-67020502 on Build GRCh38
rs777387674 Chr.1: 67020465-67020465 on Build GRCh38
rs907244829 Chr.1: 67020677-67020677 on Build GRCh38
rs199585658 Chr.1: 67020409-67020409 on Build GRCh38
rs115267525 Chr.1: 67020711-67020711 on Build GRCh38
rs527254513 Chr.1: 67020482-67020482 on Build GRCh38
rs1054238583 Chr.1: 67020560-67020560 on Build GRCh38
rs772767038 Chr.1: 67020332-67020332 on Build GRCh38
rs1021925151 Chr.1: 67020386-67020386 on Build GRCh38
rs764185366 Chr.1: 67020414-67020414 on Build GRCh38
rs938434073 Chr.1: 67020580-67020580 on Build GRCh38
rs1010475909 Chr.1: 67020373-67020373 on Build GRCh38
rs576252869 Chr.1: 67020314-67020314 on Build GRCh38
rs374553751 Chr.1: 67020318-67020318 on Build GRCh38
rs569749517 Chr.1: 67020583-67020583 on Build GRCh38
rs1010444796 Chr.1: 67020632-67020632 on Build GRCh38
rs762061949 Chr.1: 67020416-67020416 on Build GRCh38
rs894769231 Chr.1: 67020717-67020717 on Build GRCh38
rs921885243 Chr.1: 67020520-67020520 on Build GRCh38
rs907589957 Chr.1: 67020470-67020470 on Build GRCh38
rs373086951 Chr.1: 67020351-67020351 on Build GRCh38
rs975736798 Chr.1: 67020515-67020515 on Build GRCh38
rs192760258 Chr.1: 67020530-67020530 on Build GRCh38
rs772926415 Chr.1: 67020303-67020304 on Build GRCh38
rs945870775 Chr.1: 67020634-67020634 on Build GRCh38
rs148294304 Chr.1: 67020572-67020572 on Build GRCh38
rs187184697 Chr.1: 67020739-67020739 on Build GRCh38
rs149117184 Chr.1: 67020429-67020429 on Build GRCh38
rs574226071 Chr.1: 67020666-67020666 on Build GRCh38
rs746800809 Chr.1: 67020469-67020469 on Build GRCh38
rs567381817 Chr.1: 67020537-67020537 on Build GRCh38
rs765679202 Chr.1: 67020388-67020388 on Build GRCh38
rs891967794 Chr.1: 67020620-67020620 on Build GRCh38
rs764866579 Chr.1: 67020542-67020542 on Build GRCh38
rs904734796 Chr.1: 67020667-67020667 on Build GRCh38
rs543617570 Chr.1: 67020362-67020362 on Build GRCh38
rs775324564 Chr.1: 67020429-67020440 on Build GRCh38
rs552173187 Chr.1: 67020647-67020647 on Build GRCh38
rs372859338 Chr.1: 67020432-67020432 on Build GRCh38
rs538966946 Chr.1: 67020592-67020592 on Build GRCh38
rs941943076 Chr.1: 67020552-67020552 on Build GRCh38
rs201771401 Chr.1: 67020428-67020428 on Build GRCh38
rs184610399 Chr.1: 67020579-67020579 on Build GRCh38
rs909097885 Chr.1: 67020534-67020534 on Build GRCh38

View Full Product Details