Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 30491379-30491874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTAP19-3
Entrez Gene ID
337970
Gene Name
keratin associated protein 19-3
Gene Aliases
GTRHP, KAP19.3
UniGene
Hs.60480
Gene Chromosome Location
Chr.21: 30491464-30491957 on Build GRCh38
Transcripts
NM_181609.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
391848 c.161delG p.G54fs*>28 Chr.21: 30491797-30491797 on Build GRCh38
1029902 c.187G>A p.G63R Chr.21: 30491771-30491771 on Build GRCh38
4519620 c.101G>A p.R34K Chr.21: 30491857-30491857 on Build GRCh38
1029901 c.240C>A p.F80L Chr.21: 30491718-30491718 on Build GRCh38
4476447 c.206C>T p.P69L Chr.21: 30491752-30491752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773896504 Chr.21: 30491764-30491764 on Build GRCh38
rs531600798 Chr.21: 30491426-30491426 on Build GRCh38
rs755819166 Chr.21: 30491799-30491799 on Build GRCh38
rs908844068 Chr.21: 30491472-30491472 on Build GRCh38
rs2251071 Chr.21: 30491529-30491529 on Build GRCh38
rs774875471 Chr.21: 30491797-30491797 on Build GRCh38
rs1001379500 Chr.21: 30491629-30491629 on Build GRCh38
rs1048494 Chr.21: 30491760-30491760 on Build GRCh38
rs751648834 Chr.21: 30491788-30491788 on Build GRCh38
rs113286260 Chr.21: 30491804-30491804 on Build GRCh38
rs371846415 Chr.21: 30491417-30491417 on Build GRCh38
rs865951627 Chr.21: 30491828-30491828 on Build GRCh38
rs570680515 Chr.21: 30491553-30491553 on Build GRCh38
rs749197798 Chr.21: 30491807-30491807 on Build GRCh38
rs574891701 Chr.21: 30491660-30491660 on Build GRCh38
rs750978897 Chr.21: 30491734-30491734 on Build GRCh38
rs754209534 Chr.21: 30491723-30491723 on Build GRCh38
rs770688035 Chr.21: 30491444-30491444 on Build GRCh38
rs780313884 Chr.21: 30491747-30491747 on Build GRCh38
rs1042958885 Chr.21: 30491851-30491851 on Build GRCh38
rs891250953 Chr.21: 30491832-30491832 on Build GRCh38
rs373616472 Chr.21: 30491477-30491477 on Build GRCh38
rs543308267 Chr.21: 30491755-30491755 on Build GRCh38
rs765006869 Chr.21: 30491702-30491706 on Build GRCh38
rs923838456 Chr.21: 30491819-30491819 on Build GRCh38
rs78639009 Chr.21: 30491526-30491526 on Build GRCh38
rs1006931836 Chr.21: 30491836-30491836 on Build GRCh38
rs765596902 Chr.21: 30491733-30491733 on Build GRCh38
rs765890710 Chr.21: 30491690-30491690 on Build GRCh38
rs931674877 Chr.21: 30491437-30491437 on Build GRCh38
rs775212099 Chr.21: 30491771-30491771 on Build GRCh38
rs1036089123 Chr.21: 30491502-30491502 on Build GRCh38
rs796254168 Chr.21: 30491713-30491713 on Build GRCh38
rs968051324 Chr.21: 30491624-30491624 on Build GRCh38
rs1024067545 Chr.21: 30491579-30491579 on Build GRCh38
rs749248768 Chr.21: 30491759-30491759 on Build GRCh38
rs147132182 Chr.21: 30491751-30491751 on Build GRCh38
rs962646329 Chr.21: 30491646-30491646 on Build GRCh38
rs777631911 Chr.21: 30491756-30491756 on Build GRCh38
rs199527602 Chr.21: 30491781-30491781 on Build GRCh38
rs146459891 Chr.21: 30491814-30491815 on Build GRCh38
rs750918782 Chr.21: 30491775-30491775 on Build GRCh38
rs939052895 Chr.21: 30491427-30491427 on Build GRCh38
rs140298201 Chr.21: 30491772-30491772 on Build GRCh38
rs1029772685 Chr.21: 30491544-30491544 on Build GRCh38
rs759209612 Chr.21: 30491767-30491767 on Build GRCh38
rs757777704 Chr.21: 30491732-30491732 on Build GRCh38
rs990523029 Chr.21: 30491776-30491776 on Build GRCh38
rs199867409 Chr.21: 30491750-30491750 on Build GRCh38
rs191826883 Chr.21: 30491530-30491530 on Build GRCh38
rs1031070484 Chr.21: 30491644-30491644 on Build GRCh38
rs139146271 Chr.21: 30491609-30491609 on Build GRCh38
rs762885754 Chr.21: 30491782-30491808 on Build GRCh38
rs542205844 Chr.21: 30491702-30491702 on Build GRCh38
rs201803725 Chr.21: 30491855-30491855 on Build GRCh38
rs145653137 Chr.21: 30491780-30491780 on Build GRCh38
rs879244082 Chr.21: 30491487-30491487 on Build GRCh38
rs772708980 Chr.21: 30491666-30491666 on Build GRCh38
rs771686451 Chr.21: 30491770-30491770 on Build GRCh38
rs557508319 Chr.21: 30491731-30491731 on Build GRCh38
rs528825190 Chr.21: 30491796-30491796 on Build GRCh38
rs762387394 Chr.21: 30491674-30491674 on Build GRCh38
rs764493686 Chr.21: 30491705-30491705 on Build GRCh38
rs778386888 Chr.21: 30491826-30491826 on Build GRCh38
rs561996369 Chr.21: 30491835-30491835 on Build GRCh38
rs150799268 Chr.21: 30491569-30491569 on Build GRCh38
rs1018228130 Chr.21: 30491583-30491583 on Build GRCh38
rs759148318 Chr.21: 30491704-30491704 on Build GRCh38
rs781208235 Chr.21: 30491795-30491795 on Build GRCh38
rs796243575 Chr.21: 30491805-30491805 on Build GRCh38
rs1054781054 Chr.21: 30491460-30491460 on Build GRCh38
rs752702861 Chr.21: 30491711-30491712 on Build GRCh38
rs539185943 Chr.21: 30491735-30491735 on Build GRCh38
rs995722463 Chr.21: 30491679-30491679 on Build GRCh38
rs762432312 Chr.21: 30491485-30491485 on Build GRCh38
rs765158109 Chr.21: 30491410-30491410 on Build GRCh38
rs1006879718 Chr.21: 30491582-30491582 on Build GRCh38
rs755225201 Chr.21: 30491790-30491790 on Build GRCh38
rs1012749920 Chr.21: 30491571-30491572 on Build GRCh38
rs770911384 Chr.21: 30491825-30491825 on Build GRCh38
rs941789115 Chr.21: 30491481-30491481 on Build GRCh38

View Full Product Details