Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44135953-44136495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf33
Entrez Gene ID
8209
Gene Name
chromosome 21 open reading frame 33
Gene Aliases
ES1, GT335, HES1, KNPH, KNPI
UniGene
Hs.413482
Gene Chromosome Location
Chr.21: 44133612-44145723 on Build GRCh38
Transcripts
NM_001320383.1 NM_001320384.1 NM_004649.7 NM_198155.4 NR_135220.1 XM_017028479.1 XM_006723901.3 XM_006723902.3 XM_006723903.3 XM_006723904.3 XM_011546081.2 XM_017028514.1 XM_017028516.1 XM_017028517.1

Gene Symbol
LOC102724023
Entrez Gene ID
102724023
Gene Name
ES1 protein homolog, mitochondrial
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.21: 5097172-5128463 on Build GRCh38
Transcripts
NM_001320383.1 NM_001320384.1 NM_004649.7 NM_198155.4 NR_135220.1 XM_017028479.1 XM_006723901.3 XM_006723902.3 XM_006723903.3 XM_006723904.3 XM_011546081.2 XM_017028514.1 XM_017028516.1 XM_017028517.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1660180 c.238T>C p.F80L Chr.21: 44136103-44136103 on Build GRCh38
5858728 c.221G>A p.G74E Chr.21: 44136086-44136086 on Build GRCh38
7295620 c.281A>G p.K94R Chr.21: 44136146-44136146 on Build GRCh38
7286677 c.195-1G>T p.? Chr.21: 44136059-44136059 on Build GRCh38
69933 c.262C>A p.H88N Chr.21: 44136127-44136127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147740220 Chr.21: 44136129-44136129 on Build GRCh38
rs8129052 Chr.21: 44136275-44136275 on Build GRCh38
rs775923065 Chr.21: 44136115-44136115 on Build GRCh38
rs972920010 Chr.21: 44136428-44136428 on Build GRCh38
rs1031534414 Chr.21: 44136302-44136302 on Build GRCh38
rs376955628 Chr.21: 44136023-44136023 on Build GRCh38
rs1037292259 Chr.21: 44135999-44135999 on Build GRCh38
rs775786103 Chr.21: 44136044-44136044 on Build GRCh38
rs984379598 Chr.21: 44136464-44136464 on Build GRCh38
rs200282373 Chr.21: 44136053-44136053 on Build GRCh38
rs769034280 Chr.21: 44136183-44136183 on Build GRCh38
rs931285588 Chr.21: 44136242-44136242 on Build GRCh38
rs754383068 Chr.21: 44136071-44136071 on Build GRCh38
rs749884128 Chr.21: 44136110-44136110 on Build GRCh38
rs911184344 Chr.21: 44136474-44136474 on Build GRCh38
rs773834203 Chr.21: 44136197-44136197 on Build GRCh38
rs745963976 Chr.21: 44136096-44136096 on Build GRCh38
rs938889934 Chr.21: 44136224-44136224 on Build GRCh38
rs756259267 Chr.21: 44136178-44136178 on Build GRCh38
rs970132836 Chr.21: 44136355-44136355 on Build GRCh38
rs577986926 Chr.21: 44136160-44136160 on Build GRCh38
rs1017148677 Chr.21: 44136297-44136297 on Build GRCh38
rs753423959 Chr.21: 44136055-44136055 on Build GRCh38
rs763686914 Chr.21: 44136159-44136159 on Build GRCh38
rs386819105 Chr.21: 44136303-44136304 on Build GRCh38
rs770084517 Chr.21: 44136024-44136024 on Build GRCh38
rs763738263 Chr.21: 44136214-44136214 on Build GRCh38
rs138770901 Chr.21: 44136271-44136271 on Build GRCh38
rs866876783 Chr.21: 44136444-44136444 on Build GRCh38
rs1037279376 Chr.21: 44136226-44136226 on Build GRCh38
rs763237238 Chr.21: 44136125-44136125 on Build GRCh38
rs770922869 Chr.21: 44136087-44136087 on Build GRCh38
rs1048403005 Chr.21: 44136268-44136268 on Build GRCh38
rs373963980 Chr.21: 44136150-44136150 on Build GRCh38
rs540216943 Chr.21: 44136462-44136462 on Build GRCh38
rs563543219 Chr.21: 44136147-44136147 on Build GRCh38
rs768076428 Chr.21: 44136143-44136143 on Build GRCh38
rs764772419 Chr.21: 44136204-44136204 on Build GRCh38
rs752382275 Chr.21: 44136155-44136155 on Build GRCh38
rs1021214000 Chr.21: 44136335-44136335 on Build GRCh38
rs753403362 Chr.21: 44136293-44136293 on Build GRCh38
rs369169367 Chr.21: 44136054-44136054 on Build GRCh38
rs781200744 Chr.21: 44136166-44136166 on Build GRCh38
rs772308078 Chr.21: 44136193-44136193 on Build GRCh38
rs114623523 Chr.21: 44136282-44136282 on Build GRCh38
rs754911284 Chr.21: 44136206-44136225 on Build GRCh38
rs555217567 Chr.21: 44136402-44136402 on Build GRCh38
rs371193351 Chr.21: 44136182-44136182 on Build GRCh38
rs182331031 Chr.21: 44136114-44136114 on Build GRCh38
rs780258935 Chr.21: 44136181-44136181 on Build GRCh38
rs150157539 Chr.21: 44136231-44136231 on Build GRCh38
rs960221781 Chr.21: 44136379-44136379 on Build GRCh38
rs762048614 Chr.21: 44136142-44136142 on Build GRCh38
rs774652966 Chr.21: 44136184-44136184 on Build GRCh38
rs146296509 Chr.21: 44136378-44136378 on Build GRCh38
rs758019676 Chr.21: 44136156-44136156 on Build GRCh38
rs746996810 Chr.21: 44136080-44136080 on Build GRCh38
rs774882051 Chr.21: 44136133-44136133 on Build GRCh38
rs778637570 Chr.21: 44136077-44136077 on Build GRCh38
rs376878614 Chr.21: 44136325-44136325 on Build GRCh38
rs762409865 Chr.21: 44136213-44136213 on Build GRCh38
rs376869017 Chr.21: 44136165-44136165 on Build GRCh38
rs761051319 Chr.21: 44136203-44136203 on Build GRCh38
rs368518474 Chr.21: 44136016-44136016 on Build GRCh38
rs757143743 Chr.21: 44136217-44136217 on Build GRCh38
rs759837658 Chr.21: 44136032-44136032 on Build GRCh38
rs748562207 Chr.21: 44136186-44136186 on Build GRCh38
rs763471112 Chr.21: 44136045-44136045 on Build GRCh38
rs369482583 Chr.21: 44136367-44136367 on Build GRCh38
rs750575455 Chr.21: 44136176-44136177 on Build GRCh38
rs373126223 Chr.21: 44136216-44136216 on Build GRCh38
rs368912585 Chr.21: 44136148-44136148 on Build GRCh38
rs752331005 Chr.21: 44136078-44136078 on Build GRCh38
rs747943830 Chr.21: 44136009-44136009 on Build GRCh38
rs199582081 Chr.21: 44136088-44136088 on Build GRCh38
rs373399986 Chr.21: 44136097-44136097 on Build GRCh38
rs771794342 Chr.21: 44136013-44136013 on Build GRCh38
rs199985103 Chr.21: 44136174-44136174 on Build GRCh38
rs2071135 Chr.21: 44136304-44136304 on Build GRCh38
rs747006904 Chr.21: 44136445-44136445 on Build GRCh38
rs370398196 Chr.21: 44136218-44136218 on Build GRCh38
rs144227061 Chr.21: 44136079-44136079 on Build GRCh38
rs779910018 Chr.21: 44136373-44136373 on Build GRCh38
rs141880775 Chr.21: 44136303-44136303 on Build GRCh38
rs758195180 Chr.21: 44136370-44136370 on Build GRCh38
rs187124469 Chr.21: 44136280-44136280 on Build GRCh38
rs775002965 Chr.21: 44136206-44136206 on Build GRCh38
rs573228882 Chr.21: 44136411-44136411 on Build GRCh38
rs1006941943 Chr.21: 44136272-44136272 on Build GRCh38

View Full Product Details