Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44646923-44647460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTAP10-11
Entrez Gene ID
386678
Gene Name
keratin associated protein 10-11
Gene Aliases
KAP10.11, KAP18.11, KRTAP18-11, KRTAP18.11
UniGene
Hs.58076
Gene Chromosome Location
Chr.21: 44646414-44647650 on Build GRCh38
Transcripts
NM_198692.2 NM_001272037.1 NM_144991.2

Gene Symbol
TSPEAR
Entrez Gene ID
54084
Gene Name
thrombospondin type laminin G domain and EAR repeats
Gene Aliases
C21orf29, DFNB98, TSP-EAR
UniGene
Hs.660703
Gene Chromosome Location
Chr.21: 44497892-44711580 on Build GRCh38
Transcripts
NM_198692.2 NM_001272037.1 NM_144991.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3551514 c.794C>T p.S265F Chr.21: 44647252-44647252 on Build GRCh38
1031341 c.670G>A p.A224T Chr.21: 44647128-44647128 on Build GRCh38
4904359 c.795C>T p.S265S Chr.21: 44647253-44647253 on Build GRCh38
3740322 c.608G>A p.C203Y Chr.21: 44647066-44647066 on Build GRCh38
3758937 c.805C>T p.R269C Chr.21: 44647263-44647263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781873270 Chr.21: 44647246-44647246 on Build GRCh38
rs782763676 Chr.21: 44647109-44647109 on Build GRCh38
rs782424379 Chr.21: 44647266-44647266 on Build GRCh38
rs73909210 Chr.21: 44647293-44647293 on Build GRCh38
rs587773369 Chr.21: 44647128-44647128 on Build GRCh38
rs782452198 Chr.21: 44647252-44647252 on Build GRCh38
rs200236653 Chr.21: 44647183-44647183 on Build GRCh38
rs376523885 Chr.21: 44647217-44647217 on Build GRCh38
rs202058682 Chr.21: 44647077-44647077 on Build GRCh38
rs781918376 Chr.21: 44647277-44647277 on Build GRCh38
rs782767916 Chr.21: 44647294-44647294 on Build GRCh38
rs781861862 Chr.21: 44647130-44647130 on Build GRCh38
rs587759476 Chr.21: 44647113-44647113 on Build GRCh38
rs781829357 Chr.21: 44646999-44646999 on Build GRCh38
rs782340228 Chr.21: 44646960-44646960 on Build GRCh38
rs782689216 Chr.21: 44647407-44647407 on Build GRCh38
rs587745520 Chr.21: 44647005-44647005 on Build GRCh38
rs782765379 Chr.21: 44646998-44646998 on Build GRCh38
rs75548048 Chr.21: 44647187-44647187 on Build GRCh38
rs782147189 Chr.21: 44647349-44647349 on Build GRCh38
rs782542867 Chr.21: 44647301-44647302 on Build GRCh38
rs782230050 Chr.21: 44647260-44647260 on Build GRCh38
rs781808701 Chr.21: 44647140-44647140 on Build GRCh38
rs587612028 Chr.21: 44646972-44646972 on Build GRCh38
rs781811239 Chr.21: 44647297-44647297 on Build GRCh38
rs782272972 Chr.21: 44647046-44647046 on Build GRCh38
rs782307682 Chr.21: 44647031-44647031 on Build GRCh38
rs781930739 Chr.21: 44647171-44647171 on Build GRCh38
rs587598057 Chr.21: 44647009-44647009 on Build GRCh38
rs782062601 Chr.21: 44647337-44647337 on Build GRCh38
rs462007 Chr.21: 44647263-44647263 on Build GRCh38
rs782432857 Chr.21: 44647389-44647389 on Build GRCh38
rs781922188 Chr.21: 44647062-44647062 on Build GRCh38
rs782092658 Chr.21: 44647239-44647239 on Build GRCh38
rs782281354 Chr.21: 44647301-44647301 on Build GRCh38
rs782598704 Chr.21: 44647386-44647386 on Build GRCh38
rs587665055 Chr.21: 44647214-44647214 on Build GRCh38
rs375167760 Chr.21: 44647095-44647095 on Build GRCh38
rs782492646 Chr.21: 44647071-44647071 on Build GRCh38
rs782757565 Chr.21: 44647323-44647323 on Build GRCh38
rs73909208 Chr.21: 44647119-44647119 on Build GRCh38
rs368719173 Chr.21: 44647218-44647218 on Build GRCh38
rs1014766570 Chr.21: 44647155-44647155 on Build GRCh38
rs782792758 Chr.21: 44646987-44646987 on Build GRCh38
rs782409679 Chr.21: 44647318-44647318 on Build GRCh38
rs966613893 Chr.21: 44647342-44647342 on Build GRCh38
rs782605215 Chr.21: 44647026-44647026 on Build GRCh38
rs782690802 Chr.21: 44647299-44647299 on Build GRCh38
rs782544433 Chr.21: 44647011-44647011 on Build GRCh38
rs782180393 Chr.21: 44647102-44647102 on Build GRCh38
rs201197166 Chr.21: 44647051-44647051 on Build GRCh38
rs781956399 Chr.21: 44647295-44647295 on Build GRCh38
rs782009919 Chr.21: 44647321-44647321 on Build GRCh38
rs374050668 Chr.21: 44647289-44647289 on Build GRCh38
rs1035288148 Chr.21: 44647121-44647121 on Build GRCh38
rs782727146 Chr.21: 44647294-44647294 on Build GRCh38
rs587730867 Chr.21: 44647086-44647086 on Build GRCh38
rs782338819 Chr.21: 44647309-44647309 on Build GRCh38
rs782167809 Chr.21: 44647064-44647064 on Build GRCh38
rs587607760 Chr.21: 44647208-44647208 on Build GRCh38
rs201289628 Chr.21: 44647230-44647230 on Build GRCh38
rs782653309 Chr.21: 44647259-44647259 on Build GRCh38
rs960439483 Chr.21: 44646968-44646968 on Build GRCh38
rs782237768 Chr.21: 44647089-44647089 on Build GRCh38
rs200871348 Chr.21: 44647211-44647211 on Build GRCh38
rs782429833 Chr.21: 44647184-44647184 on Build GRCh38
rs782014495 Chr.21: 44647194-44647195 on Build GRCh38
rs782636477 Chr.21: 44647302-44647302 on Build GRCh38
rs782641768 Chr.21: 44647013-44647013 on Build GRCh38
rs782770425 Chr.21: 44647366-44647366 on Build GRCh38
rs782462123 Chr.21: 44647080-44647080 on Build GRCh38
rs782291466 Chr.21: 44647083-44647083 on Build GRCh38
rs782568970 Chr.21: 44647006-44647006 on Build GRCh38
rs78821735 Chr.21: 44647278-44647278 on Build GRCh38
rs782094756 Chr.21: 44646952-44646952 on Build GRCh38
rs781973105 Chr.21: 44647055-44647055 on Build GRCh38
rs782794414 Chr.21: 44647068-44647068 on Build GRCh38
rs781870524 Chr.21: 44647286-44647286 on Build GRCh38
rs199741257 Chr.21: 44647159-44647159 on Build GRCh38
rs782492153 Chr.21: 44647377-44647377 on Build GRCh38
rs782097255 Chr.21: 44647087-44647087 on Build GRCh38
rs782193170 Chr.21: 44647030-44647030 on Build GRCh38
rs376625605 Chr.21: 44647245-44647245 on Build GRCh38
rs587635293 Chr.21: 44647298-44647298 on Build GRCh38
rs77998434 Chr.21: 44647309-44647309 on Build GRCh38
rs781908750 Chr.21: 44647341-44647341 on Build GRCh38
rs782720110 Chr.21: 44647338-44647338 on Build GRCh38
rs782020061 Chr.21: 44647105-44647105 on Build GRCh38
rs782115620 Chr.21: 44646991-44646991 on Build GRCh38
rs973666864 Chr.21: 44647000-44647000 on Build GRCh38
rs200212284 Chr.21: 44647156-44647156 on Build GRCh38
rs782175405 Chr.21: 44647313-44647313 on Build GRCh38
rs781859439 Chr.21: 44647371-44647371 on Build GRCh38
rs782563110 Chr.21: 44647082-44647082 on Build GRCh38
rs781830839 Chr.21: 44647191-44647191 on Build GRCh38
rs587676741 Chr.21: 44647106-44647106 on Build GRCh38
rs375196067 Chr.21: 44647268-44647268 on Build GRCh38
rs781974731 Chr.21: 44647237-44647237 on Build GRCh38
rs781829320 Chr.21: 44647221-44647221 on Build GRCh38
rs782453810 Chr.21: 44647141-44647141 on Build GRCh38
rs782005700 Chr.21: 44647200-44647200 on Build GRCh38
rs781955765 Chr.21: 44647325-44647325 on Build GRCh38
rs587716089 Chr.21: 44647196-44647196 on Build GRCh38
rs782606646 Chr.21: 44647136-44647136 on Build GRCh38
rs202088711 Chr.21: 44647058-44647058 on Build GRCh38
rs782675510 Chr.21: 44647147-44647147 on Build GRCh38
rs782739563 Chr.21: 44647216-44647217 on Build GRCh38
rs200443384 Chr.21: 44647216-44647216 on Build GRCh38
rs782168233 Chr.21: 44647244-44647244 on Build GRCh38
rs148239295 Chr.21: 44647308-44647309 on Build GRCh38
rs782728144 Chr.21: 44647276-44647276 on Build GRCh38
rs199933627 Chr.21: 44647210-44647210 on Build GRCh38
rs781874539 Chr.21: 44647069-44647069 on Build GRCh38
rs782170677 Chr.21: 44647340-44647341 on Build GRCh38
rs782229665 Chr.21: 44647303-44647303 on Build GRCh38
rs587659517 Chr.21: 44646986-44646986 on Build GRCh38
rs587660459 Chr.21: 44647256-44647256 on Build GRCh38
rs782493673 Chr.21: 44647292-44647292 on Build GRCh38
rs782494084 Chr.21: 44647132-44647132 on Build GRCh38
rs781807354 Chr.21: 44647387-44647387 on Build GRCh38
rs587718659 Chr.21: 44647235-44647235 on Build GRCh38
rs201612439 Chr.21: 44647122-44647122 on Build GRCh38
rs782336066 Chr.21: 44647242-44647242 on Build GRCh38
rs200485359 Chr.21: 44646969-44646969 on Build GRCh38
rs782710016 Chr.21: 44647061-44647061 on Build GRCh38
rs781879038 Chr.21: 44646989-44646989 on Build GRCh38
rs782693773 Chr.21: 44647253-44647253 on Build GRCh38
rs782389438 Chr.21: 44647150-44647150 on Build GRCh38
rs782238984 Chr.21: 44647024-44647024 on Build GRCh38
rs1023024635 Chr.21: 44647163-44647163 on Build GRCh38
rs970047246 Chr.21: 44647164-44647164 on Build GRCh38
rs781911635 Chr.21: 44647298-44647299 on Build GRCh38
rs782532520 Chr.21: 44647258-44647258 on Build GRCh38
rs79740360 Chr.21: 44647251-44647251 on Build GRCh38
rs782701177 Chr.21: 44647201-44647201 on Build GRCh38
rs782182619 Chr.21: 44647076-44647076 on Build GRCh38
rs782315931 Chr.21: 44646960-44646961 on Build GRCh38
rs782001908 Chr.21: 44647046-44647046 on Build GRCh38
rs782750299 Chr.21: 44647188-44647188 on Build GRCh38
rs371766804 Chr.21: 44647204-44647204 on Build GRCh38
rs369444628 Chr.21: 44647270-44647270 on Build GRCh38
rs782736763 Chr.21: 44647430-44647430 on Build GRCh38
rs142705847 Chr.21: 44647250-44647250 on Build GRCh38
rs369120855 Chr.21: 44647381-44647381 on Build GRCh38
rs781932736 Chr.21: 44647312-44647312 on Build GRCh38
rs202157022 Chr.21: 44647215-44647215 on Build GRCh38
rs782076050 Chr.21: 44647275-44647275 on Build GRCh38
rs782788372 Chr.21: 44647283-44647283 on Build GRCh38
rs782528839 Chr.21: 44647084-44647084 on Build GRCh38
rs900191381 Chr.21: 44647094-44647094 on Build GRCh38
rs587675951 Chr.21: 44647025-44647025 on Build GRCh38
rs587763118 Chr.21: 44647166-44647166 on Build GRCh38
rs782027637 Chr.21: 44647044-44647044 on Build GRCh38

View Full Product Details