Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24224913-24225421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT5
Entrez Gene ID
2687
Gene Name
gamma-glutamyltransferase 5
Gene Aliases
GGL, GGT 5, GGT-REL, GGTLA1
UniGene
Hs.437156
Gene Chromosome Location
Chr.22: 24219654-24245142 on Build GRCh38
Transcripts
NM_001099781.2 NM_001099782.2 NM_001302464.1 NM_001302465.1 NM_004121.3 XM_005261557.2 XM_005261558.2 XM_011530133.1 XM_011530134.1 XM_011530135.1 XM_011530136.1 XM_011530137.2 XM_017028768.1 XM_017028769.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4399676 c.1466G>A p.G489E Chr.22: 24225282-24225282 on Build GRCh38
5057727 c.1446_1447insG p.S483fs*30 Chr.22: 24225301-24225302 on Build GRCh38
4164902 c.1430T>A p.I477N Chr.22: 24225318-24225318 on Build GRCh38
6891914 c.1410C>T p.S470S Chr.22: 24225338-24225338 on Build GRCh38
4164901 c.1431C>T p.I477I Chr.22: 24225317-24225317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777898121 Chr.22: 24225337-24225337 on Build GRCh38
rs1041111035 Chr.22: 24225304-24225304 on Build GRCh38
rs758005500 Chr.22: 24225106-24225106 on Build GRCh38
rs150898918 Chr.22: 24225284-24225284 on Build GRCh38
rs747658498 Chr.22: 24224978-24224978 on Build GRCh38
rs1049186603 Chr.22: 24225030-24225030 on Build GRCh38
rs779874745 Chr.22: 24225206-24225206 on Build GRCh38
rs765588510 Chr.22: 24225263-24225263 on Build GRCh38
rs528779081 Chr.22: 24225303-24225303 on Build GRCh38
rs139383582 Chr.22: 24225299-24225299 on Build GRCh38
rs755578145 Chr.22: 24225082-24225082 on Build GRCh38
rs746671447 Chr.22: 24225129-24225129 on Build GRCh38
rs767531469 Chr.22: 24225054-24225054 on Build GRCh38
rs374683064 Chr.22: 24224980-24224980 on Build GRCh38
rs778547176 Chr.22: 24224947-24224947 on Build GRCh38
rs745874439 Chr.22: 24225014-24225014 on Build GRCh38
rs774527296 Chr.22: 24225017-24225017 on Build GRCh38
rs755737041 Chr.22: 24225388-24225388 on Build GRCh38
rs777095964 Chr.22: 24225223-24225223 on Build GRCh38
rs777545484 Chr.22: 24224981-24224981 on Build GRCh38
rs200684960 Chr.22: 24225222-24225222 on Build GRCh38
rs752800899 Chr.22: 24225323-24225324 on Build GRCh38
rs542440435 Chr.22: 24225396-24225396 on Build GRCh38
rs894850599 Chr.22: 24225188-24225188 on Build GRCh38
rs866363144 Chr.22: 24225270-24225270 on Build GRCh38
rs769612704 Chr.22: 24225016-24225016 on Build GRCh38
rs754142074 Chr.22: 24225074-24225074 on Build GRCh38
rs745618291 Chr.22: 24225198-24225198 on Build GRCh38
rs765401891 Chr.22: 24225257-24225258 on Build GRCh38
rs761249346 Chr.22: 24225379-24225379 on Build GRCh38
rs747032399 Chr.22: 24225342-24225342 on Build GRCh38
rs1030793569 Chr.22: 24225274-24225274 on Build GRCh38
rs574777300 Chr.22: 24225101-24225101 on Build GRCh38
rs999205973 Chr.22: 24225265-24225265 on Build GRCh38
rs371500873 Chr.22: 24225066-24225066 on Build GRCh38
rs201616574 Chr.22: 24225219-24225219 on Build GRCh38
rs892550683 Chr.22: 24224948-24224948 on Build GRCh38
rs952661223 Chr.22: 24225022-24225022 on Build GRCh38
rs777378390 Chr.22: 24225119-24225119 on Build GRCh38
rs977879543 Chr.22: 24224955-24224955 on Build GRCh38
rs372732410 Chr.22: 24225006-24225006 on Build GRCh38
rs776743050 Chr.22: 24225350-24225350 on Build GRCh38
rs1036547052 Chr.22: 24225092-24225092 on Build GRCh38
rs759800101 Chr.22: 24225234-24225234 on Build GRCh38
rs182867617 Chr.22: 24225132-24225132 on Build GRCh38
rs751162471 Chr.22: 24225377-24225377 on Build GRCh38
rs771235441 Chr.22: 24225346-24225346 on Build GRCh38
rs749181027 Chr.22: 24225212-24225212 on Build GRCh38
rs1056441583 Chr.22: 24225162-24225162 on Build GRCh38
rs781106041 Chr.22: 24225135-24225135 on Build GRCh38
rs892435899 Chr.22: 24225247-24225247 on Build GRCh38
rs985183445 Chr.22: 24225375-24225375 on Build GRCh38
rs1052458766 Chr.22: 24224943-24224943 on Build GRCh38
rs200146637 Chr.22: 24225356-24225356 on Build GRCh38
rs529621965 Chr.22: 24225359-24225359 on Build GRCh38
rs1026801373 Chr.22: 24225351-24225351 on Build GRCh38
rs369915142 Chr.22: 24225204-24225204 on Build GRCh38
rs764663086 Chr.22: 24225370-24225370 on Build GRCh38
rs112092021 Chr.22: 24225190-24225190 on Build GRCh38
rs199510644 Chr.22: 24225214-24225214 on Build GRCh38
rs767011018 Chr.22: 24225382-24225382 on Build GRCh38
rs776231440 Chr.22: 24225013-24225013 on Build GRCh38
rs376709868 Chr.22: 24225369-24225369 on Build GRCh38
rs573432117 Chr.22: 24224942-24224942 on Build GRCh38
rs755159959 Chr.22: 24225138-24225138 on Build GRCh38
rs765632350 Chr.22: 24225086-24225086 on Build GRCh38
rs564711431 Chr.22: 24225295-24225295 on Build GRCh38
rs773450659 Chr.22: 24225055-24225055 on Build GRCh38
rs768572532 Chr.22: 24225331-24225331 on Build GRCh38
rs760823413 Chr.22: 24225065-24225065 on Build GRCh38
rs763495821 Chr.22: 24225283-24225283 on Build GRCh38
rs553312804 Chr.22: 24224985-24224985 on Build GRCh38
rs750040366 Chr.22: 24225300-24225300 on Build GRCh38
rs923745322 Chr.22: 24224959-24224959 on Build GRCh38
rs899927211 Chr.22: 24225317-24225317 on Build GRCh38
rs1003134270 Chr.22: 24225159-24225159 on Build GRCh38
rs113295783 Chr.22: 24225243-24225243 on Build GRCh38
rs143616956 Chr.22: 24225383-24225383 on Build GRCh38
rs150033594 Chr.22: 24225302-24225302 on Build GRCh38
rs756973423 Chr.22: 24225296-24225296 on Build GRCh38
rs7288201 Chr.22: 24225325-24225325 on Build GRCh38
rs369769344 Chr.22: 24225124-24225124 on Build GRCh38
rs370492248 Chr.22: 24225275-24225275 on Build GRCh38
rs761855801 Chr.22: 24225027-24225027 on Build GRCh38
rs1008344955 Chr.22: 24224956-24224956 on Build GRCh38
rs140052476 Chr.22: 24225355-24225355 on Build GRCh38
rs375273797 Chr.22: 24225123-24225123 on Build GRCh38

View Full Product Details