Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24555513-24556006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUCD1
Entrez Gene ID
83606
Gene Name
guanylyl cyclase domain containing 1
Gene Aliases
C22orf13, LLN4
UniGene
Hs.9850
Gene Chromosome Location
Chr.22: 24540423-24555935 on Build GRCh38
Transcripts
NM_001284251.1 NM_001284252.1 NM_001284253.1 NR_104286.1 XM_005261761.1 NM_001278656.1 NM_004175.4 NR_103819.1

Gene Symbol
SNRPD3
Entrez Gene ID
6634
Gene Name
small nuclear ribonucleoprotein D3 polypeptide
Gene Aliases
SMD3, Sm-D3
UniGene
Hs.356549
Gene Chromosome Location
Chr.22: 24555650-24574964 on Build GRCh38
Transcripts
NM_001284251.1 NM_001284252.1 NM_001284253.1 NR_104286.1 XM_005261761.1 NM_001278656.1 NM_004175.4 NR_103819.1

SNP Detail

SNP ID Chromosome Location
rs1045593323 Chr.22: 24555697-24555697 on Build GRCh38
rs918460482 Chr.22: 24555907-24555907 on Build GRCh38
rs1022939107 Chr.22: 24555785-24555785 on Build GRCh38
rs892897442 Chr.22: 24555988-24555988 on Build GRCh38
rs551696359 Chr.22: 24555865-24555865 on Build GRCh38
rs764659175 Chr.22: 24555807-24555807 on Build GRCh38
rs929916139 Chr.22: 24555914-24555914 on Build GRCh38
rs937106666 Chr.22: 24555708-24555708 on Build GRCh38
rs539241589 Chr.22: 24555861-24555861 on Build GRCh38
rs770429268 Chr.22: 24555664-24555664 on Build GRCh38
rs189468388 Chr.22: 24555948-24555948 on Build GRCh38
rs986814062 Chr.22: 24555852-24555852 on Build GRCh38
rs577035501 Chr.22: 24555935-24555935 on Build GRCh38
rs898942610 Chr.22: 24555678-24555678 on Build GRCh38
rs906916294 Chr.22: 24555699-24555699 on Build GRCh38
rs566602144 Chr.22: 24555896-24555896 on Build GRCh38
rs974120486 Chr.22: 24555905-24555905 on Build GRCh38
rs763429424 Chr.22: 24555788-24555788 on Build GRCh38
rs560611560 Chr.22: 24555545-24555545 on Build GRCh38
rs1055579288 Chr.22: 24555987-24555987 on Build GRCh38
rs988854366 Chr.22: 24555557-24555557 on Build GRCh38
rs561844745 Chr.22: 24555605-24555605 on Build GRCh38
rs929775218 Chr.22: 24555681-24555681 on Build GRCh38
rs930422975 Chr.22: 24555934-24555934 on Build GRCh38
rs1011142525 Chr.22: 24555779-24555779 on Build GRCh38
rs905780860 Chr.22: 24555815-24555815 on Build GRCh38
rs964273922 Chr.22: 24555904-24555904 on Build GRCh38
rs761426090 Chr.22: 24555817-24555817 on Build GRCh38
rs372146842 Chr.22: 24555928-24555928 on Build GRCh38
rs967307855 Chr.22: 24555552-24555552 on Build GRCh38
rs920067862 Chr.22: 24555930-24555930 on Build GRCh38
rs978304505 Chr.22: 24555574-24555574 on Build GRCh38
rs767090231 Chr.22: 24555821-24555821 on Build GRCh38
rs1029821141 Chr.22: 24555812-24555812 on Build GRCh38
rs930361266 Chr.22: 24555691-24555691 on Build GRCh38
rs377656284 Chr.22: 24555809-24555809 on Build GRCh38
rs144974630 Chr.22: 24555855-24555855 on Build GRCh38
rs757072755 Chr.22: 24555614-24555614 on Build GRCh38
rs747082658 Chr.22: 24555578-24555578 on Build GRCh38
rs367997605 Chr.22: 24555824-24555824 on Build GRCh38
rs1020292741 Chr.22: 24555832-24555832 on Build GRCh38
rs546233946 Chr.22: 24555589-24555589 on Build GRCh38
rs892815292 Chr.22: 24555735-24555735 on Build GRCh38
rs1010710518 Chr.22: 24555831-24555831 on Build GRCh38
rs955456274 Chr.22: 24555840-24555840 on Build GRCh38
rs374369179 Chr.22: 24555610-24555610 on Build GRCh38
rs750133832 Chr.22: 24555846-24555846 on Build GRCh38
rs949790039 Chr.22: 24555949-24555949 on Build GRCh38
rs1014176355 Chr.22: 24555799-24555799 on Build GRCh38
rs181265967 Chr.22: 24555977-24555977 on Build GRCh38
rs907615662 Chr.22: 24555941-24555941 on Build GRCh38
rs1032791612 Chr.22: 24555823-24555823 on Build GRCh38
rs755794985 Chr.22: 24555854-24555854 on Build GRCh38
rs114059315 Chr.22: 24555588-24555588 on Build GRCh38
rs759703475 Chr.22: 24555625-24555625 on Build GRCh38
rs181923376 Chr.22: 24555641-24555641 on Build GRCh38
rs184920490 Chr.22: 24555709-24555709 on Build GRCh38
rs886075121 Chr.22: 24555793-24555793 on Build GRCh38
rs1000070163 Chr.22: 24555808-24555808 on Build GRCh38
rs985420907 Chr.22: 24555605-24555605 on Build GRCh38
rs759712056 Chr.22: 24555618-24555618 on Build GRCh38
rs963005243 Chr.22: 24555897-24555897 on Build GRCh38
rs558188803 Chr.22: 24555562-24555562 on Build GRCh38
rs1038720777 Chr.22: 24555643-24555643 on Build GRCh38

View Full Product Details