Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93908959-93909449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCLM
Entrez Gene ID
2730
Gene Name
glutamate-cysteine ligase modifier subunit
Gene Aliases
GLCLR
UniGene
Hs.315562
Gene Chromosome Location
Chr.1: 93885199-93909598 on Build GRCh38
Transcripts
NM_001308253.1 NM_002061.3 XM_011541261.2 XM_017001056.1 XM_017001057.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7329644 c.113C>T p.T38M Chr.1: 93909051-93909051 on Build GRCh38
4944499 c.24C>G p.A8A Chr.1: 93909140-93909140 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776524476 Chr.1: 93909130-93909130 on Build GRCh38
rs752115103 Chr.1: 93909202-93909202 on Build GRCh38
rs781244231 Chr.1: 93909000-93909001 on Build GRCh38
rs2273407 Chr.1: 93908988-93908988 on Build GRCh38
rs2273406 Chr.1: 93909020-93909020 on Build GRCh38
rs17878517 Chr.1: 93909372-93909372 on Build GRCh38
rs895362414 Chr.1: 93909047-93909047 on Build GRCh38
rs948352652 Chr.1: 93909101-93909101 on Build GRCh38
rs1024281562 Chr.1: 93909050-93909050 on Build GRCh38
rs749080940 Chr.1: 93909053-93909053 on Build GRCh38
rs1021873879 Chr.1: 93909295-93909295 on Build GRCh38
rs924030349 Chr.1: 93909002-93909002 on Build GRCh38
rs992548316 Chr.1: 93909388-93909388 on Build GRCh38
rs1031756930 Chr.1: 93909148-93909148 on Build GRCh38
rs111406294 Chr.1: 93909147-93909147 on Build GRCh38
rs948303488 Chr.1: 93909392-93909392 on Build GRCh38
rs916613254 Chr.1: 93909390-93909390 on Build GRCh38
rs529996540 Chr.1: 93909254-93909254 on Build GRCh38
rs565331853 Chr.1: 93909069-93909069 on Build GRCh38
rs743119 Chr.1: 93909186-93909186 on Build GRCh38
rs548438884 Chr.1: 93909368-93909368 on Build GRCh38
rs767815533 Chr.1: 93909044-93909044 on Build GRCh38
rs747972711 Chr.1: 93909086-93909086 on Build GRCh38
rs17883607 Chr.1: 93909200-93909201 on Build GRCh38
rs1013604100 Chr.1: 93909024-93909024 on Build GRCh38
rs532594815 Chr.1: 93909115-93909115 on Build GRCh38
rs543559869 Chr.1: 93909059-93909059 on Build GRCh38
rs12401915 Chr.1: 93909179-93909179 on Build GRCh38
rs868831657 Chr.1: 93909325-93909325 on Build GRCh38
rs898759395 Chr.1: 93909415-93909415 on Build GRCh38
rs764495299 Chr.1: 93909201-93909201 on Build GRCh38
rs868785388 Chr.1: 93909398-93909398 on Build GRCh38
rs530693140 Chr.1: 93909401-93909401 on Build GRCh38
rs759853135 Chr.1: 93909136-93909136 on Build GRCh38
rs978186419 Chr.1: 93908997-93908997 on Build GRCh38
rs977233095 Chr.1: 93909343-93909343 on Build GRCh38
rs1038617261 Chr.1: 93909005-93909005 on Build GRCh38
rs778838905 Chr.1: 93909081-93909081 on Build GRCh38
rs541424485 Chr.1: 93909143-93909143 on Build GRCh38
rs967144397 Chr.1: 93909304-93909304 on Build GRCh38
rs879021892 Chr.1: 93909387-93909387 on Build GRCh38
rs1017963949 Chr.1: 93909222-93909222 on Build GRCh38
rs776063708 Chr.1: 93909164-93909164 on Build GRCh38
rs930102770 Chr.1: 93909417-93909417 on Build GRCh38
rs866646303 Chr.1: 93909373-93909373 on Build GRCh38
rs770111684 Chr.1: 93909138-93909138 on Build GRCh38
rs990116908 Chr.1: 93909272-93909272 on Build GRCh38
rs1007874959 Chr.1: 93909216-93909216 on Build GRCh38

View Full Product Details