Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37074530-37075038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS6
Entrez Gene ID
164656
Gene Name
transmembrane protease, serine 6
Gene Aliases
IRIDA
UniGene
Hs.370885
Gene Chromosome Location
Chr.22: 37065436-37109701 on Build GRCh38
Transcripts
NM_001289000.1 NM_001289001.1 NM_153609.3 XM_011529989.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3405648 c.1376G>A p.G459E Chr.22: 37074675-37074675 on Build GRCh38
1416107 c.1394C>A p.S465Y Chr.22: 37074684-37074684 on Build GRCh38
3554085 c.1387C>T p.L463F Chr.22: 37074691-37074691 on Build GRCh38
108022 c.1437C>T p.D479D Chr.22: 37074641-37074641 on Build GRCh38
323897 c.1417G>T p.A473S Chr.22: 37074661-37074661 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183182083 Chr.22: 37074583-37074583 on Build GRCh38
rs143732650 Chr.22: 37074625-37074625 on Build GRCh38
rs771281945 Chr.22: 37074588-37074588 on Build GRCh38
rs990517716 Chr.22: 37074952-37074952 on Build GRCh38
rs903236196 Chr.22: 37074772-37074772 on Build GRCh38
rs965546418 Chr.22: 37074862-37074862 on Build GRCh38
rs566696914 Chr.22: 37074775-37074775 on Build GRCh38
rs750412369 Chr.22: 37074608-37074608 on Build GRCh38
rs564708213 Chr.22: 37074833-37074833 on Build GRCh38
rs775819791 Chr.22: 37074789-37074789 on Build GRCh38
rs139616588 Chr.22: 37074811-37074811 on Build GRCh38
rs763728125 Chr.22: 37075001-37075004 on Build GRCh38
rs986085081 Chr.22: 37074969-37074969 on Build GRCh38
rs760761085 Chr.22: 37074739-37074739 on Build GRCh38
rs910433348 Chr.22: 37074982-37074982 on Build GRCh38
rs2072860 Chr.22: 37074564-37074564 on Build GRCh38
rs370678539 Chr.22: 37074571-37074571 on Build GRCh38
rs566684087 Chr.22: 37075000-37075000 on Build GRCh38
rs773168777 Chr.22: 37074855-37074855 on Build GRCh38
rs369540809 Chr.22: 37074758-37074758 on Build GRCh38
rs373512251 Chr.22: 37074891-37074892 on Build GRCh38
rs753536777 Chr.22: 37074620-37074620 on Build GRCh38
rs546574834 Chr.22: 37074611-37074611 on Build GRCh38
rs778717629 Chr.22: 37074631-37074631 on Build GRCh38
rs1023148037 Chr.22: 37074808-37074808 on Build GRCh38
rs1019820622 Chr.22: 37074845-37074845 on Build GRCh38
rs374912833 Chr.22: 37074832-37074832 on Build GRCh38
rs865982171 Chr.22: 37074696-37074696 on Build GRCh38
rs1010384071 Chr.22: 37074875-37074875 on Build GRCh38
rs1048916755 Chr.22: 37075011-37075011 on Build GRCh38
rs1015874603 Chr.22: 37074846-37074846 on Build GRCh38
rs889777261 Chr.22: 37075014-37075019 on Build GRCh38
rs777411575 Chr.22: 37074760-37074760 on Build GRCh38
rs1035884377 Chr.22: 37074787-37074787 on Build GRCh38
rs781531526 Chr.22: 37074653-37074653 on Build GRCh38
rs748331733 Chr.22: 37074560-37074562 on Build GRCh38
rs571221688 Chr.22: 37074786-37074786 on Build GRCh38
rs148119956 Chr.22: 37074676-37074676 on Build GRCh38
rs78640241 Chr.22: 37074598-37074598 on Build GRCh38
rs375137058 Chr.22: 37074745-37074745 on Build GRCh38
rs770196209 Chr.22: 37074594-37074594 on Build GRCh38
rs560740989 Chr.22: 37074794-37074794 on Build GRCh38
rs755949077 Chr.22: 37074610-37074610 on Build GRCh38
rs762043265 Chr.22: 37074804-37074805 on Build GRCh38
rs538294270 Chr.22: 37074910-37074910 on Build GRCh38
rs949240440 Chr.22: 37074996-37074996 on Build GRCh38
rs774666379 Chr.22: 37074717-37074717 on Build GRCh38
rs532005335 Chr.22: 37074866-37074866 on Build GRCh38
rs371707140 Chr.22: 37074729-37074729 on Build GRCh38
rs576812488 Chr.22: 37074882-37074883 on Build GRCh38
rs373885799 Chr.22: 37074587-37074587 on Build GRCh38
rs933276467 Chr.22: 37074744-37074744 on Build GRCh38
rs575076546 Chr.22: 37074759-37074759 on Build GRCh38
rs751536169 Chr.22: 37074599-37074599 on Build GRCh38
rs755409444 Chr.22: 37074553-37074553 on Build GRCh38
rs937767763 Chr.22: 37074790-37074790 on Build GRCh38
rs758544017 Chr.22: 37074751-37074751 on Build GRCh38
rs773570300 Chr.22: 37074635-37074635 on Build GRCh38
rs777214750 Chr.22: 37074592-37074592 on Build GRCh38
rs151074901 Chr.22: 37074641-37074641 on Build GRCh38
rs762514821 Chr.22: 37074962-37074962 on Build GRCh38
rs761569553 Chr.22: 37075000-37075001 on Build GRCh38
rs759598363 Chr.22: 37075003-37075004 on Build GRCh38
rs201823626 Chr.22: 37074593-37074593 on Build GRCh38
rs959406664 Chr.22: 37074931-37074931 on Build GRCh38
rs778387386 Chr.22: 37074756-37074756 on Build GRCh38
rs746448716 Chr.22: 37074673-37074673 on Build GRCh38
rs1026954426 Chr.22: 37074819-37074819 on Build GRCh38
rs995916391 Chr.22: 37074812-37074812 on Build GRCh38
rs923158208 Chr.22: 37074912-37074912 on Build GRCh38
rs529751538 Chr.22: 37074920-37074920 on Build GRCh38
rs565920151 Chr.22: 37075015-37075015 on Build GRCh38
rs111807510 Chr.22: 37074595-37074595 on Build GRCh38
rs368147579 Chr.22: 37074637-37074637 on Build GRCh38
rs548035232 Chr.22: 37074943-37074943 on Build GRCh38
rs766419811 Chr.22: 37074742-37074742 on Build GRCh38
rs542016025 Chr.22: 37074767-37074767 on Build GRCh38
rs767320092 Chr.22: 37074604-37074604 on Build GRCh38
rs747559770 Chr.22: 37074632-37074632 on Build GRCh38
rs79816125 Chr.22: 37074600-37074600 on Build GRCh38
rs940535872 Chr.22: 37074623-37074623 on Build GRCh38
rs768818027 Chr.22: 37074699-37074699 on Build GRCh38
rs13056055 Chr.22: 37074809-37074809 on Build GRCh38
rs113697947 Chr.22: 37074768-37074768 on Build GRCh38
rs141869125 Chr.22: 37074689-37074689 on Build GRCh38
rs1006974873 Chr.22: 37074840-37074840 on Build GRCh38
rs954117655 Chr.22: 37074713-37074713 on Build GRCh38
rs372156816 Chr.22: 37074728-37074728 on Build GRCh38
rs111225958 Chr.22: 37075001-37075002 on Build GRCh38
rs775769858 Chr.22: 37074682-37074682 on Build GRCh38
rs755966392 Chr.22: 37074561-37074561 on Build GRCh38
rs917638154 Chr.22: 37074963-37074963 on Build GRCh38
rs768268119 Chr.22: 37074576-37074576 on Build GRCh38
rs1043115385 Chr.22: 37075002-37075002 on Build GRCh38
rs1036363839 Chr.22: 37074589-37074589 on Build GRCh38
rs995492515 Chr.22: 37074635-37074635 on Build GRCh38
rs775005599 Chr.22: 37074846-37074846 on Build GRCh38
rs543898364 Chr.22: 37074903-37074903 on Build GRCh38
rs971956588 Chr.22: 37074719-37074719 on Build GRCh38

View Full Product Details