Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37612891-37613290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGA1
Entrez Gene ID
26088
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 1
Gene Aliases
-
UniGene
Hs.499158
Gene Chromosome Location
Chr.22: 37607235-37633564 on Build GRCh38
Transcripts
NM_001001560.2 NM_001172687.1 NM_001172688.1 NM_013365.4 XM_005261517.3 XM_005261518.1 XM_005261520.1 XM_006724229.1 XM_011530122.1 XM_011530123.1 XM_011530124.1 XM_017028760.1 XM_017028761.1 XM_017028762.1 XM_017028763.1 XM_017028764.1

SNP Detail

SNP ID Chromosome Location
rs549666721 Chr.22: 37612964-37612964 on Build GRCh38
rs761851445 Chr.22: 37613247-37613247 on Build GRCh38
rs923403131 Chr.22: 37613204-37613204 on Build GRCh38
rs964529408 Chr.22: 37613180-37613180 on Build GRCh38
rs1042590219 Chr.22: 37613223-37613223 on Build GRCh38
rs1012741302 Chr.22: 37612984-37612984 on Build GRCh38
rs974881337 Chr.22: 37613186-37613186 on Build GRCh38
rs535196586 Chr.22: 37612966-37612966 on Build GRCh38
rs765352175 Chr.22: 37613255-37613255 on Build GRCh38
rs114580473 Chr.22: 37613120-37613120 on Build GRCh38
rs940954212 Chr.22: 37613115-37613115 on Build GRCh38
rs950402936 Chr.22: 37613091-37613091 on Build GRCh38
rs902479214 Chr.22: 37612948-37612948 on Build GRCh38
rs1050459754 Chr.22: 37613198-37613198 on Build GRCh38
rs948170165 Chr.22: 37613026-37613026 on Build GRCh38
rs936649757 Chr.22: 37613235-37613235 on Build GRCh38
rs1053819977 Chr.22: 37613236-37613236 on Build GRCh38
rs1022837512 Chr.22: 37613013-37613013 on Build GRCh38
rs971299705 Chr.22: 37613031-37613031 on Build GRCh38
rs1006685122 Chr.22: 37613246-37613246 on Build GRCh38
rs564390935 Chr.22: 37613107-37613107 on Build GRCh38
rs1036643994 Chr.22: 37613116-37613116 on Build GRCh38
rs761336 Chr.22: 37613234-37613234 on Build GRCh38
rs565426113 Chr.22: 37612981-37612981 on Build GRCh38
rs933499321 Chr.22: 37613220-37613220 on Build GRCh38
rs933360226 Chr.22: 37613197-37613197 on Build GRCh38
rs1042622696 Chr.22: 37612941-37612941 on Build GRCh38
rs999831731 Chr.22: 37613268-37613268 on Build GRCh38
rs968371692 Chr.22: 37612915-37612915 on Build GRCh38
rs74974951 Chr.22: 37613175-37613175 on Build GRCh38
rs977761096 Chr.22: 37612957-37612957 on Build GRCh38
rs759966599 Chr.22: 37613029-37613030 on Build GRCh38
rs909444716 Chr.22: 37613070-37613070 on Build GRCh38
rs991906067 Chr.22: 37612996-37612996 on Build GRCh38
rs184055745 Chr.22: 37612972-37612972 on Build GRCh38
rs551963085 Chr.22: 37612971-37612971 on Build GRCh38
rs933614627 Chr.22: 37612928-37612928 on Build GRCh38
rs1026237295 Chr.22: 37613065-37613065 on Build GRCh38
rs555622208 Chr.22: 37613183-37613183 on Build GRCh38
rs908945180 Chr.22: 37613124-37613124 on Build GRCh38
rs187632405 Chr.22: 37613063-37613063 on Build GRCh38
rs57600472 Chr.22: 37612965-37612965 on Build GRCh38
rs916368543 Chr.22: 37613010-37613010 on Build GRCh38
rs7511450 Chr.22: 37612974-37612974 on Build GRCh38

View Full Product Details