Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41658159-41658586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XRCC6
Entrez Gene ID
2547
Gene Name
X-ray repair cross complementing 6
Gene Aliases
CTC75, CTCBF, G22P1, KU70, ML8, TLAA
UniGene
Hs.292493
Gene Chromosome Location
Chr.22: 41621163-41664048 on Build GRCh38
Transcripts
NM_001288976.1 NM_001288977.1 NM_001288978.1 NM_001469.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
303582 c.1443C>T p.P481P Chr.22: 41658273-41658273 on Build GRCh38
385169 c.1499C>T p.P500L Chr.22: 41658329-41658329 on Build GRCh38
4104501 c.1493T>G p.M498R Chr.22: 41658323-41658323 on Build GRCh38
5859482 c.1471G>A p.E491K Chr.22: 41658301-41658301 on Build GRCh38
5611206 c.1422-1G>T p.? Chr.22: 41658251-41658251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878900250 Chr.22: 41658192-41658192 on Build GRCh38
rs912659770 Chr.22: 41658555-41658555 on Build GRCh38
rs975096171 Chr.22: 41658439-41658439 on Build GRCh38
rs921854326 Chr.22: 41658526-41658526 on Build GRCh38
rs770785582 Chr.22: 41658201-41658201 on Build GRCh38
rs983523013 Chr.22: 41658506-41658506 on Build GRCh38
rs769531721 Chr.22: 41658197-41658197 on Build GRCh38
rs774400267 Chr.22: 41658204-41658204 on Build GRCh38
rs560913351 Chr.22: 41658426-41658426 on Build GRCh38
rs374917433 Chr.22: 41658351-41658351 on Build GRCh38
rs72547483 Chr.22: 41658361-41658361 on Build GRCh38
rs540130873 Chr.22: 41658528-41658528 on Build GRCh38
rs767776641 Chr.22: 41658228-41658228 on Build GRCh38
rs774174738 Chr.22: 41658372-41658372 on Build GRCh38
rs1019014080 Chr.22: 41658508-41658508 on Build GRCh38
rs372273063 Chr.22: 41658223-41658223 on Build GRCh38
rs556993005 Chr.22: 41658364-41658364 on Build GRCh38
rs189779012 Chr.22: 41658385-41658385 on Build GRCh38
rs538301994 Chr.22: 41658236-41658238 on Build GRCh38
rs4084339 Chr.22: 41658287-41658287 on Build GRCh38
rs759777154 Chr.22: 41658216-41658216 on Build GRCh38
rs749345195 Chr.22: 41658347-41658348 on Build GRCh38
rs369043204 Chr.22: 41658402-41658402 on Build GRCh38
rs754651274 Chr.22: 41658350-41658350 on Build GRCh38
rs200180852 Chr.22: 41658369-41658369 on Build GRCh38
rs200669332 Chr.22: 41658362-41658362 on Build GRCh38
rs750813587 Chr.22: 41658452-41658452 on Build GRCh38
rs762450127 Chr.22: 41658315-41658315 on Build GRCh38
rs754380678 Chr.22: 41658303-41658303 on Build GRCh38
rs1023208212 Chr.22: 41658179-41658179 on Build GRCh38
rs534724306 Chr.22: 41658235-41658235 on Build GRCh38
rs553591070 Chr.22: 41658242-41658242 on Build GRCh38
rs987294359 Chr.22: 41658554-41658554 on Build GRCh38
rs1045477625 Chr.22: 41658190-41658190 on Build GRCh38
rs1035864190 Chr.22: 41658318-41658318 on Build GRCh38
rs750789414 Chr.22: 41658335-41658335 on Build GRCh38
rs139850731 Chr.22: 41658273-41658273 on Build GRCh38
rs573024174 Chr.22: 41658488-41658488 on Build GRCh38
rs772279023 Chr.22: 41658388-41658388 on Build GRCh38
rs553304643 Chr.22: 41658332-41658332 on Build GRCh38
rs1027653904 Chr.22: 41658470-41658470 on Build GRCh38
rs1015924753 Chr.22: 41658384-41658384 on Build GRCh38
rs954721466 Chr.22: 41658529-41658529 on Build GRCh38
rs950801264 Chr.22: 41658473-41658473 on Build GRCh38
rs536576177 Chr.22: 41658476-41658476 on Build GRCh38
rs1001294599 Chr.22: 41658328-41658328 on Build GRCh38
rs765648597 Chr.22: 41658436-41658436 on Build GRCh38
rs777806911 Chr.22: 41658367-41658367 on Build GRCh38
rs768736700 Chr.22: 41658369-41658371 on Build GRCh38
rs757515014 Chr.22: 41658323-41658323 on Build GRCh38
rs752760439 Chr.22: 41658232-41658232 on Build GRCh38
rs113005024 Chr.22: 41658218-41658218 on Build GRCh38
rs1034163867 Chr.22: 41658330-41658330 on Build GRCh38

View Full Product Details