Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46912006-46912501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D22A
Entrez Gene ID
25771
Gene Name
TBC1 domain family member 22A
Gene Aliases
C22orf4, HSC79E021
UniGene
Hs.435044
Gene Chromosome Location
Chr.22: 46762617-47224975 on Build GRCh38
Transcripts
NM_001284303.1 NM_001284304.1 NM_001284305.1 NM_014346.3 NR_104292.1 XM_005261496.1 XM_011530099.1 XM_011530100.2 XM_011530101.2 XM_011530102.2 XM_017028742.1 XM_017028743.1 XM_017028744.1 XM_017028745.1 XM_017028746.1 XM_017028747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
189366 c.935G>A p.R312H Chr.22: 46912108-46912108 on Build GRCh38
7393119 c.829G>A p.D277N Chr.22: 46912143-46912143 on Build GRCh38
7423420 c.805A>T p.S269C Chr.22: 46912119-46912119 on Build GRCh38
7121969 c.787G>A p.A263T Chr.22: 46912101-46912101 on Build GRCh38
7423419 c.946A>T p.S316C Chr.22: 46912119-46912119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756620844 Chr.22: 46912148-46912148 on Build GRCh38
rs28694630 Chr.22: 46912429-46912429 on Build GRCh38
rs75872785 Chr.22: 46912038-46912038 on Build GRCh38
rs757282399 Chr.22: 46912181-46912181 on Build GRCh38
rs974527911 Chr.22: 46912260-46912261 on Build GRCh38
rs1051774336 Chr.22: 46912426-46912426 on Build GRCh38
rs559010732 Chr.22: 46912052-46912052 on Build GRCh38
rs758017404 Chr.22: 46912469-46912469 on Build GRCh38
rs745361736 Chr.22: 46912103-46912103 on Build GRCh38
rs567756861 Chr.22: 46912213-46912213 on Build GRCh38
rs953262044 Chr.22: 46912182-46912182 on Build GRCh38
rs898296254 Chr.22: 46912128-46912142 on Build GRCh38
rs773347703 Chr.22: 46912136-46912136 on Build GRCh38
rs936427654 Chr.22: 46912401-46912401 on Build GRCh38
rs758068427 Chr.22: 46912367-46912367 on Build GRCh38
rs762431913 Chr.22: 46912128-46912128 on Build GRCh38
rs757123843 Chr.22: 46912054-46912054 on Build GRCh38
rs142368974 Chr.22: 46912053-46912053 on Build GRCh38
rs751794345 Chr.22: 46912152-46912152 on Build GRCh38
rs918596050 Chr.22: 46912272-46912272 on Build GRCh38
rs530158950 Chr.22: 46912284-46912284 on Build GRCh38
rs779710071 Chr.22: 46912219-46912219 on Build GRCh38
rs988342620 Chr.22: 46912107-46912107 on Build GRCh38
rs747395220 Chr.22: 46912111-46912111 on Build GRCh38
rs754601580 Chr.22: 46912223-46912223 on Build GRCh38
rs9615420 Chr.22: 46912326-46912326 on Build GRCh38
rs200163466 Chr.22: 46912109-46912109 on Build GRCh38
rs749613020 Chr.22: 46912319-46912319 on Build GRCh38
rs992161351 Chr.22: 46912291-46912291 on Build GRCh38
rs997439283 Chr.22: 46912414-46912414 on Build GRCh38
rs991829335 Chr.22: 46912288-46912288 on Build GRCh38
rs548444280 Chr.22: 46912149-46912149 on Build GRCh38
rs374879123 Chr.22: 46912341-46912341 on Build GRCh38
rs926385658 Chr.22: 46912235-46912235 on Build GRCh38
rs929591536 Chr.22: 46912298-46912298 on Build GRCh38
rs918122422 Chr.22: 46912290-46912290 on Build GRCh38
rs746668887 Chr.22: 46912086-46912087 on Build GRCh38
rs566826749 Chr.22: 46912366-46912366 on Build GRCh38
rs746702904 Chr.22: 46912222-46912222 on Build GRCh38
rs948284666 Chr.22: 46912339-46912339 on Build GRCh38
rs9616149 Chr.22: 46912176-46912176 on Build GRCh38
rs891014974 Chr.22: 46912446-46912446 on Build GRCh38
rs937649855 Chr.22: 46912261-46912261 on Build GRCh38
rs767628265 Chr.22: 46912060-46912060 on Build GRCh38
rs5769241 Chr.22: 46912188-46912188 on Build GRCh38
rs771756934 Chr.22: 46912108-46912108 on Build GRCh38
rs145498680 Chr.22: 46912333-46912333 on Build GRCh38
rs373469514 Chr.22: 46912066-46912066 on Build GRCh38
rs764033398 Chr.22: 46912169-46912169 on Build GRCh38
rs768967302 Chr.22: 46912113-46912113 on Build GRCh38
rs140392080 Chr.22: 46912127-46912127 on Build GRCh38
rs778539219 Chr.22: 46912102-46912102 on Build GRCh38
rs371872564 Chr.22: 46912086-46912086 on Build GRCh38
rs900629834 Chr.22: 46912407-46912407 on Build GRCh38
rs369108625 Chr.22: 46912173-46912173 on Build GRCh38
rs780434178 Chr.22: 46912214-46912214 on Build GRCh38
rs759803136 Chr.22: 46912160-46912160 on Build GRCh38
rs997619854 Chr.22: 46912421-46912421 on Build GRCh38
rs770198561 Chr.22: 46912132-46912132 on Build GRCh38
rs369229931 Chr.22: 46912237-46912237 on Build GRCh38
rs765013600 Chr.22: 46912195-46912195 on Build GRCh38
rs112701218 Chr.22: 46912233-46912233 on Build GRCh38
rs1009500881 Chr.22: 46912452-46912452 on Build GRCh38
rs61074215 Chr.22: 46912405-46912405 on Build GRCh38
rs371300371 Chr.22: 46912185-46912185 on Build GRCh38

View Full Product Details