Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10164757-10165258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRAK2
Entrez Gene ID
3656
Gene Name
interleukin 1 receptor associated kinase 2
Gene Aliases
IRAK-2
UniGene
Hs.449207
Gene Chromosome Location
Chr.3: 10164879-10243743 on Build GRCh38
Transcripts
NM_001570.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7447390 c.77G>A p.W26* Chr.3: 10165031-10165031 on Build GRCh38
6229488 c.37C>T p.L13L Chr.3: 10164991-10164991 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781024442 Chr.3: 10165085-10165085 on Build GRCh38
rs146076327 Chr.3: 10164953-10164953 on Build GRCh38
rs767038966 Chr.3: 10164973-10164973 on Build GRCh38
rs572730889 Chr.3: 10164857-10164857 on Build GRCh38
rs1055254300 Chr.3: 10164803-10164803 on Build GRCh38
rs1000248896 Chr.3: 10165138-10165138 on Build GRCh38
rs779997859 Chr.3: 10164997-10164997 on Build GRCh38
rs201125447 Chr.3: 10164971-10164971 on Build GRCh38
rs763606474 Chr.3: 10164980-10164980 on Build GRCh38
rs748394830 Chr.3: 10165020-10165020 on Build GRCh38
rs745812166 Chr.3: 10164926-10164926 on Build GRCh38
rs964861347 Chr.3: 10164820-10164820 on Build GRCh38
rs200268545 Chr.3: 10164984-10164984 on Build GRCh38
rs540248368 Chr.3: 10164861-10164861 on Build GRCh38
rs765361322 Chr.3: 10164955-10164955 on Build GRCh38
rs772064442 Chr.3: 10164927-10164927 on Build GRCh38
rs969876243 Chr.3: 10164854-10164854 on Build GRCh38
rs1019512149 Chr.3: 10164913-10164913 on Build GRCh38
rs11465852 Chr.3: 10165170-10165170 on Build GRCh38
rs991277667 Chr.3: 10164979-10164979 on Build GRCh38
rs747170520 Chr.3: 10165231-10165231 on Build GRCh38
rs371308540 Chr.3: 10164909-10164909 on Build GRCh38
rs748031391 Chr.3: 10164996-10164997 on Build GRCh38
rs536880697 Chr.3: 10165175-10165175 on Build GRCh38
rs186431561 Chr.3: 10165160-10165160 on Build GRCh38
rs776649062 Chr.3: 10165017-10165017 on Build GRCh38
rs758309915 Chr.3: 10164990-10164990 on Build GRCh38
rs968775508 Chr.3: 10164904-10164904 on Build GRCh38
rs949384848 Chr.3: 10164983-10164983 on Build GRCh38
rs181986744 Chr.3: 10164793-10164793 on Build GRCh38
rs756817971 Chr.3: 10164922-10164922 on Build GRCh38
rs199769069 Chr.3: 10165071-10165071 on Build GRCh38
rs11465851 Chr.3: 10165163-10165163 on Build GRCh38
rs749280257 Chr.3: 10164873-10164873 on Build GRCh38
rs763536692 Chr.3: 10164965-10164965 on Build GRCh38
rs915629780 Chr.3: 10164850-10164850 on Build GRCh38
rs754934207 Chr.3: 10165084-10165084 on Build GRCh38
rs549192265 Chr.3: 10165007-10165007 on Build GRCh38
rs966602810 Chr.3: 10164917-10164917 on Build GRCh38
rs745440775 Chr.3: 10164985-10164985 on Build GRCh38
rs532457605 Chr.3: 10164940-10164940 on Build GRCh38
rs112190415 Chr.3: 10164870-10164870 on Build GRCh38
rs778659393 Chr.3: 10164925-10164925 on Build GRCh38
rs1046399172 Chr.3: 10164993-10164993 on Build GRCh38
rs747938054 Chr.3: 10165094-10165094 on Build GRCh38
rs749893720 Chr.3: 10165078-10165078 on Build GRCh38
rs970317126 Chr.3: 10165183-10165183 on Build GRCh38
rs571566202 Chr.3: 10165105-10165105 on Build GRCh38
rs1018787788 Chr.3: 10164805-10164805 on Build GRCh38
rs746846013 Chr.3: 10164944-10164944 on Build GRCh38
rs763168994 Chr.3: 10165039-10165039 on Build GRCh38
rs907872401 Chr.3: 10165192-10165192 on Build GRCh38
rs968827742 Chr.3: 10165127-10165127 on Build GRCh38
rs995663150 Chr.3: 10164878-10164878 on Build GRCh38
rs1025006891 Chr.3: 10165174-10165174 on Build GRCh38
rs766957274 Chr.3: 10165048-10165048 on Build GRCh38
rs757925483 Chr.3: 10165082-10165082 on Build GRCh38
rs769965191 Chr.3: 10165028-10165028 on Build GRCh38
rs768508288 Chr.3: 10165012-10165012 on Build GRCh38
rs899325438 Chr.3: 10165067-10165067 on Build GRCh38
rs768720778 Chr.3: 10164948-10164948 on Build GRCh38
rs776932620 Chr.3: 10164949-10164949 on Build GRCh38
rs773490044 Chr.3: 10164962-10164962 on Build GRCh38
rs978541290 Chr.3: 10164866-10164866 on Build GRCh38
rs755429241 Chr.3: 10164976-10164976 on Build GRCh38
rs756655853 Chr.3: 10164982-10164982 on Build GRCh38
rs546602804 Chr.3: 10165074-10165074 on Build GRCh38
rs1047368044 Chr.3: 10164787-10164787 on Build GRCh38
rs375575112 Chr.3: 10165061-10165061 on Build GRCh38
rs970767627 Chr.3: 10165181-10165181 on Build GRCh38
rs1008007396 Chr.3: 10165104-10165104 on Build GRCh38
rs978025301 Chr.3: 10164932-10164932 on Build GRCh38
rs777526312 Chr.3: 10164919-10164919 on Build GRCh38
rs767379826 Chr.3: 10165044-10165045 on Build GRCh38
rs751945187 Chr.3: 10164974-10164974 on Build GRCh38
rs868091278 Chr.3: 10164996-10164996 on Build GRCh38
rs753072005 Chr.3: 10165062-10165062 on Build GRCh38
rs759765333 Chr.3: 10165057-10165057 on Build GRCh38
rs561460688 Chr.3: 10165056-10165056 on Build GRCh38
rs773400153 Chr.3: 10165038-10165038 on Build GRCh38
rs1009221225 Chr.3: 10164794-10164794 on Build GRCh38
rs1028381756 Chr.3: 10164897-10164897 on Build GRCh38
rs1032263063 Chr.3: 10165165-10165165 on Build GRCh38
rs770023382 Chr.3: 10165097-10165097 on Build GRCh38
rs371052497 Chr.3: 10165209-10165209 on Build GRCh38
rs768396161 Chr.3: 10164905-10164905 on Build GRCh38
rs1022117683 Chr.3: 10165114-10165114 on Build GRCh38
rs748710439 Chr.3: 10164921-10164921 on Build GRCh38
rs371307726 Chr.3: 10165083-10165083 on Build GRCh38
rs996438567 Chr.3: 10165100-10165100 on Build GRCh38

View Full Product Details