Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 44735276-44735789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF501
Entrez Gene ID
115560
Gene Name
zinc finger protein 501
Gene Aliases
ZNF, ZNF52
UniGene
Hs.401045
Gene Chromosome Location
Chr.3: 44729606-44737083 on Build GRCh38
Transcripts
NM_001258280.1 NM_145044.3

SNP Detail

SNP ID Chromosome Location
rs969919792 Chr.3: 44735653-44735653 on Build GRCh38
rs889979239 Chr.3: 44735523-44735523 on Build GRCh38
rs947017821 Chr.3: 44735745-44735745 on Build GRCh38
rs928425862 Chr.3: 44735701-44735701 on Build GRCh38
rs943185788 Chr.3: 44735392-44735392 on Build GRCh38
rs146462196 Chr.3: 44735465-44735465 on Build GRCh38
rs571677368 Chr.3: 44735488-44735488 on Build GRCh38
rs538190503 Chr.3: 44735428-44735428 on Build GRCh38
rs1025656741 Chr.3: 44735582-44735582 on Build GRCh38
rs375975679 Chr.3: 44735611-44735611 on Build GRCh38
rs905576800 Chr.3: 44735766-44735766 on Build GRCh38
rs975919894 Chr.3: 44735654-44735654 on Build GRCh38
rs923103473 Chr.3: 44735684-44735684 on Build GRCh38
rs529509614 Chr.3: 44735487-44735487 on Build GRCh38
rs947655922 Chr.3: 44735512-44735512 on Build GRCh38
rs938392816 Chr.3: 44735463-44735463 on Build GRCh38
rs979882827 Chr.3: 44735449-44735449 on Build GRCh38
rs1011353522 Chr.3: 44735474-44735474 on Build GRCh38
rs928403469 Chr.3: 44735455-44735455 on Build GRCh38
rs914254662 Chr.3: 44735722-44735722 on Build GRCh38
rs1002333735 Chr.3: 44735515-44735515 on Build GRCh38
rs555871918 Chr.3: 44735748-44735748 on Build GRCh38
rs745488903 Chr.3: 44735357-44735358 on Build GRCh38
rs371787199 Chr.3: 44735582-44735585 on Build GRCh38
rs959832663 Chr.3: 44735705-44735705 on Build GRCh38
rs1044154076 Chr.3: 44735765-44735765 on Build GRCh38
rs994625796 Chr.3: 44735567-44735567 on Build GRCh38
rs934568040 Chr.3: 44735427-44735427 on Build GRCh38
rs182998471 Chr.3: 44735546-44735546 on Build GRCh38
rs1040272110 Chr.3: 44735397-44735397 on Build GRCh38
rs142246848 Chr.3: 44735678-44735682 on Build GRCh38
rs188699749 Chr.3: 44735671-44735671 on Build GRCh38
rs762613751 Chr.3: 44735489-44735489 on Build GRCh38
rs1052827356 Chr.3: 44735462-44735462 on Build GRCh38
rs554332475 Chr.3: 44735547-44735547 on Build GRCh38
rs879177721 Chr.3: 44735556-44735556 on Build GRCh38
rs910599361 Chr.3: 44735613-44735613 on Build GRCh38
rs555199418 Chr.3: 44735652-44735652 on Build GRCh38
rs934617782 Chr.3: 44735716-44735716 on Build GRCh38
rs901258736 Chr.3: 44735560-44735560 on Build GRCh38
rs59932401 Chr.3: 44735353-44735353 on Build GRCh38
rs143348081 Chr.3: 44735718-44735718 on Build GRCh38
rs139343016 Chr.3: 44735650-44735650 on Build GRCh38
rs909069335 Chr.3: 44735368-44735368 on Build GRCh38
rs767546245 Chr.3: 44735581-44735581 on Build GRCh38
rs766321834 Chr.3: 44735497-44735497 on Build GRCh38
rs1043323317 Chr.3: 44735737-44735737 on Build GRCh38
rs567456458 Chr.3: 44735408-44735408 on Build GRCh38
rs755030522 Chr.3: 44735562-44735562 on Build GRCh38
rs562053419 Chr.3: 44735458-44735458 on Build GRCh38
rs566120980 Chr.3: 44735732-44735732 on Build GRCh38
rs950671140 Chr.3: 44735310-44735310 on Build GRCh38
rs952804834 Chr.3: 44735608-44735608 on Build GRCh38
rs1006171880 Chr.3: 44735618-44735618 on Build GRCh38
rs757608956 Chr.3: 44735580-44735580 on Build GRCh38

View Full Product Details