Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46883223-46883714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTH1R
Entrez Gene ID
5745
Gene Name
parathyroid hormone 1 receptor
Gene Aliases
PFE, PTHR, PTHR1
UniGene
Hs.1019
Gene Chromosome Location
Chr.3: 46877690-46903799 on Build GRCh38
Transcripts
NM_000316.2 NM_001184744.1 XM_011533967.2 XM_011533968.2 XM_017006932.1 XM_017006933.1 XM_017006934.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6278959 c.49C>T p.P17S Chr.3: 46883608-46883608 on Build GRCh38
4798386 c.11C>T p.A4V Chr.3: 46883570-46883570 on Build GRCh38
6199722 c.32C>A p.A11E Chr.3: 46883591-46883591 on Build GRCh38
5582264 c.26G>A p.G9D Chr.3: 46883585-46883585 on Build GRCh38
7386096 c.44G>A p.C15Y Chr.3: 46883603-46883603 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1039854062 Chr.3: 46883283-46883283 on Build GRCh38
rs934768900 Chr.3: 46883667-46883667 on Build GRCh38
rs942845504 Chr.3: 46883282-46883282 on Build GRCh38
rs924774746 Chr.3: 46883655-46883655 on Build GRCh38
rs752702883 Chr.3: 46883544-46883544 on Build GRCh38
rs199993250 Chr.3: 46883675-46883675 on Build GRCh38
rs772304416 Chr.3: 46883611-46883611 on Build GRCh38
rs934206321 Chr.3: 46883382-46883382 on Build GRCh38
rs567440911 Chr.3: 46883593-46883593 on Build GRCh38
rs775859348 Chr.3: 46883515-46883517 on Build GRCh38
rs979244821 Chr.3: 46883259-46883259 on Build GRCh38
rs959937327 Chr.3: 46883578-46883578 on Build GRCh38
rs923024127 Chr.3: 46883342-46883342 on Build GRCh38
rs559917886 Chr.3: 46883346-46883346 on Build GRCh38
rs749062175 Chr.3: 46883610-46883610 on Build GRCh38
rs571514959 Chr.3: 46883661-46883661 on Build GRCh38
rs967777439 Chr.3: 46883520-46883520 on Build GRCh38
rs200603325 Chr.3: 46883646-46883646 on Build GRCh38
rs926145116 Chr.3: 46883279-46883279 on Build GRCh38
rs766284000 Chr.3: 46883664-46883664 on Build GRCh38
rs530204092 Chr.3: 46883377-46883377 on Build GRCh38
rs780469309 Chr.3: 46883616-46883616 on Build GRCh38
rs1001332385 Chr.3: 46883451-46883451 on Build GRCh38
rs899774952 Chr.3: 46883680-46883680 on Build GRCh38
rs975589924 Chr.3: 46883585-46883585 on Build GRCh38
rs777569371 Chr.3: 46883583-46883583 on Build GRCh38
rs1014568862 Chr.3: 46883508-46883508 on Build GRCh38
rs756013791 Chr.3: 46883565-46883565 on Build GRCh38
rs1051456293 Chr.3: 46883383-46883384 on Build GRCh38
rs1035372423 Chr.3: 46883558-46883558 on Build GRCh38
rs967512144 Chr.3: 46883250-46883250 on Build GRCh38
rs960003432 Chr.3: 46883490-46883490 on Build GRCh38
rs886086563 Chr.3: 46883388-46883388 on Build GRCh38
rs931215495 Chr.3: 46883693-46883693 on Build GRCh38
rs899042391 Chr.3: 46883410-46883410 on Build GRCh38
rs987920318 Chr.3: 46883669-46883669 on Build GRCh38
rs1020315567 Chr.3: 46883517-46883517 on Build GRCh38
rs547769292 Chr.3: 46883336-46883336 on Build GRCh38
rs978882525 Chr.3: 46883521-46883521 on Build GRCh38
rs201663714 Chr.3: 46883523-46883523 on Build GRCh38
rs913201556 Chr.3: 46883245-46883245 on Build GRCh38
rs751447035 Chr.3: 46883679-46883679 on Build GRCh38
rs1023200257 Chr.3: 46883625-46883625 on Build GRCh38
rs200851488 Chr.3: 46883676-46883676 on Build GRCh38
rs199757613 Chr.3: 46883547-46883547 on Build GRCh38
rs1004664667 Chr.3: 46883407-46883407 on Build GRCh38
rs1034118126 Chr.3: 46883483-46883483 on Build GRCh38
rs200710310 Chr.3: 46883643-46883643 on Build GRCh38

View Full Product Details