Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49808303-49808811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBA7
Entrez Gene ID
7318
Gene Name
ubiquitin like modifier activating enzyme 7
Gene Aliases
D8, UBA1B, UBE1L, UBE2
UniGene
Hs.16695
Gene Chromosome Location
Chr.3: 49805205-49813958 on Build GRCh38
Transcripts
NM_003335.2 XM_005265430.2 XM_006713321.2 XM_011534070.1 XM_011534071.1 XM_011534072.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1231521 c.2399G>A p.G800D Chr.3: 49808417-49808417 on Build GRCh38
4803156 c.2358G>T p.Q786H Chr.3: 49808458-49808458 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1045423429 Chr.3: 49808575-49808575 on Build GRCh38
rs911742773 Chr.3: 49808466-49808466 on Build GRCh38
rs758122771 Chr.3: 49808375-49808375 on Build GRCh38
rs1048779133 Chr.3: 49808613-49808613 on Build GRCh38
rs953192011 Chr.3: 49808656-49808656 on Build GRCh38
rs760979719 Chr.3: 49808475-49808475 on Build GRCh38
rs1000797550 Chr.3: 49808736-49808736 on Build GRCh38
rs201545211 Chr.3: 49808483-49808483 on Build GRCh38
rs944249809 Chr.3: 49808424-49808425 on Build GRCh38
rs764743288 Chr.3: 49808480-49808480 on Build GRCh38
rs147771273 Chr.3: 49808571-49808571 on Build GRCh38
rs914903541 Chr.3: 49808387-49808387 on Build GRCh38
rs764324446 Chr.3: 49808479-49808479 on Build GRCh38
rs1045688619 Chr.3: 49808689-49808689 on Build GRCh38
rs779956373 Chr.3: 49808377-49808377 on Build GRCh38
rs750949798 Chr.3: 49808632-49808632 on Build GRCh38
rs759941295 Chr.3: 49808631-49808631 on Build GRCh38
rs754106128 Chr.3: 49808482-49808482 on Build GRCh38
rs1021942279 Chr.3: 49808634-49808634 on Build GRCh38
rs948320182 Chr.3: 49808685-49808685 on Build GRCh38
rs769469402 Chr.3: 49808415-49808415 on Build GRCh38
rs1048367795 Chr.3: 49808508-49808508 on Build GRCh38
rs760182417 Chr.3: 49808419-49808419 on Build GRCh38
rs746791357 Chr.3: 49808385-49808385 on Build GRCh38
rs930075639 Chr.3: 49808500-49808500 on Build GRCh38
rs985722635 Chr.3: 49808660-49808660 on Build GRCh38
rs532181851 Chr.3: 49808515-49808515 on Build GRCh38
rs896684134 Chr.3: 49808645-49808645 on Build GRCh38
rs368439645 Chr.3: 49808523-49808523 on Build GRCh38
rs968973609 Chr.3: 49808403-49808403 on Build GRCh38
rs750346004 Chr.3: 49808510-49808510 on Build GRCh38
rs1012154809 Chr.3: 49808570-49808570 on Build GRCh38
rs956805045 Chr.3: 49808333-49808333 on Build GRCh38
rs202039284 Chr.3: 49808359-49808359 on Build GRCh38
rs918775416 Chr.3: 49808776-49808776 on Build GRCh38
rs529901000 Chr.3: 49808614-49808614 on Build GRCh38
rs778797747 Chr.3: 49808342-49808342 on Build GRCh38
rs763436628 Chr.3: 49808417-49808417 on Build GRCh38
rs369872655 Chr.3: 49808713-49808713 on Build GRCh38
rs552615981 Chr.3: 49808686-49808686 on Build GRCh38
rs369476968 Chr.3: 49808487-49808487 on Build GRCh38
rs2234386 Chr.3: 49808409-49808409 on Build GRCh38
rs986139590 Chr.3: 49808408-49808409 on Build GRCh38
rs911425292 Chr.3: 49808677-49808677 on Build GRCh38
rs554277198 Chr.3: 49808734-49808734 on Build GRCh38
rs759203296 Chr.3: 49808484-49808484 on Build GRCh38
rs543591062 Chr.3: 49808414-49808414 on Build GRCh38
rs750396891 Chr.3: 49808370-49808370 on Build GRCh38
rs772474523 Chr.3: 49808423-49808423 on Build GRCh38
rs921854556 Chr.3: 49808562-49808562 on Build GRCh38
rs867682060 Chr.3: 49808459-49808459 on Build GRCh38
rs775860371 Chr.3: 49808450-49808450 on Build GRCh38
rs763046595 Chr.3: 49808411-49808411 on Build GRCh38
rs1041199902 Chr.3: 49808434-49808434 on Build GRCh38
rs2234385 Chr.3: 49808608-49808608 on Build GRCh38
rs949736364 Chr.3: 49808581-49808581 on Build GRCh38
rs774994589 Chr.3: 49808418-49808418 on Build GRCh38

View Full Product Details