Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52435823-52436330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA3G
Entrez Gene ID
56920
Gene Name
semaphorin 3G
Gene Aliases
sem2
UniGene
Hs.59729
Gene Chromosome Location
Chr.3: 52433252-52445027 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5471777 c.2084G>A p.R695Q Chr.3: 52435868-52435868 on Build GRCh38
5011113 c.2024T>C p.V675A Chr.3: 52435928-52435928 on Build GRCh38
7413483 c.2065C>T p.P689S Chr.3: 52435887-52435887 on Build GRCh38
4724712 c.2054C>T p.P685L Chr.3: 52435898-52435898 on Build GRCh38
5568701 c.1936A>G p.S646G Chr.3: 52436016-52436016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780096547 Chr.3: 52435849-52435849 on Build GRCh38
rs200593686 Chr.3: 52435878-52435878 on Build GRCh38
rs758693843 Chr.3: 52436064-52436064 on Build GRCh38
rs747528320 Chr.3: 52435995-52435995 on Build GRCh38
rs747545193 Chr.3: 52436122-52436122 on Build GRCh38
rs779447141 Chr.3: 52436036-52436036 on Build GRCh38
rs750442070 Chr.3: 52435847-52435847 on Build GRCh38
rs757711856 Chr.3: 52436080-52436080 on Build GRCh38
rs760629721 Chr.3: 52436011-52436011 on Build GRCh38
rs529286895 Chr.3: 52436220-52436220 on Build GRCh38
rs780501064 Chr.3: 52436049-52436051 on Build GRCh38
rs771698538 Chr.3: 52436099-52436099 on Build GRCh38
rs572086601 Chr.3: 52435979-52435979 on Build GRCh38
rs772056522 Chr.3: 52436066-52436066 on Build GRCh38
rs200142915 Chr.3: 52436003-52436003 on Build GRCh38
rs762144595 Chr.3: 52436289-52436289 on Build GRCh38
rs111410797 Chr.3: 52436141-52436141 on Build GRCh38
rs747862589 Chr.3: 52436095-52436095 on Build GRCh38
rs755223081 Chr.3: 52435951-52435951 on Build GRCh38
rs967798793 Chr.3: 52435872-52435872 on Build GRCh38
rs138549917 Chr.3: 52435873-52435873 on Build GRCh38
rs369858134 Chr.3: 52436040-52436040 on Build GRCh38
rs533040208 Chr.3: 52436154-52436154 on Build GRCh38
rs758884166 Chr.3: 52436024-52436024 on Build GRCh38
rs115134928 Chr.3: 52436065-52436065 on Build GRCh38
rs185476879 Chr.3: 52435940-52435940 on Build GRCh38
rs982779922 Chr.3: 52436050-52436050 on Build GRCh38
rs572843419 Chr.3: 52436037-52436037 on Build GRCh38
rs141710411 Chr.3: 52436059-52436059 on Build GRCh38
rs755167126 Chr.3: 52436013-52436013 on Build GRCh38
rs182689922 Chr.3: 52436157-52436157 on Build GRCh38
rs746749617 Chr.3: 52436110-52436110 on Build GRCh38
rs151151732 Chr.3: 52436045-52436045 on Build GRCh38
rs892255744 Chr.3: 52436151-52436151 on Build GRCh38
rs748767828 Chr.3: 52436087-52436087 on Build GRCh38
rs752584880 Chr.3: 52435950-52435950 on Build GRCh38
rs549046685 Chr.3: 52436307-52436307 on Build GRCh38
rs142042017 Chr.3: 52436025-52436025 on Build GRCh38
rs139348169 Chr.3: 52435987-52435987 on Build GRCh38
rs564324191 Chr.3: 52436109-52436109 on Build GRCh38
rs149805640 Chr.3: 52435925-52435925 on Build GRCh38
rs1046803879 Chr.3: 52435972-52435972 on Build GRCh38
rs769191980 Chr.3: 52436241-52436241 on Build GRCh38
rs767461567 Chr.3: 52436008-52436008 on Build GRCh38
rs779388206 Chr.3: 52435868-52435868 on Build GRCh38
rs758627697 Chr.3: 52435853-52435854 on Build GRCh38
rs561801072 Chr.3: 52436044-52436044 on Build GRCh38
rs768949976 Chr.3: 52435866-52435866 on Build GRCh38
rs143787641 Chr.3: 52435898-52435898 on Build GRCh38
rs776680926 Chr.3: 52436124-52436124 on Build GRCh38
rs769509391 Chr.3: 52435879-52435879 on Build GRCh38
rs776607886 Chr.3: 52436047-52436047 on Build GRCh38
rs757773529 Chr.3: 52436026-52436026 on Build GRCh38
rs149754805 Chr.3: 52436001-52436001 on Build GRCh38
rs377268528 Chr.3: 52436039-52436039 on Build GRCh38
rs1050546735 Chr.3: 52435894-52435894 on Build GRCh38
rs534522733 Chr.3: 52436015-52436015 on Build GRCh38
rs370902272 Chr.3: 52436094-52436094 on Build GRCh38
rs750529472 Chr.3: 52435931-52435931 on Build GRCh38
rs773054985 Chr.3: 52436042-52436042 on Build GRCh38
rs1055485857 Chr.3: 52436145-52436145 on Build GRCh38
rs958576111 Chr.3: 52435865-52435865 on Build GRCh38
rs903062564 Chr.3: 52436196-52436196 on Build GRCh38
rs1033622827 Chr.3: 52436245-52436245 on Build GRCh38
rs936884639 Chr.3: 52436137-52436137 on Build GRCh38
rs929655488 Chr.3: 52436082-52436082 on Build GRCh38
rs780159666 Chr.3: 52435949-52435949 on Build GRCh38
rs778376244 Chr.3: 52435993-52435993 on Build GRCh38
rs370282481 Chr.3: 52435892-52435892 on Build GRCh38
rs936182262 Chr.3: 52435886-52435886 on Build GRCh38
rs766435587 Chr.3: 52436012-52436012 on Build GRCh38
rs1024897058 Chr.3: 52436014-52436014 on Build GRCh38
rs35027771 Chr.3: 52436162-52436163 on Build GRCh38
rs776959647 Chr.3: 52436284-52436284 on Build GRCh38
rs992710184 Chr.3: 52435867-52435867 on Build GRCh38
rs763946717 Chr.3: 52435897-52435897 on Build GRCh38
rs1014078000 Chr.3: 52436162-52436162 on Build GRCh38
rs751897478 Chr.3: 52436072-52436072 on Build GRCh38
rs748459524 Chr.3: 52435983-52435983 on Build GRCh38
rs200816034 Chr.3: 52436062-52436062 on Build GRCh38
rs769231842 Chr.3: 52435997-52435997 on Build GRCh38
rs780489027 Chr.3: 52436111-52436111 on Build GRCh38
rs1034226112 Chr.3: 52435863-52435863 on Build GRCh38
rs560280805 Chr.3: 52436169-52436169 on Build GRCh38
rs974549370 Chr.3: 52436034-52436034 on Build GRCh38
rs919890825 Chr.3: 52435880-52435880 on Build GRCh38
rs201353450 Chr.3: 52435990-52435990 on Build GRCh38
rs145809970 Chr.3: 52436058-52436058 on Build GRCh38
rs748729663 Chr.3: 52435869-52435869 on Build GRCh38
rs944200880 Chr.3: 52435930-52435930 on Build GRCh38
rs761552999 Chr.3: 52436007-52436007 on Build GRCh38
rs199738568 Chr.3: 52435871-52435871 on Build GRCh38
rs1024225883 Chr.3: 52436184-52436184 on Build GRCh38
rs558407652 Chr.3: 52436002-52436002 on Build GRCh38
rs761755344 Chr.3: 52435917-52435917 on Build GRCh38
rs775064868 Chr.3: 52435999-52435999 on Build GRCh38
rs963524841 Chr.3: 52436246-52436246 on Build GRCh38
rs145646348 Chr.3: 52436017-52436017 on Build GRCh38
rs749681098 Chr.3: 52435858-52435858 on Build GRCh38
rs376737486 Chr.3: 52436083-52436083 on Build GRCh38
rs773067139 Chr.3: 52436010-52436010 on Build GRCh38

View Full Product Details