Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108154845-108155378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A24
Entrez Gene ID
29957
Gene Name
solute carrier family 25 member 24
Gene Aliases
APC1, SCAMC-1
UniGene
Hs.656870
Gene Chromosome Location
Chr.1: 108134722-108200358 on Build GRCh38
Transcripts
NM_013386.4 NM_213651.2 XM_017001104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6507582 c.810G>T p.W270C Chr.1: 108154995-108154995 on Build GRCh38
7316159 c.686C>G p.S229* Chr.1: 108155119-108155119 on Build GRCh38
7316160 c.629C>G p.S210* Chr.1: 108155119-108155119 on Build GRCh38
4386031 c.699G>T p.R233S Chr.1: 108155049-108155049 on Build GRCh38
5842949 c.742C>T p.R248C Chr.1: 108155063-108155063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750575976 Chr.1: 108155173-108155173 on Build GRCh38
rs75988329 Chr.1: 108154875-108154875 on Build GRCh38
rs1019335357 Chr.1: 108155269-108155269 on Build GRCh38
rs953059981 Chr.1: 108155333-108155333 on Build GRCh38
rs773534747 Chr.1: 108155175-108155180 on Build GRCh38
rs772717897 Chr.1: 108155103-108155103 on Build GRCh38
rs937129710 Chr.1: 108155168-108155168 on Build GRCh38
rs991574131 Chr.1: 108155114-108155114 on Build GRCh38
rs770168846 Chr.1: 108155073-108155077 on Build GRCh38
rs745354492 Chr.1: 108155073-108155073 on Build GRCh38
rs781269573 Chr.1: 108155051-108155051 on Build GRCh38
rs768975202 Chr.1: 108154958-108154958 on Build GRCh38
rs762832650 Chr.1: 108155158-108155158 on Build GRCh38
rs374820142 Chr.1: 108155017-108155017 on Build GRCh38
rs1048995680 Chr.1: 108155207-108155207 on Build GRCh38
rs777816933 Chr.1: 108154900-108154900 on Build GRCh38
rs376441576 Chr.1: 108155159-108155159 on Build GRCh38
rs937451500 Chr.1: 108154882-108154882 on Build GRCh38
rs567851928 Chr.1: 108155197-108155197 on Build GRCh38
rs769099966 Chr.1: 108155084-108155084 on Build GRCh38
rs544533376 Chr.1: 108154967-108154967 on Build GRCh38
rs150730278 Chr.1: 108155034-108155034 on Build GRCh38
rs530981056 Chr.1: 108155130-108155130 on Build GRCh38
rs919198878 Chr.1: 108155185-108155185 on Build GRCh38
rs780408418 Chr.1: 108154941-108154941 on Build GRCh38
rs754013981 Chr.1: 108155046-108155046 on Build GRCh38
rs888765978 Chr.1: 108154992-108154992 on Build GRCh38
rs771653438 Chr.1: 108154970-108154970 on Build GRCh38
rs1048734028 Chr.1: 108154982-108154982 on Build GRCh38
rs930570550 Chr.1: 108155203-108155203 on Build GRCh38
rs779262141 Chr.1: 108155089-108155089 on Build GRCh38
rs78616848 Chr.1: 108154934-108154934 on Build GRCh38
rs373291702 Chr.1: 108155129-108155129 on Build GRCh38
rs1034717631 Chr.1: 108154898-108154898 on Build GRCh38
rs1007343871 Chr.1: 108155033-108155033 on Build GRCh38
rs191042201 Chr.1: 108155334-108155334 on Build GRCh38
rs534341793 Chr.1: 108155281-108155281 on Build GRCh38
rs748185865 Chr.1: 108155066-108155067 on Build GRCh38
rs756120780 Chr.1: 108155059-108155059 on Build GRCh38
rs373695269 Chr.1: 108155151-108155151 on Build GRCh38
rs749707615 Chr.1: 108154953-108154953 on Build GRCh38
rs1000441653 Chr.1: 108155355-108155355 on Build GRCh38
rs907044439 Chr.1: 108155154-108155154 on Build GRCh38
rs760099162 Chr.1: 108154978-108154978 on Build GRCh38
rs369827745 Chr.1: 108155109-108155109 on Build GRCh38
rs772782322 Chr.1: 108154973-108154973 on Build GRCh38
rs770380638 Chr.1: 108155124-108155124 on Build GRCh38
rs147531188 Chr.1: 108155090-108155090 on Build GRCh38
rs17021146 Chr.1: 108155278-108155278 on Build GRCh38
rs910252997 Chr.1: 108155222-108155222 on Build GRCh38
rs372564561 Chr.1: 108154967-108154967 on Build GRCh38
rs529189711 Chr.1: 108154906-108154906 on Build GRCh38
rs752609677 Chr.1: 108155155-108155155 on Build GRCh38
rs1040122805 Chr.1: 108155233-108155233 on Build GRCh38
rs200993204 Chr.1: 108155063-108155063 on Build GRCh38
rs893012738 Chr.1: 108155217-108155217 on Build GRCh38
rs1011623420 Chr.1: 108154948-108154948 on Build GRCh38
rs771778301 Chr.1: 108155091-108155091 on Build GRCh38
rs55861360 Chr.1: 108155240-108155240 on Build GRCh38
rs753688650 Chr.1: 108155011-108155011 on Build GRCh38
rs750578672 Chr.1: 108155058-108155058 on Build GRCh38
rs931596898 Chr.1: 108155104-108155104 on Build GRCh38
rs7555964 Chr.1: 108155223-108155223 on Build GRCh38
rs143348106 Chr.1: 108155062-108155062 on Build GRCh38
rs1001453680 Chr.1: 108154886-108154886 on Build GRCh38
rs978966222 Chr.1: 108155069-108155069 on Build GRCh38
rs559724952 Chr.1: 108154989-108154989 on Build GRCh38
rs368649872 Chr.1: 108154966-108154966 on Build GRCh38
rs991523465 Chr.1: 108154899-108154899 on Build GRCh38
rs149634691 Chr.1: 108155294-108155294 on Build GRCh38

View Full Product Details