Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109317662-109318158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SORT1
Entrez Gene ID
6272
Gene Name
sortilin 1
Gene Aliases
Gp95, LDLCQ6, NT3, NTR3
UniGene
Hs.485195
Gene Chromosome Location
Chr.1: 109309565-109397945 on Build GRCh38
Transcripts
NM_001205228.1 NM_002959.6 XM_005271100.2 XM_005271101.2 XM_005271102.1 XM_006710812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6992604 c.2056T>G p.S686A Chr.1: 109317938-109317938 on Build GRCh38
6120116 c.2122G>C p.E708Q Chr.1: 109317872-109317872 on Build GRCh38
4569854 c.2102T>A p.F701Y Chr.1: 109317892-109317892 on Build GRCh38
5968722 c.2075C>G p.P692R Chr.1: 109317919-109317919 on Build GRCh38
7147297 c.2092G>T p.D698Y Chr.1: 109317902-109317902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538506736 Chr.1: 109318077-109318077 on Build GRCh38
rs994783665 Chr.1: 109317794-109317794 on Build GRCh38
rs746392929 Chr.1: 109317883-109317883 on Build GRCh38
rs897312339 Chr.1: 109317741-109317741 on Build GRCh38
rs923729189 Chr.1: 109317955-109317955 on Build GRCh38
rs781038788 Chr.1: 109317815-109317815 on Build GRCh38
rs776130114 Chr.1: 109318005-109318005 on Build GRCh38
rs1056650635 Chr.1: 109317749-109317749 on Build GRCh38
rs1040203447 Chr.1: 109317740-109317740 on Build GRCh38
rs1038493888 Chr.1: 109317779-109317779 on Build GRCh38
rs1022565865 Chr.1: 109317835-109317835 on Build GRCh38
rs1031545324 Chr.1: 109317972-109317972 on Build GRCh38
rs961128998 Chr.1: 109317862-109317862 on Build GRCh38
rs371846577 Chr.1: 109317810-109317810 on Build GRCh38
rs761536586 Chr.1: 109317917-109317917 on Build GRCh38
rs762049797 Chr.1: 109318017-109318017 on Build GRCh38
rs769552486 Chr.1: 109317700-109317701 on Build GRCh38
rs72646581 Chr.1: 109317800-109317800 on Build GRCh38
rs376283085 Chr.1: 109318016-109318016 on Build GRCh38
rs889808974 Chr.1: 109317756-109317756 on Build GRCh38
rs545224556 Chr.1: 109317685-109317685 on Build GRCh38
rs770374728 Chr.1: 109317897-109317897 on Build GRCh38
rs750330704 Chr.1: 109317950-109317950 on Build GRCh38
rs775409345 Chr.1: 109317899-109317899 on Build GRCh38
rs999827457 Chr.1: 109318125-109318125 on Build GRCh38
rs771436530 Chr.1: 109317878-109317878 on Build GRCh38
rs897226595 Chr.1: 109317781-109317781 on Build GRCh38
rs941489744 Chr.1: 109318023-109318023 on Build GRCh38
rs774559468 Chr.1: 109317906-109317906 on Build GRCh38
rs778776913 Chr.1: 109317866-109317866 on Build GRCh38
rs755597688 Chr.1: 109317825-109317825 on Build GRCh38
rs548333462 Chr.1: 109318009-109318009 on Build GRCh38
rs895424129 Chr.1: 109318096-109318096 on Build GRCh38
rs909114705 Chr.1: 109318042-109318042 on Build GRCh38
rs918643612 Chr.1: 109318053-109318053 on Build GRCh38
rs370017732 Chr.1: 109317903-109317903 on Build GRCh38
rs529657236 Chr.1: 109317882-109317882 on Build GRCh38
rs1023820714 Chr.1: 109317895-109317895 on Build GRCh38
rs1044100675 Chr.1: 109318117-109318117 on Build GRCh38
rs1005525770 Chr.1: 109317758-109317758 on Build GRCh38
rs776757996 Chr.1: 109317804-109317804 on Build GRCh38
rs747313158 Chr.1: 109317875-109317875 on Build GRCh38
rs767234330 Chr.1: 109317939-109317939 on Build GRCh38
rs534188785 Chr.1: 109318127-109318127 on Build GRCh38
rs1031847828 Chr.1: 109318126-109318126 on Build GRCh38
rs184219286 Chr.1: 109317870-109317870 on Build GRCh38
rs376624400 Chr.1: 109318039-109318039 on Build GRCh38
rs878870232 Chr.1: 109318064-109318064 on Build GRCh38
rs1000343259 Chr.1: 109317946-109317946 on Build GRCh38
rs747827454 Chr.1: 109317988-109317988 on Build GRCh38
rs554983678 Chr.1: 109318080-109318080 on Build GRCh38
rs904182597 Chr.1: 109318120-109318120 on Build GRCh38
rs941366971 Chr.1: 109318052-109318052 on Build GRCh38
rs748881374 Chr.1: 109317842-109317842 on Build GRCh38
rs753396170 Chr.1: 109317981-109317981 on Build GRCh38
rs372630842 Chr.1: 109317976-109317976 on Build GRCh38
rs761669018 Chr.1: 109317870-109317872 on Build GRCh38
rs574737101 Chr.1: 109318092-109318092 on Build GRCh38
rs772291182 Chr.1: 109317731-109317731 on Build GRCh38
rs530818700 Chr.1: 109317818-109317818 on Build GRCh38
rs191308023 Chr.1: 109317987-109317987 on Build GRCh38
rs754667912 Chr.1: 109317846-109317846 on Build GRCh38
rs973230956 Chr.1: 109318026-109318026 on Build GRCh38
rs369638121 Chr.1: 109317995-109317995 on Build GRCh38
rs755987119 Chr.1: 109317952-109317952 on Build GRCh38
rs757656184 Chr.1: 109317997-109317997 on Build GRCh38
rs948291252 Chr.1: 109318097-109318097 on Build GRCh38
rs569382221 Chr.1: 109318041-109318041 on Build GRCh38
rs372874714 Chr.1: 109317999-109317999 on Build GRCh38
rs375586089 Chr.1: 109317982-109317982 on Build GRCh38
rs770188286 Chr.1: 109318004-109318004 on Build GRCh38
rs187592600 Chr.1: 109317872-109317872 on Build GRCh38
rs780309284 Chr.1: 109317742-109317742 on Build GRCh38
rs1050731107 Chr.1: 109317750-109317750 on Build GRCh38
rs865939290 Chr.1: 109317887-109317887 on Build GRCh38
rs745593507 Chr.1: 109317705-109317705 on Build GRCh38
rs751340689 Chr.1: 109317809-109317809 on Build GRCh38
rs376554239 Chr.1: 109317826-109317826 on Build GRCh38
rs897278873 Chr.1: 109318057-109318057 on Build GRCh38
rs546247342 Chr.1: 109317902-109317902 on Build GRCh38
rs749297798 Chr.1: 109318007-109318007 on Build GRCh38

View Full Product Details