Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58867738-58868252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf67
Entrez Gene ID
200844
Gene Name
chromosome 3 open reading frame 67
Gene Aliases
-
UniGene
Hs.368434
Gene Chromosome Location
Chr.3: 58716415-59050078 on Build GRCh38
Transcripts
NM_198463.2 XM_005264929.2 XM_006713013.3 XM_011533451.2 XM_011533453.2 XM_011533455.2 XM_011533456.1 XM_011533457.1 XM_011533458.2 XM_011533459.2 XM_011533460.2 XM_011533461.1 XM_011533463.2 XM_017005848.1 XM_017005849.1 XM_017005850.1 XM_017005851.1 NR_110820.1

Gene Symbol
C3orf67-AS1
Entrez Gene ID
101929238
Gene Name
C3orf67 antisense RNA 1
Gene Aliases
-
UniGene
Hs.683953
Gene Chromosome Location
Chr.3: 58824471-59019093 on Build GRCh38
Transcripts
NM_198463.2 XM_005264929.2 XM_006713013.3 XM_011533451.2 XM_011533453.2 XM_011533455.2 XM_011533456.1 XM_011533457.1 XM_011533458.2 XM_011533459.2 XM_011533460.2 XM_011533461.1 XM_011533463.2 XM_017005848.1 XM_017005849.1 XM_017005850.1 XM_017005851.1 NR_110820.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3596893 c.720G>A p.R240R Chr.3: 58867857-58867857 on Build GRCh38
4337533 c.727delA p.S243fs*39 Chr.3: 58867850-58867850 on Build GRCh38
4787773 c.745A>G p.T249A Chr.3: 58867832-58867832 on Build GRCh38
5686146 c.666G>T p.P222P Chr.3: 58867911-58867911 on Build GRCh38
4326592 c.647A>G p.N216S Chr.3: 58867930-58867930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951720468 Chr.3: 58868193-58868193 on Build GRCh38
rs545080053 Chr.3: 58868122-58868122 on Build GRCh38
rs879016659 Chr.3: 58867934-58867934 on Build GRCh38
rs776779350 Chr.3: 58867853-58867853 on Build GRCh38
rs75606030 Chr.3: 58867988-58867988 on Build GRCh38
rs199595225 Chr.3: 58867848-58867848 on Build GRCh38
rs1049342795 Chr.3: 58867861-58867861 on Build GRCh38
rs940584280 Chr.3: 58867897-58867897 on Build GRCh38
rs565431731 Chr.3: 58867834-58867834 on Build GRCh38
rs889170090 Chr.3: 58868063-58868063 on Build GRCh38
rs191456822 Chr.3: 58867951-58867951 on Build GRCh38
rs751464849 Chr.3: 58867884-58867884 on Build GRCh38
rs776930506 Chr.3: 58867858-58867858 on Build GRCh38
rs199964995 Chr.3: 58867970-58867970 on Build GRCh38
rs748140208 Chr.3: 58867994-58867994 on Build GRCh38
rs772490363 Chr.3: 58867863-58867865 on Build GRCh38
rs776648816 Chr.3: 58867958-58867958 on Build GRCh38
rs772549901 Chr.3: 58867948-58867948 on Build GRCh38
rs28522900 Chr.3: 58868108-58868108 on Build GRCh38
rs747008118 Chr.3: 58867950-58867950 on Build GRCh38
rs972100964 Chr.3: 58867793-58867793 on Build GRCh38
rs183754764 Chr.3: 58867792-58867792 on Build GRCh38
rs1023872868 Chr.3: 58868060-58868060 on Build GRCh38
rs546596755 Chr.3: 58867818-58867818 on Build GRCh38
rs920239394 Chr.3: 58868159-58868159 on Build GRCh38
rs772709211 Chr.3: 58867903-58867903 on Build GRCh38
rs944570410 Chr.3: 58867857-58867857 on Build GRCh38
rs774264787 Chr.3: 58867973-58867973 on Build GRCh38
rs766576605 Chr.3: 58867844-58867844 on Build GRCh38
rs755234359 Chr.3: 58867930-58867930 on Build GRCh38
rs9852739 Chr.3: 58868109-58868109 on Build GRCh38
rs779796914 Chr.3: 58867814-58867814 on Build GRCh38
rs774784196 Chr.3: 58867825-58867825 on Build GRCh38
rs780843258 Chr.3: 58867796-58867796 on Build GRCh38
rs187794466 Chr.3: 58867949-58867949 on Build GRCh38
rs774736533 Chr.3: 58867775-58867782 on Build GRCh38
rs202187076 Chr.3: 58867986-58867986 on Build GRCh38
rs141866639 Chr.3: 58868217-58868217 on Build GRCh38
rs962064845 Chr.3: 58868147-58868147 on Build GRCh38
rs1001048317 Chr.3: 58868082-58868082 on Build GRCh38
rs772635829 Chr.3: 58867833-58867833 on Build GRCh38
rs762594054 Chr.3: 58867859-58867859 on Build GRCh38
rs540988078 Chr.3: 58868107-58868107 on Build GRCh38
rs368243575 Chr.3: 58867939-58867939 on Build GRCh38
rs1035197950 Chr.3: 58868123-58868123 on Build GRCh38
rs969577384 Chr.3: 58868061-58868061 on Build GRCh38
rs145324023 Chr.3: 58867852-58867852 on Build GRCh38
rs530588619 Chr.3: 58868194-58868194 on Build GRCh38
rs757245451 Chr.3: 58867887-58867887 on Build GRCh38
rs756759335 Chr.3: 58867790-58867790 on Build GRCh38
rs746167323 Chr.3: 58867818-58867841 on Build GRCh38
rs988345285 Chr.3: 58868189-58868189 on Build GRCh38
rs760532998 Chr.3: 58867800-58867800 on Build GRCh38
rs763951309 Chr.3: 58867867-58867867 on Build GRCh38
rs767611378 Chr.3: 58867889-58867889 on Build GRCh38
rs370413637 Chr.3: 58867771-58867771 on Build GRCh38
rs201674194 Chr.3: 58867767-58867767 on Build GRCh38
rs141916956 Chr.3: 58867912-58867912 on Build GRCh38
rs745428162 Chr.3: 58867798-58867798 on Build GRCh38
rs961733735 Chr.3: 58867774-58867774 on Build GRCh38
rs775107395 Chr.3: 58867871-58867873 on Build GRCh38
rs182285671 Chr.3: 58868032-58868032 on Build GRCh38
rs1019936993 Chr.3: 58868212-58868212 on Build GRCh38
rs143379870 Chr.3: 58867911-58867911 on Build GRCh38
rs887695512 Chr.3: 58867878-58867878 on Build GRCh38
rs971071485 Chr.3: 58868220-58868220 on Build GRCh38
rs755681592 Chr.3: 58867904-58867904 on Build GRCh38
rs762315869 Chr.3: 58867826-58867826 on Build GRCh38
rs971825078 Chr.3: 58868156-58868156 on Build GRCh38
rs6790436 Chr.3: 58868021-58868021 on Build GRCh38
rs563276729 Chr.3: 58868192-58868192 on Build GRCh38

View Full Product Details