Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 75737629-75738145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF717
Entrez Gene ID
100131827
Gene Name
zinc finger protein 717
Gene Aliases
X17, ZNF838
UniGene
Hs.556877
Gene Chromosome Location
Chr.3: 75729961-75785583 on Build GRCh38
Transcripts
NM_001128223.2 NM_001290208.1 NM_001290209.1 NM_001290210.1 NM_001324026.1 NM_001324027.1 NM_001324028.1 XM_005264711.3 XM_006712912.3 XM_011533244.2 XM_011533245.2 XM_011533246.2 XM_011533248.2 XM_011533249.2 XM_011533250.2 XM_011533251.1 XM_017005486.1 XM_017005487.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4158466 c.1505C>T p.T502I Chr.3: 75738118-75738118 on Build GRCh38
5479989 c.1788G>A p.K596K Chr.3: 75737814-75737814 on Build GRCh38
3746643 c.1622G>A p.G541E Chr.3: 75738001-75738001 on Build GRCh38
4327492 c.1626C>A p.Y542* Chr.3: 75737976-75737976 on Build GRCh38
4158457 c.1559G>A p.R520K Chr.3: 75738043-75738043 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs73843012 Chr.3: 75738035-75738035 on Build GRCh38
rs755692982 Chr.3: 75737910-75737910 on Build GRCh38
rs752455744 Chr.3: 75737686-75737686 on Build GRCh38
rs1045936818 Chr.3: 75737896-75737896 on Build GRCh38
rs62250107 Chr.3: 75737906-75737906 on Build GRCh38
rs796390112 Chr.3: 75737782-75737782 on Build GRCh38
rs2918517 Chr.3: 75737791-75737791 on Build GRCh38
rs78906544 Chr.3: 75737765-75737765 on Build GRCh38
rs758183144 Chr.3: 75737764-75737764 on Build GRCh38
rs200945611 Chr.3: 75738023-75738023 on Build GRCh38
rs748519426 Chr.3: 75738013-75738013 on Build GRCh38
rs370022573 Chr.3: 75738074-75738074 on Build GRCh38
rs866543533 Chr.3: 75737960-75737960 on Build GRCh38
rs768920912 Chr.3: 75737834-75737834 on Build GRCh38
rs73843010 Chr.3: 75738015-75738015 on Build GRCh38
rs746594210 Chr.3: 75737704-75737704 on Build GRCh38
rs374047309 Chr.3: 75737731-75737731 on Build GRCh38
rs748885031 Chr.3: 75737819-75737819 on Build GRCh38
rs201263569 Chr.3: 75737667-75737675 on Build GRCh38
rs62250106 Chr.3: 75737898-75737898 on Build GRCh38
rs767520430 Chr.3: 75737744-75737744 on Build GRCh38
rs770035668 Chr.3: 75738116-75738116 on Build GRCh38
rs751464511 Chr.3: 75737769-75737769 on Build GRCh38
rs144168413 Chr.3: 75737744-75737744 on Build GRCh38
rs77829652 Chr.3: 75738114-75738114 on Build GRCh38
rs80019510 Chr.3: 75738113-75738113 on Build GRCh38
rs78806516 Chr.3: 75737892-75737892 on Build GRCh38
rs533798223 Chr.3: 75737899-75737899 on Build GRCh38
rs796686632 Chr.3: 75738071-75738073 on Build GRCh38
rs763817112 Chr.3: 75737883-75737883 on Build GRCh38
rs79550710 Chr.3: 75738084-75738084 on Build GRCh38
rs751722073 Chr.3: 75737664-75737664 on Build GRCh38
rs76263679 Chr.3: 75737864-75737864 on Build GRCh38
rs79516801 Chr.3: 75737715-75737715 on Build GRCh38
rs753571758 Chr.3: 75737984-75737984 on Build GRCh38
rs62250105 Chr.3: 75737694-75737694 on Build GRCh38
rs773299358 Chr.3: 75737747-75737747 on Build GRCh38
rs201606605 Chr.3: 75738067-75738067 on Build GRCh38
rs770854670 Chr.3: 75737697-75737697 on Build GRCh38
rs868115346 Chr.3: 75737733-75737733 on Build GRCh38
rs1615736 Chr.3: 75737737-75737737 on Build GRCh38
rs769540917 Chr.3: 75737718-75737718 on Build GRCh38
rs780694948 Chr.3: 75737709-75737709 on Build GRCh38
rs77490669 Chr.3: 75737965-75737965 on Build GRCh38
rs202060190 Chr.3: 75738043-75738043 on Build GRCh38
rs76496075 Chr.3: 75737840-75737840 on Build GRCh38
rs764447978 Chr.3: 75737856-75737856 on Build GRCh38
rs78806516 Chr.3: 75737892-75737892 on Build GRCh38
rs74340838 Chr.3: 75737948-75737949 on Build GRCh38
rs757180235 Chr.3: 75737770-75737770 on Build GRCh38
rs112847134 Chr.3: 75738073-75738073 on Build GRCh38
rs77322475 Chr.3: 75738008-75738008 on Build GRCh38
rs772607103 Chr.3: 75737780-75737781 on Build GRCh38
rs774731924 Chr.3: 75737835-75737835 on Build GRCh38
rs778098228 Chr.3: 75737694-75737694 on Build GRCh38
rs761797378 Chr.3: 75737671-75737671 on Build GRCh38
rs748630837 Chr.3: 75737838-75737838 on Build GRCh38
rs774543835 Chr.3: 75737742-75737742 on Build GRCh38
rs754845657 Chr.3: 75737659-75737660 on Build GRCh38
rs533309906 Chr.3: 75737730-75737730 on Build GRCh38
rs79982156 Chr.3: 75738057-75738057 on Build GRCh38
rs73843011 Chr.3: 75738018-75738018 on Build GRCh38
rs79560192 Chr.3: 75737994-75737994 on Build GRCh38
rs200125588 Chr.3: 75737990-75737990 on Build GRCh38
rs79635065 Chr.3: 75737676-75737676 on Build GRCh38
rs550801385 Chr.3: 75737673-75737673 on Build GRCh38
rs77378861 Chr.3: 75737930-75737930 on Build GRCh38
rs774073278 Chr.3: 75738118-75738118 on Build GRCh38
rs138214171 Chr.3: 75738048-75738048 on Build GRCh38
rs76111663 Chr.3: 75738089-75738089 on Build GRCh38
rs78763958 Chr.3: 75738077-75738077 on Build GRCh38
rs773327801 Chr.3: 75737851-75737851 on Build GRCh38
rs764677392 Chr.3: 75737659-75737659 on Build GRCh38
rs567032467 Chr.3: 75738064-75738064 on Build GRCh38
rs573869621 Chr.3: 75737855-75737856 on Build GRCh38
rs796543994 Chr.3: 75738113-75738114 on Build GRCh38
rs76729736 Chr.3: 75737881-75737881 on Build GRCh38
rs780608527 Chr.3: 75737773-75737773 on Build GRCh38
rs77322475 Chr.3: 75738008-75738008 on Build GRCh38
rs752543553 Chr.3: 75737761-75737761 on Build GRCh38
rs78828372 Chr.3: 75737748-75737748 on Build GRCh38
rs760727342 Chr.3: 75737857-75737857 on Build GRCh38
rs76347222 Chr.3: 75737814-75737814 on Build GRCh38
rs370463977 Chr.3: 75738093-75738093 on Build GRCh38
rs796859243 Chr.3: 75737896-75737897 on Build GRCh38
rs776841176 Chr.3: 75737875-75737875 on Build GRCh38
rs78680336 Chr.3: 75738071-75738071 on Build GRCh38
rs763597454 Chr.3: 75737657-75737657 on Build GRCh38
rs755889773 Chr.3: 75737801-75737801 on Build GRCh38
rs11122676 Chr.3: 75737681-75737681 on Build GRCh38
rs867694150 Chr.3: 75737854-75737855 on Build GRCh38
rs373869907 Chr.3: 75738102-75738102 on Build GRCh38
rs76911771 Chr.3: 75738108-75738108 on Build GRCh38
rs557324871 Chr.3: 75737697-75737698 on Build GRCh38
rs74740396 Chr.3: 75737976-75737976 on Build GRCh38
rs369928577 Chr.3: 75737891-75737892 on Build GRCh38
rs779655396 Chr.3: 75737912-75737912 on Build GRCh38
rs924792891 Chr.3: 75738068-75738068 on Build GRCh38
rs76263679 Chr.3: 75737864-75737864 on Build GRCh38
rs200631700 Chr.3: 75737759-75737759 on Build GRCh38
rs776047719 Chr.3: 75737719-75737719 on Build GRCh38
rs139448820 Chr.3: 75737682-75737682 on Build GRCh38
rs75063165 Chr.3: 75738053-75738053 on Build GRCh38
rs745352079 Chr.3: 75737800-75737800 on Build GRCh38
rs770351960 Chr.3: 75737708-75737708 on Build GRCh38
rs113151880 Chr.3: 75738085-75738085 on Build GRCh38
rs75032682 Chr.3: 75737666-75737666 on Build GRCh38
rs149887072 Chr.3: 75737842-75737842 on Build GRCh38
rs776210532 Chr.3: 75738116-75738117 on Build GRCh38
rs57577072 Chr.3: 75737671-75737679 on Build GRCh38
rs750627087 Chr.3: 75737675-75737675 on Build GRCh38
rs774728033 Chr.3: 75737734-75737734 on Build GRCh38
rs775808698 Chr.3: 75737794-75737795 on Build GRCh38
rs76826286 Chr.3: 75737741-75737741 on Build GRCh38
rs79089658 Chr.3: 75738070-75738070 on Build GRCh38
rs76921234 Chr.3: 75737703-75737703 on Build GRCh38
rs13314309 Chr.3: 75737803-75737803 on Build GRCh38
rs759746427 Chr.3: 75737876-75737876 on Build GRCh38
rs779851479 Chr.3: 75737808-75737808 on Build GRCh38
rs77101176 Chr.3: 75737768-75737768 on Build GRCh38
rs76713735 Chr.3: 75738051-75738051 on Build GRCh38
rs772596336 Chr.3: 75737845-75737845 on Build GRCh38
rs745547929 Chr.3: 75737713-75737713 on Build GRCh38
rs373780316 Chr.3: 75737893-75737894 on Build GRCh38
rs921548486 Chr.3: 75737832-75737832 on Build GRCh38

View Full Product Details