Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 167687439-167687960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDCD10
Entrez Gene ID
11235
Gene Name
programmed cell death 10
Gene Aliases
CCM3, TFAR15
UniGene
Hs.478150
Gene Chromosome Location
Chr.3: 167683891-167735690 on Build GRCh38
Transcripts
NM_007217.3 NM_145859.1 NM_145860.1 XM_005247086.4 XM_005247087.4 XM_005247088.3 XM_006713485.3 XM_011512368.2 XM_011512369.2 XM_017005644.1 XM_017005645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6899241 c.433G>A p.V145M Chr.3: 167687656-167687656 on Build GRCh38
5665997 c.396G>T p.K132N Chr.3: 167687693-167687693 on Build GRCh38
40445 c.396G>A p.K132K Chr.3: 167687693-167687693 on Build GRCh38
1219904 c.418G>T p.E140* Chr.3: 167687671-167687671 on Build GRCh38
4333452 c.396-1G>T p.? Chr.3: 167687694-167687694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886041888 Chr.3: 167687621-167687622 on Build GRCh38
rs189238762 Chr.3: 167687741-167687741 on Build GRCh38
rs757890205 Chr.3: 167687715-167687715 on Build GRCh38
rs1019961358 Chr.3: 167687769-167687769 on Build GRCh38
rs751245377 Chr.3: 167687723-167687723 on Build GRCh38
rs561395243 Chr.3: 167687773-167687773 on Build GRCh38
rs375997950 Chr.3: 167687564-167687564 on Build GRCh38
rs760436661 Chr.3: 167687648-167687648 on Build GRCh38
rs909147593 Chr.3: 167687855-167687855 on Build GRCh38
rs755309660 Chr.3: 167687570-167687570 on Build GRCh38
rs367943169 Chr.3: 167687630-167687630 on Build GRCh38
rs748452548 Chr.3: 167687736-167687736 on Build GRCh38
rs534305257 Chr.3: 167687717-167687755 on Build GRCh38
rs984714966 Chr.3: 167687839-167687839 on Build GRCh38
rs965773573 Chr.3: 167687776-167687776 on Build GRCh38
rs573388501 Chr.3: 167687462-167687462 on Build GRCh38
rs551036976 Chr.3: 167687871-167687871 on Build GRCh38
rs528548367 Chr.3: 167687778-167687778 on Build GRCh38
rs566904958 Chr.3: 167687825-167687825 on Build GRCh38
rs1027643458 Chr.3: 167687529-167687529 on Build GRCh38
rs771126809 Chr.3: 167687619-167687619 on Build GRCh38
rs754584968 Chr.3: 167687727-167687727 on Build GRCh38
rs748685407 Chr.3: 167687581-167687581 on Build GRCh38
rs528131910 Chr.3: 167687812-167687812 on Build GRCh38
rs747369642 Chr.3: 167687565-167687565 on Build GRCh38
rs534849184 Chr.3: 167687491-167687491 on Build GRCh38
rs372343719 Chr.3: 167687866-167687866 on Build GRCh38
rs770216793 Chr.3: 167687583-167687583 on Build GRCh38
rs1001808798 Chr.3: 167687503-167687503 on Build GRCh38
rs776531423 Chr.3: 167687655-167687655 on Build GRCh38
rs778554068 Chr.3: 167687912-167687912 on Build GRCh38
rs781486214 Chr.3: 167687571-167687571 on Build GRCh38
rs778672630 Chr.3: 167687884-167687884 on Build GRCh38
rs543027351 Chr.3: 167687755-167687755 on Build GRCh38
rs891514632 Chr.3: 167687876-167687876 on Build GRCh38
rs975678958 Chr.3: 167687794-167687794 on Build GRCh38
rs749965663 Chr.3: 167687698-167687698 on Build GRCh38
rs761849146 Chr.3: 167687673-167687673 on Build GRCh38
rs976585767 Chr.3: 167687633-167687633 on Build GRCh38
rs116787813 Chr.3: 167687745-167687745 on Build GRCh38
rs765249687 Chr.3: 167687693-167687693 on Build GRCh38
rs765993541 Chr.3: 167687716-167687716 on Build GRCh38
rs952060772 Chr.3: 167687562-167687562 on Build GRCh38
rs185195114 Chr.3: 167687728-167687728 on Build GRCh38
rs1045889337 Chr.3: 167687882-167687882 on Build GRCh38
rs950115795 Chr.3: 167687880-167687880 on Build GRCh38
rs1033241006 Chr.3: 167687919-167687919 on Build GRCh38
rs200143822 Chr.3: 167687478-167687478 on Build GRCh38
rs985376581 Chr.3: 167687810-167687810 on Build GRCh38
rs370669838 Chr.3: 167687475-167687475 on Build GRCh38
rs367568906 Chr.3: 167687606-167687606 on Build GRCh38
rs1001965213 Chr.3: 167687902-167687902 on Build GRCh38
rs201857174 Chr.3: 167687650-167687650 on Build GRCh38
rs111922591 Chr.3: 167687783-167687783 on Build GRCh38
rs773292567 Chr.3: 167687603-167687603 on Build GRCh38
rs921597776 Chr.3: 167687818-167687818 on Build GRCh38
rs878990589 Chr.3: 167687865-167687865 on Build GRCh38
rs73173064 Chr.3: 167687826-167687826 on Build GRCh38
rs1024320562 Chr.3: 167687530-167687530 on Build GRCh38
rs780322739 Chr.3: 167687548-167687548 on Build GRCh38
rs940513596 Chr.3: 167687856-167687856 on Build GRCh38

View Full Product Details