Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 172750605-172751114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECT2
Entrez Gene ID
1894
Gene Name
epithelial cell transforming 2
Gene Aliases
ARHGEF31
UniGene
Hs.518299
Gene Chromosome Location
Chr.3: 172750682-172829273 on Build GRCh38
Transcripts
NM_001258315.1 NM_001258316.1 NM_018098.5 XM_005247176.1 XM_006713523.3 XM_006713524.3 XM_011512514.2 XM_011512515.2 XM_011512516.1 XM_017005830.1 XM_017005831.1 XM_017005832.1 XM_017005833.1 XM_017005834.1 XM_017005835.1 XM_017005836.1 XM_017005837.1 XM_017005838.1

SNP Detail

SNP ID Chromosome Location
rs960119122 Chr.3: 172751003-172751003 on Build GRCh38
rs1011494255 Chr.3: 172750717-172750717 on Build GRCh38
rs190106783 Chr.3: 172750664-172750664 on Build GRCh38
rs981218629 Chr.3: 172750755-172750755 on Build GRCh38
rs1003971799 Chr.3: 172750799-172750799 on Build GRCh38
rs150915278 Chr.3: 172750661-172750661 on Build GRCh38
rs1015733183 Chr.3: 172750905-172750905 on Build GRCh38
rs928493827 Chr.3: 172751000-172751000 on Build GRCh38
rs986768631 Chr.3: 172751058-172751058 on Build GRCh38
rs773724081 Chr.3: 172750643-172750643 on Build GRCh38
rs139251438 Chr.3: 172750867-172750867 on Build GRCh38
rs768500387 Chr.3: 172750657-172750657 on Build GRCh38
rs368942417 Chr.3: 172750957-172750957 on Build GRCh38
rs541640984 Chr.3: 172750892-172750892 on Build GRCh38
rs890611848 Chr.3: 172750895-172750895 on Build GRCh38
rs962837052 Chr.3: 172750965-172750965 on Build GRCh38
rs756021890 Chr.3: 172751059-172751059 on Build GRCh38
rs979942782 Chr.3: 172750796-172750796 on Build GRCh38
rs576868078 Chr.3: 172750768-172750768 on Build GRCh38
rs929990552 Chr.3: 172750826-172750826 on Build GRCh38
rs561731764 Chr.3: 172751021-172751021 on Build GRCh38
rs181812072 Chr.3: 172750841-172750841 on Build GRCh38
rs918615114 Chr.3: 172750819-172750819 on Build GRCh38
rs1018065349 Chr.3: 172750685-172750685 on Build GRCh38
rs941647091 Chr.3: 172750797-172750797 on Build GRCh38
rs919806394 Chr.3: 172750779-172750779 on Build GRCh38
rs932246183 Chr.3: 172751074-172751074 on Build GRCh38
rs913249674 Chr.3: 172750837-172750837 on Build GRCh38
rs995540223 Chr.3: 172750940-172750940 on Build GRCh38
rs549824755 Chr.3: 172751006-172751006 on Build GRCh38
rs958459708 Chr.3: 172750770-172750770 on Build GRCh38
rs947492408 Chr.3: 172751042-172751042 on Build GRCh38
rs1006940297 Chr.3: 172750864-172750864 on Build GRCh38
rs3830582 Chr.3: 172750683-172750683 on Build GRCh38
rs1007998140 Chr.3: 172750896-172750896 on Build GRCh38
rs938807286 Chr.3: 172750810-172750810 on Build GRCh38
rs891205823 Chr.3: 172750845-172750845 on Build GRCh38
rs969980666 Chr.3: 172750990-172750990 on Build GRCh38
rs35511310 Chr.3: 172750850-172750850 on Build GRCh38
rs916721207 Chr.3: 172750784-172750784 on Build GRCh38
rs1053594349 Chr.3: 172750815-172750815 on Build GRCh38
rs34511822 Chr.3: 172750960-172750961 on Build GRCh38
rs564381936 Chr.3: 172750949-172750949 on Build GRCh38
rs529019912 Chr.3: 172751080-172751080 on Build GRCh38
rs991246027 Chr.3: 172750942-172750942 on Build GRCh38
rs567039165 Chr.3: 172750663-172750663 on Build GRCh38
rs1020004185 Chr.3: 172750718-172750718 on Build GRCh38
rs1024246257 Chr.3: 172750944-172750944 on Build GRCh38
rs572246457 Chr.3: 172750919-172750919 on Build GRCh38
rs111679156 Chr.3: 172751040-172751040 on Build GRCh38
rs961313075 Chr.3: 172750748-172750748 on Build GRCh38
rs967019417 Chr.3: 172750753-172750753 on Build GRCh38
rs1008381391 Chr.3: 172750682-172750682 on Build GRCh38
rs546157932 Chr.3: 172750925-172750925 on Build GRCh38
rs1020202225 Chr.3: 172750894-172750894 on Build GRCh38
rs111316186 Chr.3: 172750858-172750858 on Build GRCh38
rs1007153363 Chr.3: 172750667-172750667 on Build GRCh38
rs779116926 Chr.3: 172750979-172750979 on Build GRCh38
rs918856151 Chr.3: 172751018-172751018 on Build GRCh38
rs558527519 Chr.3: 172750670-172750670 on Build GRCh38
rs922271862 Chr.3: 172750648-172750661 on Build GRCh38
rs531285537 Chr.3: 172750999-172750999 on Build GRCh38
rs5854482 Chr.3: 172750682-172750682 on Build GRCh38
rs568163995 Chr.3: 172750647-172750647 on Build GRCh38
rs958708315 Chr.3: 172750937-172750937 on Build GRCh38
rs913268630 Chr.3: 172751032-172751032 on Build GRCh38
rs996424221 Chr.3: 172750868-172750869 on Build GRCh38
rs892969475 Chr.3: 172750875-172750875 on Build GRCh38
rs1015286015 Chr.3: 172750703-172750703 on Build GRCh38
rs536418020 Chr.3: 172750649-172750649 on Build GRCh38
rs560322504 Chr.3: 172750906-172750906 on Build GRCh38
rs974028969 Chr.3: 172750751-172750751 on Build GRCh38
rs957522172 Chr.3: 172750688-172750688 on Build GRCh38
rs1002558224 Chr.3: 172750914-172750914 on Build GRCh38
rs569596523 Chr.3: 172750641-172750641 on Build GRCh38
rs1038571104 Chr.3: 172750808-172750808 on Build GRCh38
rs1011525506 Chr.3: 172750884-172750884 on Build GRCh38
rs974182151 Chr.3: 172750994-172750994 on Build GRCh38
rs544211108 Chr.3: 172750783-172750783 on Build GRCh38
rs776388828 Chr.3: 172750903-172750903 on Build GRCh38
rs897159604 Chr.3: 172750639-172750639 on Build GRCh38

View Full Product Details