Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 182819851-182820348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP11B
Entrez Gene ID
23200
Gene Name
ATPase phospholipid transporting 11B (putative)
Gene Aliases
ATPIF, ATPIR
UniGene
Hs.478429
Gene Chromosome Location
Chr.3: 182793503-182923010 on Build GRCh38
Transcripts
NM_014616.2 XM_005247241.2 XM_011512593.2 XM_011512594.2 XM_017006009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5589757 c.46C>T p.Q16* Chr.3: 182820278-182820278 on Build GRCh38
6607113 c.100T>C p.Y34H Chr.3: 182820332-182820332 on Build GRCh38
3206411 c.96C>T p.G32G Chr.3: 182820328-182820328 on Build GRCh38
7447445 c.40C>G p.P14A Chr.3: 182820272-182820272 on Build GRCh38
1421160 c.114A>C p.K38N Chr.3: 182820346-182820346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959180783 Chr.3: 182820288-182820288 on Build GRCh38
rs1008857188 Chr.3: 182820320-182820320 on Build GRCh38
rs948901510 Chr.3: 182820253-182820253 on Build GRCh38
rs563139955 Chr.3: 182820060-182820060 on Build GRCh38
rs772419308 Chr.3: 182820255-182820255 on Build GRCh38
rs1057292542 Chr.3: 182820155-182820155 on Build GRCh38
rs963872533 Chr.3: 182820002-182820002 on Build GRCh38
rs894189454 Chr.3: 182820222-182820222 on Build GRCh38
rs752437985 Chr.3: 182820302-182820302 on Build GRCh38
rs762903585 Chr.3: 182819897-182819897 on Build GRCh38
rs759464404 Chr.3: 182820289-182820289 on Build GRCh38
rs763031361 Chr.3: 182819889-182819889 on Build GRCh38
rs201120571 Chr.3: 182820201-182820201 on Build GRCh38
rs746234826 Chr.3: 182820243-182820243 on Build GRCh38
rs917565544 Chr.3: 182819906-182819906 on Build GRCh38
rs766540640 Chr.3: 182820141-182820141 on Build GRCh38
rs1016489016 Chr.3: 182819905-182819905 on Build GRCh38
rs375065862 Chr.3: 182820274-182820274 on Build GRCh38
rs1039633243 Chr.3: 182820081-182820081 on Build GRCh38
rs141226430 Chr.3: 182820277-182820277 on Build GRCh38
rs951180814 Chr.3: 182820233-182820233 on Build GRCh38
rs763753686 Chr.3: 182820305-182820305 on Build GRCh38
rs79910998 Chr.3: 182820041-182820041 on Build GRCh38
rs993696814 Chr.3: 182819934-182819934 on Build GRCh38
rs975223222 Chr.3: 182820005-182820006 on Build GRCh38
rs530658729 Chr.3: 182820125-182820125 on Build GRCh38
rs774188470 Chr.3: 182819972-182819972 on Build GRCh38
rs765991771 Chr.3: 182820230-182820233 on Build GRCh38
rs189462496 Chr.3: 182819943-182819943 on Build GRCh38
rs753301030 Chr.3: 182820308-182820308 on Build GRCh38
rs150296662 Chr.3: 182820301-182820301 on Build GRCh38
rs1041763821 Chr.3: 182820048-182820048 on Build GRCh38
rs761899019 Chr.3: 182820273-182820273 on Build GRCh38
rs372793473 Chr.3: 182820123-182820123 on Build GRCh38
rs754311233 Chr.3: 182820161-182820161 on Build GRCh38
rs1041618503 Chr.3: 182820035-182820036 on Build GRCh38
rs899720155 Chr.3: 182820135-182820135 on Build GRCh38
rs1027205154 Chr.3: 182820171-182820171 on Build GRCh38
rs150709291 Chr.3: 182820256-182820256 on Build GRCh38
rs137914384 Chr.3: 182820303-182820303 on Build GRCh38
rs185261902 Chr.3: 182820000-182820000 on Build GRCh38
rs914672816 Chr.3: 182820014-182820014 on Build GRCh38
rs776599975 Chr.3: 182820272-182820272 on Build GRCh38
rs779333950 Chr.3: 182820230-182820230 on Build GRCh38
rs142985880 Chr.3: 182820140-182820140 on Build GRCh38
rs191150085 Chr.3: 182820189-182820189 on Build GRCh38
rs768818214 Chr.3: 182820269-182820269 on Build GRCh38
rs757726180 Chr.3: 182820224-182820224 on Build GRCh38
rs924312950 Chr.3: 182820036-182820036 on Build GRCh38
rs868139726 Chr.3: 182820031-182820031 on Build GRCh38
rs1054994631 Chr.3: 182820010-182820010 on Build GRCh38
rs759079076 Chr.3: 182820052-182820052 on Build GRCh38
rs947317508 Chr.3: 182820019-182820019 on Build GRCh38
rs766663172 Chr.3: 182820074-182820074 on Build GRCh38
rs774430043 Chr.3: 182820284-182820284 on Build GRCh38
rs927524100 Chr.3: 182819956-182819956 on Build GRCh38
rs756717697 Chr.3: 182820317-182820317 on Build GRCh38
rs938354046 Chr.3: 182820091-182820092 on Build GRCh38
rs747173362 Chr.3: 182820260-182820260 on Build GRCh38
rs558173533 Chr.3: 182819892-182819892 on Build GRCh38
rs562180031 Chr.3: 182819950-182819950 on Build GRCh38
rs914746851 Chr.3: 182820314-182820314 on Build GRCh38
rs766108022 Chr.3: 182819908-182819908 on Build GRCh38
rs1008173976 Chr.3: 182820077-182820077 on Build GRCh38
rs761105008 Chr.3: 182819984-182819984 on Build GRCh38
rs182061200 Chr.3: 182819944-182819944 on Build GRCh38
rs987992401 Chr.3: 182819964-182819964 on Build GRCh38
rs946097477 Chr.3: 182820045-182820045 on Build GRCh38

View Full Product Details