Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 185082611-185083117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf70
Entrez Gene ID
285382
Gene Name
chromosome 3 open reading frame 70
Gene Aliases
-
UniGene
Hs.380048
Gene Chromosome Location
Chr.3: 185078050-185153014 on Build GRCh38
Transcripts
NM_001025266.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667745 c.681G>A p.P227P Chr.3: 185083079-185083079 on Build GRCh38
3331708 c.735G>A p.T245T Chr.3: 185083025-185083025 on Build GRCh38
4632335 c.708G>A p.S236S Chr.3: 185083052-185083052 on Build GRCh38
5001153 c.715G>T p.D239Y Chr.3: 185083045-185083045 on Build GRCh38
5864065 c.701C>T p.S234F Chr.3: 185083059-185083059 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778071741 Chr.3: 185083061-185083061 on Build GRCh38
rs766838018 Chr.3: 185082751-185082751 on Build GRCh38
rs192437094 Chr.3: 185082975-185082975 on Build GRCh38
rs997965444 Chr.3: 185082659-185082659 on Build GRCh38
rs771483286 Chr.3: 185083069-185083069 on Build GRCh38
rs551593933 Chr.3: 185082856-185082856 on Build GRCh38
rs989092978 Chr.3: 185083071-185083071 on Build GRCh38
rs961185956 Chr.3: 185082707-185082707 on Build GRCh38
rs531623074 Chr.3: 185082675-185082675 on Build GRCh38
rs972506246 Chr.3: 185082709-185082709 on Build GRCh38
rs773729356 Chr.3: 185082996-185082996 on Build GRCh38
rs1031912810 Chr.3: 185082640-185082640 on Build GRCh38
rs1016888870 Chr.3: 185082695-185082695 on Build GRCh38
rs796585535 Chr.3: 185082931-185082931 on Build GRCh38
rs960810375 Chr.3: 185082782-185082782 on Build GRCh38
rs201641850 Chr.3: 185083048-185083048 on Build GRCh38
rs889446152 Chr.3: 185082946-185082946 on Build GRCh38
rs752121599 Chr.3: 185083010-185083010 on Build GRCh38
rs534156954 Chr.3: 185082884-185082884 on Build GRCh38
rs1028379808 Chr.3: 185082674-185082674 on Build GRCh38
rs908404535 Chr.3: 185082813-185082814 on Build GRCh38
rs752352893 Chr.3: 185083092-185083093 on Build GRCh38
rs933723883 Chr.3: 185082899-185082899 on Build GRCh38
rs775400044 Chr.3: 185083026-185083026 on Build GRCh38
rs146317967 Chr.3: 185083078-185083078 on Build GRCh38
rs770082686 Chr.3: 185083059-185083059 on Build GRCh38
rs778783502 Chr.3: 185082970-185082970 on Build GRCh38
rs533311417 Chr.3: 185082828-185082828 on Build GRCh38
rs760133055 Chr.3: 185083075-185083075 on Build GRCh38
rs900886965 Chr.3: 185082649-185082649 on Build GRCh38
rs970124016 Chr.3: 185082839-185082839 on Build GRCh38
rs891237945 Chr.3: 185082898-185082898 on Build GRCh38
rs1053290803 Chr.3: 185082648-185082648 on Build GRCh38
rs939914419 Chr.3: 185082847-185082847 on Build GRCh38
rs187840065 Chr.3: 185082869-185082869 on Build GRCh38
rs979417016 Chr.3: 185082867-185082867 on Build GRCh38
rs891982325 Chr.3: 185082943-185082943 on Build GRCh38
rs763742130 Chr.3: 185083092-185083092 on Build GRCh38
rs766994274 Chr.3: 185083001-185083001 on Build GRCh38
rs780699508 Chr.3: 185083015-185083023 on Build GRCh38
rs769884881 Chr.3: 185082980-185082980 on Build GRCh38
rs142152683 Chr.3: 185083056-185083056 on Build GRCh38
rs756981203 Chr.3: 185083043-185083043 on Build GRCh38
rs777563746 Chr.3: 185082900-185082900 on Build GRCh38
rs1005861955 Chr.3: 185082688-185082688 on Build GRCh38
rs543744858 Chr.3: 185083080-185083080 on Build GRCh38
rs201783027 Chr.3: 185083053-185083053 on Build GRCh38
rs538616874 Chr.3: 185082981-185082981 on Build GRCh38
rs967746143 Chr.3: 185082672-185082672 on Build GRCh38
rs767125323 Chr.3: 185083007-185083007 on Build GRCh38
rs199721046 Chr.3: 185082972-185082972 on Build GRCh38
rs934904934 Chr.3: 185082879-185082879 on Build GRCh38
rs1005250675 Chr.3: 185082966-185082966 on Build GRCh38
rs549148730 Chr.3: 185082893-185082893 on Build GRCh38
rs544162149 Chr.3: 185082754-185082754 on Build GRCh38
rs774855344 Chr.3: 185083073-185083073 on Build GRCh38
rs368971235 Chr.3: 185082969-185082969 on Build GRCh38
rs542880085 Chr.3: 185083016-185083016 on Build GRCh38
rs200241839 Chr.3: 185082999-185082999 on Build GRCh38
rs1021939591 Chr.3: 185082660-185082660 on Build GRCh38
rs781381496 Chr.3: 185083055-185083055 on Build GRCh38
rs375797500 Chr.3: 185082973-185082973 on Build GRCh38
rs753665859 Chr.3: 185083025-185083025 on Build GRCh38
rs113106000 Chr.3: 185083005-185083005 on Build GRCh38
rs756606694 Chr.3: 185082710-185082710 on Build GRCh38
rs184467596 Chr.3: 185083017-185083017 on Build GRCh38
rs572830042 Chr.3: 185082785-185082785 on Build GRCh38
rs139649690 Chr.3: 185083079-185083079 on Build GRCh38
rs774939051 Chr.3: 185083004-185083004 on Build GRCh38
rs550067264 Chr.3: 185082723-185082723 on Build GRCh38
rs756872479 Chr.3: 185083052-185083052 on Build GRCh38
rs567485772 Chr.3: 185082945-185082945 on Build GRCh38

View Full Product Details