Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 47936470-47936965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGA1
Entrez Gene ID
1259
Gene Name
cyclic nucleotide gated channel alpha 1
Gene Aliases
CNCG, CNCG1, CNG-1, CNG1, RCNC1, RCNCa, RCNCalpha, RP49
UniGene
Hs.1323
Gene Chromosome Location
Chr.4: 47935015-48016718 on Build GRCh38
Transcripts
NM_000087.3 NM_001142564.1 XM_005248049.4 XM_011513623.2 XM_017007712.1 NR_125879.1

Gene Symbol
LOC101927157
Entrez Gene ID
101927157
Gene Name
uncharacterized LOC101927157
Gene Aliases
-
UniGene
Hs.134190
Gene Chromosome Location
Chr.4: 47914227-47990737 on Build GRCh38
Transcripts
NM_000087.3 NM_001142564.1 XM_005248049.4 XM_011513623.2 XM_017007712.1 NR_125879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1429894 c.1541G>A p.R514Q Chr.4: 47936953-47936953 on Build GRCh38
5709260 c.2014T>G p.L672V Chr.4: 47936687-47936687 on Build GRCh38
4531963 c.1633G>A p.G545S Chr.4: 47936861-47936861 on Build GRCh38
7205491 c.2053G>T p.A685S Chr.4: 47936648-47936648 on Build GRCh38
387427 c.1973G>T p.R658I Chr.4: 47936521-47936521 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17573673 Chr.4: 47936862-47936862 on Build GRCh38
rs371440311 Chr.4: 47936552-47936552 on Build GRCh38
rs768363051 Chr.4: 47936795-47936795 on Build GRCh38
rs942857951 Chr.4: 47936677-47936677 on Build GRCh38
rs945651304 Chr.4: 47936663-47936663 on Build GRCh38
rs549705696 Chr.4: 47936543-47936543 on Build GRCh38
rs768694789 Chr.4: 47936736-47936739 on Build GRCh38
rs1042384 Chr.4: 47936709-47936709 on Build GRCh38
rs778589744 Chr.4: 47936881-47936881 on Build GRCh38
rs759904304 Chr.4: 47936839-47936839 on Build GRCh38
rs764456378 Chr.4: 47936934-47936934 on Build GRCh38
rs754004038 Chr.4: 47936590-47936590 on Build GRCh38
rs529542511 Chr.4: 47936508-47936508 on Build GRCh38
rs750370709 Chr.4: 47936749-47936749 on Build GRCh38
rs894837753 Chr.4: 47936536-47936536 on Build GRCh38
rs189580090 Chr.4: 47936570-47936570 on Build GRCh38
rs750411022 Chr.4: 47936778-47936778 on Build GRCh38
rs376959147 Chr.4: 47936609-47936609 on Build GRCh38
rs773644867 Chr.4: 47936701-47936701 on Build GRCh38
rs761341105 Chr.4: 47936695-47936695 on Build GRCh38
rs199992510 Chr.4: 47936593-47936593 on Build GRCh38
rs747824246 Chr.4: 47936882-47936882 on Build GRCh38
rs762706687 Chr.4: 47936550-47936550 on Build GRCh38
rs867972995 Chr.4: 47936815-47936815 on Build GRCh38
rs745803581 Chr.4: 47936629-47936629 on Build GRCh38
rs527236057 Chr.4: 47936861-47936861 on Build GRCh38
rs777233831 Chr.4: 47936889-47936889 on Build GRCh38
rs781234380 Chr.4: 47936618-47936618 on Build GRCh38
rs768123452 Chr.4: 47936566-47936566 on Build GRCh38
rs765450800 Chr.4: 47936715-47936715 on Build GRCh38
rs780203611 Chr.4: 47936500-47936500 on Build GRCh38
rs182234888 Chr.4: 47936853-47936853 on Build GRCh38
rs763233407 Chr.4: 47936924-47936924 on Build GRCh38
rs766717829 Chr.4: 47936713-47936713 on Build GRCh38
rs370636592 Chr.4: 47936636-47936636 on Build GRCh38
rs756433866 Chr.4: 47936584-47936584 on Build GRCh38
rs552871598 Chr.4: 47936885-47936885 on Build GRCh38
rs753754324 Chr.4: 47936880-47936880 on Build GRCh38
rs1018800266 Chr.4: 47936900-47936900 on Build GRCh38
rs375049806 Chr.4: 47936714-47936714 on Build GRCh38
rs771604546 Chr.4: 47936700-47936700 on Build GRCh38
rs748884455 Chr.4: 47936664-47936664 on Build GRCh38
rs752892318 Chr.4: 47936719-47936719 on Build GRCh38
rs758429898 Chr.4: 47936615-47936615 on Build GRCh38
rs771469417 Chr.4: 47936830-47936830 on Build GRCh38
rs537196863 Chr.4: 47936681-47936681 on Build GRCh38
rs548116727 Chr.4: 47936509-47936509 on Build GRCh38
rs770457952 Chr.4: 47936899-47936899 on Build GRCh38
rs761166989 Chr.4: 47936818-47936818 on Build GRCh38
rs577544200 Chr.4: 47936918-47936918 on Build GRCh38
rs551835238 Chr.4: 47936547-47936547 on Build GRCh38
rs775781523 Chr.4: 47936922-47936922 on Build GRCh38
rs756184834 Chr.4: 47936751-47936751 on Build GRCh38
rs373782782 Chr.4: 47936608-47936608 on Build GRCh38
rs776305454 Chr.4: 47936780-47936793 on Build GRCh38
rs201633176 Chr.4: 47936784-47936784 on Build GRCh38
rs746572064 Chr.4: 47936892-47936892 on Build GRCh38
rs540881872 Chr.4: 47936513-47936513 on Build GRCh38
rs758436235 Chr.4: 47936728-47936728 on Build GRCh38
rs762191770 Chr.4: 47936561-47936561 on Build GRCh38
rs151223668 Chr.4: 47936809-47936809 on Build GRCh38
rs567961453 Chr.4: 47936816-47936816 on Build GRCh38
rs761185333 Chr.4: 47936712-47936712 on Build GRCh38
rs769653731 Chr.4: 47936640-47936640 on Build GRCh38
rs1018767693 Chr.4: 47936576-47936576 on Build GRCh38
rs774732831 Chr.4: 47936938-47936938 on Build GRCh38
rs770967964 Chr.4: 47936525-47936525 on Build GRCh38
rs750954043 Chr.4: 47936567-47936567 on Build GRCh38
rs776837749 Chr.4: 47936831-47936831 on Build GRCh38
rs77505978 Chr.4: 47936601-47936601 on Build GRCh38
rs778010980 Chr.4: 47936729-47936729 on Build GRCh38
rs768391117 Chr.4: 47936679-47936679 on Build GRCh38
rs765523630 Chr.4: 47936840-47936840 on Build GRCh38
rs770180675 Chr.4: 47936921-47936921 on Build GRCh38
rs766736379 Chr.4: 47936588-47936588 on Build GRCh38
rs752905054 Chr.4: 47936849-47936849 on Build GRCh38
rs370912894 Chr.4: 47936876-47936876 on Build GRCh38
rs373120472 Chr.4: 47936860-47936860 on Build GRCh38
rs747005447 Chr.4: 47936562-47936563 on Build GRCh38
rs886059422 Chr.4: 47936770-47936770 on Build GRCh38
rs749152335 Chr.4: 47936792-47936792 on Build GRCh38
rs566054486 Chr.4: 47936515-47936515 on Build GRCh38
rs867327430 Chr.4: 47936545-47936545 on Build GRCh38
rs778592209 Chr.4: 47936800-47936800 on Build GRCh38
rs373946784 Chr.4: 47936912-47936912 on Build GRCh38
rs776545639 Chr.4: 47936542-47936542 on Build GRCh38
rs746908022 Chr.4: 47936521-47936521 on Build GRCh38
rs779609032 Chr.4: 47936877-47936877 on Build GRCh38
rs756311008 Chr.4: 47936497-47936497 on Build GRCh38
rs376378429 Chr.4: 47936548-47936548 on Build GRCh38

View Full Product Details