Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 105630814-105631330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF38
Entrez Gene ID
54848
Gene Name
Rho guanine nucleotide exchange factor 38
Gene Aliases
-
UniGene
Hs.585224
Gene Chromosome Location
Chr.4: 105552603-105682539 on Build GRCh38
Transcripts
NM_001242729.1 NM_017700.1 XM_006714244.3 XM_011532050.2 XM_011532051.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6604314 c.572A>G p.H191R Chr.4: 105630961-105630961 on Build GRCh38
6241312 c.642T>A p.C214* Chr.4: 105631031-105631031 on Build GRCh38
6604313 c.572A>G p.H191R Chr.4: 105630961-105630961 on Build GRCh38
1485577 c.510_512delAGA p.E171delE Chr.4: 105630899-105630901 on Build GRCh38
7194552 c.656+6G>T p.? Chr.4: 105631051-105631051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573839061 Chr.4: 105631157-105631157 on Build GRCh38
rs764760665 Chr.4: 105630974-105630974 on Build GRCh38
rs778790444 Chr.4: 105631276-105631276 on Build GRCh38
rs545198673 Chr.4: 105631159-105631159 on Build GRCh38
rs565361044 Chr.4: 105631171-105631171 on Build GRCh38
rs371602710 Chr.4: 105630913-105630913 on Build GRCh38
rs752413604 Chr.4: 105631094-105631094 on Build GRCh38
rs751148437 Chr.4: 105631087-105631087 on Build GRCh38
rs533175469 Chr.4: 105631207-105631207 on Build GRCh38
rs994849620 Chr.4: 105631111-105631111 on Build GRCh38
rs772495615 Chr.4: 105631032-105631032 on Build GRCh38
rs200163297 Chr.4: 105631011-105631011 on Build GRCh38
rs754803940 Chr.4: 105630890-105630890 on Build GRCh38
rs911777052 Chr.4: 105630880-105630880 on Build GRCh38
rs761088099 Chr.4: 105630973-105630973 on Build GRCh38
rs769017240 Chr.4: 105631057-105631057 on Build GRCh38
rs928513860 Chr.4: 105631223-105631223 on Build GRCh38
rs2044536 Chr.4: 105631257-105631257 on Build GRCh38
rs374218147 Chr.4: 105631026-105631026 on Build GRCh38
rs1055350198 Chr.4: 105631231-105631231 on Build GRCh38
rs183665500 Chr.4: 105631219-105631219 on Build GRCh38
rs376484696 Chr.4: 105631224-105631224 on Build GRCh38
rs985735786 Chr.4: 105630846-105630846 on Build GRCh38
rs765715238 Chr.4: 105631072-105631073 on Build GRCh38
rs975793457 Chr.4: 105630936-105630936 on Build GRCh38
rs150102971 Chr.4: 105630912-105630912 on Build GRCh38
rs767549444 Chr.4: 105630933-105630933 on Build GRCh38
rs747862229 Chr.4: 105630889-105630889 on Build GRCh38
rs965498760 Chr.4: 105631141-105631141 on Build GRCh38
rs776005575 Chr.4: 105630962-105630962 on Build GRCh38
rs201736725 Chr.4: 105630955-105630955 on Build GRCh38
rs762444769 Chr.4: 105631066-105631066 on Build GRCh38
rs529138678 Chr.4: 105631245-105631245 on Build GRCh38
rs1036832609 Chr.4: 105631233-105631233 on Build GRCh38
rs191755011 Chr.4: 105631214-105631214 on Build GRCh38
rs543016276 Chr.4: 105631020-105631020 on Build GRCh38
rs756028271 Chr.4: 105631008-105631008 on Build GRCh38
rs761191711 Chr.4: 105631052-105631052 on Build GRCh38
rs369856984 Chr.4: 105631017-105631017 on Build GRCh38
rs775744775 Chr.4: 105631043-105631043 on Build GRCh38
rs769261226 Chr.4: 105630889-105630889 on Build GRCh38
rs753665160 Chr.4: 105631198-105631198 on Build GRCh38
rs780855992 Chr.4: 105631001-105631001 on Build GRCh38
rs570195843 Chr.4: 105631120-105631120 on Build GRCh38
rs752819357 Chr.4: 105631086-105631086 on Build GRCh38
rs565587749 Chr.4: 105631268-105631268 on Build GRCh38
rs751277609 Chr.4: 105630998-105630998 on Build GRCh38
rs762227222 Chr.4: 105630963-105630964 on Build GRCh38
rs55893297 Chr.4: 105631219-105631219 on Build GRCh38
rs886511531 Chr.4: 105631298-105631298 on Build GRCh38
rs112405522 Chr.4: 105630942-105630942 on Build GRCh38
rs906758419 Chr.4: 105631052-105631052 on Build GRCh38
rs889773361 Chr.4: 105631274-105631274 on Build GRCh38
rs61757395 Chr.4: 105631010-105631010 on Build GRCh38
rs187226727 Chr.4: 105631064-105631064 on Build GRCh38
rs757952103 Chr.4: 105630988-105630988 on Build GRCh38
rs771256401 Chr.4: 105630946-105630946 on Build GRCh38
rs938258547 Chr.4: 105631228-105631228 on Build GRCh38
rs964978380 Chr.4: 105631166-105631166 on Build GRCh38
rs200568523 Chr.4: 105631006-105631006 on Build GRCh38
rs767295133 Chr.4: 105631092-105631092 on Build GRCh38
rs779775423 Chr.4: 105630866-105630866 on Build GRCh38
rs974726952 Chr.4: 105631194-105631194 on Build GRCh38
rs189615529 Chr.4: 105630865-105630865 on Build GRCh38
rs1036885541 Chr.4: 105631031-105631031 on Build GRCh38
rs924385630 Chr.4: 105630941-105630941 on Build GRCh38
rs34152356 Chr.4: 105630961-105630961 on Build GRCh38
rs142871068 Chr.4: 105630935-105630935 on Build GRCh38
rs777028244 Chr.4: 105631225-105631225 on Build GRCh38
rs989917644 Chr.4: 105631178-105631178 on Build GRCh38
rs571536985 Chr.4: 105630991-105630991 on Build GRCh38
rs534181396 Chr.4: 105631270-105631274 on Build GRCh38
rs749920485 Chr.4: 105630986-105630986 on Build GRCh38
rs759218987 Chr.4: 105631089-105631089 on Build GRCh38
rs748050616 Chr.4: 105630924-105630924 on Build GRCh38
rs764358182 Chr.4: 105631185-105631185 on Build GRCh38
rs746780541 Chr.4: 105630867-105630867 on Build GRCh38
rs1048508551 Chr.4: 105631270-105631270 on Build GRCh38
rs774704317 Chr.4: 105631023-105631023 on Build GRCh38
rs749851425 Chr.4: 105631143-105631144 on Build GRCh38
rs537232897 Chr.4: 105631000-105631000 on Build GRCh38
rs368669982 Chr.4: 105630949-105630949 on Build GRCh38
rs978206091 Chr.4: 105631147-105631147 on Build GRCh38
rs1026374887 Chr.4: 105631139-105631139 on Build GRCh38
rs745751515 Chr.4: 105631279-105631279 on Build GRCh38
rs1019854611 Chr.4: 105631121-105631121 on Build GRCh38
rs573186872 Chr.4: 105631077-105631077 on Build GRCh38
rs147678698 Chr.4: 105630960-105630960 on Build GRCh38
rs1009661747 Chr.4: 105631027-105631027 on Build GRCh38
rs777318246 Chr.4: 105630939-105630945 on Build GRCh38

View Full Product Details