Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 105682349-105682846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS12
Entrez Gene ID
57117
Gene Name
integrator complex subunit 12
Gene Aliases
INT12, PHF22, SBBI22
UniGene
Hs.480454
Gene Chromosome Location
Chr.4: 105682627-105708725 on Build GRCh38
Transcripts
NM_001142471.1 NM_020395.3 XM_005263148.4 XM_011532143.2 XM_011532145.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
337121 c.1282C>T p.P428S Chr.4: 105682840-105682840 on Build GRCh38
1618228 c.1360A>G p.K454E Chr.4: 105682762-105682762 on Build GRCh38
1309599 c.1347G>A p.Q449Q Chr.4: 105682775-105682775 on Build GRCh38
3945854 c.1336A>T p.K446* Chr.4: 105682786-105682786 on Build GRCh38
4598772 c.1366G>A p.A456T Chr.4: 105682756-105682756 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944197411 Chr.4: 105682660-105682660 on Build GRCh38
rs576536666 Chr.4: 105682630-105682630 on Build GRCh38
rs974974537 Chr.4: 105682609-105682609 on Build GRCh38
rs778085922 Chr.4: 105682816-105682816 on Build GRCh38
rs1009824577 Chr.4: 105682376-105682376 on Build GRCh38
rs372755279 Chr.4: 105682695-105682695 on Build GRCh38
rs1033614533 Chr.4: 105682820-105682820 on Build GRCh38
rs763615024 Chr.4: 105682730-105682730 on Build GRCh38
rs777729920 Chr.4: 105682551-105682551 on Build GRCh38
rs768714170 Chr.4: 105682682-105682682 on Build GRCh38
rs769883106 Chr.4: 105682698-105682698 on Build GRCh38
rs1039945603 Chr.4: 105682667-105682667 on Build GRCh38
rs753268208 Chr.4: 105682742-105682742 on Build GRCh38
rs147779765 Chr.4: 105682382-105682382 on Build GRCh38
rs912754464 Chr.4: 105682623-105682623 on Build GRCh38
rs755843749 Chr.4: 105682373-105682373 on Build GRCh38
rs756834645 Chr.4: 105682749-105682749 on Build GRCh38
rs766863487 Chr.4: 105682712-105682712 on Build GRCh38
rs777943422 Chr.4: 105682692-105682692 on Build GRCh38
rs76612485 Chr.4: 105682596-105682596 on Build GRCh38
rs539096809 Chr.4: 105682639-105682639 on Build GRCh38
rs913782598 Chr.4: 105682444-105682444 on Build GRCh38
rs936672634 Chr.4: 105682676-105682676 on Build GRCh38
rs748144462 Chr.4: 105682814-105682814 on Build GRCh38
rs778333714 Chr.4: 105682708-105682708 on Build GRCh38
rs779826195 Chr.4: 105682787-105682787 on Build GRCh38
rs535829080 Chr.4: 105682675-105682675 on Build GRCh38
rs1055248349 Chr.4: 105682549-105682549 on Build GRCh38
rs759996806 Chr.4: 105682728-105682728 on Build GRCh38
rs533829755 Chr.4: 105682548-105682548 on Build GRCh38
rs966663265 Chr.4: 105682464-105682464 on Build GRCh38
rs979244688 Chr.4: 105682487-105682487 on Build GRCh38
rs568242491 Chr.4: 105682431-105682431 on Build GRCh38
rs751457421 Chr.4: 105682789-105682789 on Build GRCh38
rs1008278286 Chr.4: 105682794-105682794 on Build GRCh38
rs34234581 Chr.4: 105682778-105682778 on Build GRCh38
rs750131305 Chr.4: 105682782-105682782 on Build GRCh38
rs370808431 Chr.4: 105682694-105682694 on Build GRCh38
rs1027938964 Chr.4: 105682430-105682430 on Build GRCh38
rs955183428 Chr.4: 105682433-105682433 on Build GRCh38
rs79866548 Chr.4: 105682704-105682704 on Build GRCh38
rs986487783 Chr.4: 105682442-105682442 on Build GRCh38
rs774933016 Chr.4: 105682719-105682719 on Build GRCh38
rs973068486 Chr.4: 105682556-105682556 on Build GRCh38
rs886677354 Chr.4: 105682621-105682621 on Build GRCh38
rs754894181 Chr.4: 105682796-105682796 on Build GRCh38
rs1047736253 Chr.4: 105682598-105682598 on Build GRCh38
rs376992002 Chr.4: 105682785-105682785 on Build GRCh38
rs987626081 Chr.4: 105682618-105682618 on Build GRCh38
rs925129464 Chr.4: 105682504-105682504 on Build GRCh38
rs370294113 Chr.4: 105682798-105682798 on Build GRCh38
rs919003788 Chr.4: 105682568-105682568 on Build GRCh38
rs62320293 Chr.4: 105682729-105682729 on Build GRCh38

View Full Product Details