Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 107895169-107895690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101929595
Entrez Gene ID
101929595
Gene Name
uncharacterized LOC101929595
Gene Aliases
-
UniGene
Hs.678014
Gene Chromosome Location
Chr.4: 107863479-107932119 on Build GRCh38
Transcripts
NR_125929.1 NM_001136257.1 NM_001136258.1 NM_152621.5 XM_011531698.2 XM_011531699.2 XM_011531700.2 XM_011531701.2 XM_011531702.2 XM_017007839.1 XM_017007840.1 XM_017007841.1

Gene Symbol
SGMS2
Entrez Gene ID
166929
Gene Name
sphingomyelin synthase 2
Gene Aliases
SMS2
UniGene
Hs.595423
Gene Chromosome Location
Chr.4: 107823645-107915047 on Build GRCh38
Transcripts
NR_125929.1 NM_001136257.1 NM_001136258.1 NM_152621.5 XM_011531698.2 XM_011531699.2 XM_011531700.2 XM_011531701.2 XM_011531702.2 XM_017007839.1 XM_017007840.1 XM_017007841.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5525562 c.126C>T p.P42P Chr.4: 107895679-107895679 on Build GRCh38
1426274 c.81C>T p.P27P Chr.4: 107895634-107895634 on Build GRCh38
5717206 c.6T>G p.D2E Chr.4: 107895559-107895559 on Build GRCh38
4121209 c.68C>G p.T23S Chr.4: 107895621-107895621 on Build GRCh38
7180959 c.100G>C p.E34Q Chr.4: 107895653-107895653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043992508 Chr.4: 107895310-107895310 on Build GRCh38
rs763589073 Chr.4: 107895496-107895496 on Build GRCh38
rs553158004 Chr.4: 107895539-107895539 on Build GRCh38
rs565028161 Chr.4: 107895616-107895616 on Build GRCh38
rs1020264186 Chr.4: 107895208-107895208 on Build GRCh38
rs1041013030 Chr.4: 107895428-107895428 on Build GRCh38
rs1013707876 Chr.4: 107895456-107895456 on Build GRCh38
rs951764006 Chr.4: 107895360-107895360 on Build GRCh38
rs574040888 Chr.4: 107895461-107895461 on Build GRCh38
rs960810406 Chr.4: 107895282-107895282 on Build GRCh38
rs572555808 Chr.4: 107895552-107895552 on Build GRCh38
rs558104539 Chr.4: 107895267-107895267 on Build GRCh38
rs17038199 Chr.4: 107895256-107895256 on Build GRCh38
rs754215424 Chr.4: 107895438-107895439 on Build GRCh38
rs746612884 Chr.4: 107895562-107895562 on Build GRCh38
rs778400449 Chr.4: 107895548-107895548 on Build GRCh38
rs762688604 Chr.4: 107895524-107895525 on Build GRCh38
rs1041759879 Chr.4: 107895512-107895512 on Build GRCh38
rs762913325 Chr.4: 107895606-107895606 on Build GRCh38
rs988552262 Chr.4: 107895643-107895643 on Build GRCh38
rs1040260449 Chr.4: 107895385-107895385 on Build GRCh38
rs756574341 Chr.4: 107895546-107895546 on Build GRCh38
rs893235626 Chr.4: 107895483-107895483 on Build GRCh38
rs773253543 Chr.4: 107895590-107895590 on Build GRCh38
rs751873442 Chr.4: 107895516-107895516 on Build GRCh38
rs933060192 Chr.4: 107895272-107895272 on Build GRCh38
rs557209731 Chr.4: 107895404-107895404 on Build GRCh38
rs916334503 Chr.4: 107895348-107895348 on Build GRCh38
rs17038204 Chr.4: 107895615-107895615 on Build GRCh38
rs542802930 Chr.4: 107895474-107895474 on Build GRCh38
rs1006021272 Chr.4: 107895473-107895473 on Build GRCh38
rs762047509 Chr.4: 107895632-107895632 on Build GRCh38
rs17038201 Chr.4: 107895279-107895279 on Build GRCh38
rs775062299 Chr.4: 107895361-107895361 on Build GRCh38
rs1016187101 Chr.4: 107895196-107895196 on Build GRCh38
rs781649926 Chr.4: 107895535-107895535 on Build GRCh38
rs990619281 Chr.4: 107895342-107895342 on Build GRCh38
rs754329411 Chr.4: 107895656-107895656 on Build GRCh38
rs768306630 Chr.4: 107895563-107895563 on Build GRCh38
rs886312194 Chr.4: 107895466-107895466 on Build GRCh38
rs761667531 Chr.4: 107895571-107895571 on Build GRCh38
rs779568386 Chr.4: 107895561-107895561 on Build GRCh38
rs530407088 Chr.4: 107895622-107895622 on Build GRCh38
rs984486214 Chr.4: 107895650-107895650 on Build GRCh38
rs759776066 Chr.4: 107895621-107895621 on Build GRCh38
rs764476029 Chr.4: 107895645-107895645 on Build GRCh38
rs999826812 Chr.4: 107895277-107895277 on Build GRCh38
rs143392363 Chr.4: 107895383-107895383 on Build GRCh38
rs1011640476 Chr.4: 107895492-107895492 on Build GRCh38
rs923025857 Chr.4: 107895269-107895269 on Build GRCh38
rs1055111946 Chr.4: 107895386-107895386 on Build GRCh38
rs914099641 Chr.4: 107895306-107895306 on Build GRCh38
rs767159178 Chr.4: 107895309-107895309 on Build GRCh38
rs200702483 Chr.4: 107895534-107895534 on Build GRCh38
rs769608601 Chr.4: 107895572-107895572 on Build GRCh38
rs756549531 Chr.4: 107895635-107895635 on Build GRCh38
rs149647259 Chr.4: 107895198-107895198 on Build GRCh38
rs753098107 Chr.4: 107895634-107895634 on Build GRCh38
rs372458980 Chr.4: 107895502-107895502 on Build GRCh38
rs867220165 Chr.4: 107895658-107895658 on Build GRCh38

View Full Product Details