Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 113452863-113453410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK2D
Entrez Gene ID
817
Gene Name
calcium/calmodulin dependent protein kinase II delta
Gene Aliases
CAMKD
UniGene
Hs.144114
Gene Chromosome Location
Chr.4: 113451032-113761927 on Build GRCh38
Transcripts
NM_001221.3 NM_001321566.1 NM_001321567.1 NM_001321568.1 NM_001321569.1 NM_001321570.1 NM_001321571.1 NM_001321572.1 NM_001321573.1 NM_001321574.1 NM_001321575.1 NM_001321576.1 NM_001321577.1 NM_001321578.1 NM_001321579.1 NM_001321580.1 NM_001321581.1 NM_001321582.1 NM_001321583.1 NM_001321584.1 NM_001321585.1 NM_001321586.1 NM_001321587.1 NM_001321588.1 NM_001321589.1 NM_001321590.1 NM_001321591.1 NM_172127.2 NM_172128.2 XM_005263253.3 XM_005263254.3 XM_006714330.2 XM_011532289.1 XM_011532290.1 XM_011532291.1 XM_011532292.2 XM_011532295.1 XM_017008672.1 XM_017008673.1 XM_017008674.1 XM_017008675.1 XM_017008676.1 XM_017008677.1 XM_017008678.1

SNP Detail

SNP ID Chromosome Location
rs776050246 Chr.4: 113453215-113453215 on Build GRCh38
rs903634362 Chr.4: 113452964-113452964 on Build GRCh38
rs559806299 Chr.4: 113452974-113452977 on Build GRCh38
rs894884299 Chr.4: 113453340-113453340 on Build GRCh38
rs144440179 Chr.4: 113453375-113453375 on Build GRCh38
rs946278659 Chr.4: 113453242-113453242 on Build GRCh38
rs1054815815 Chr.4: 113452981-113452981 on Build GRCh38
rs529172085 Chr.4: 113453266-113453266 on Build GRCh38
rs944463884 Chr.4: 113453151-113453151 on Build GRCh38
rs374476598 Chr.4: 113452977-113452980 on Build GRCh38
rs1043229083 Chr.4: 113452933-113452933 on Build GRCh38
rs887959702 Chr.4: 113452903-113452903 on Build GRCh38
rs949104118 Chr.4: 113453360-113453360 on Build GRCh38
rs1000858439 Chr.4: 113453367-113453367 on Build GRCh38
rs533021875 Chr.4: 113453379-113453379 on Build GRCh38
rs1013661487 Chr.4: 113453010-113453010 on Build GRCh38
rs1014088485 Chr.4: 113453324-113453324 on Build GRCh38
rs933573437 Chr.4: 113452893-113452893 on Build GRCh38
rs886368872 Chr.4: 113453376-113453376 on Build GRCh38
rs185318044 Chr.4: 113452894-113452894 on Build GRCh38
rs1024064291 Chr.4: 113453032-113453032 on Build GRCh38
rs578134564 Chr.4: 113453105-113453105 on Build GRCh38
rs954769887 Chr.4: 113453221-113453221 on Build GRCh38
rs894775513 Chr.4: 113452997-113452997 on Build GRCh38
rs969375803 Chr.4: 113453045-113453045 on Build GRCh38
rs896620931 Chr.4: 113453319-113453319 on Build GRCh38
rs770234725 Chr.4: 113453285-113453285 on Build GRCh38
rs960778271 Chr.4: 113452991-113452991 on Build GRCh38
rs181038184 Chr.4: 113453083-113453083 on Build GRCh38
rs974757601 Chr.4: 113453182-113453182 on Build GRCh38
rs554019451 Chr.4: 113453201-113453201 on Build GRCh38
rs554946624 Chr.4: 113452951-113452951 on Build GRCh38
rs556690812 Chr.4: 113453091-113453091 on Build GRCh38
rs1016051936 Chr.4: 113453383-113453383 on Build GRCh38
rs969805920 Chr.4: 113453356-113453356 on Build GRCh38
rs1005041795 Chr.4: 113452907-113452907 on Build GRCh38
rs75070532 Chr.4: 113453381-113453381 on Build GRCh38
rs913019321 Chr.4: 113453108-113453108 on Build GRCh38
rs72903949 Chr.4: 113453238-113453238 on Build GRCh38
rs1005092701 Chr.4: 113453241-113453241 on Build GRCh38
rs540107231 Chr.4: 113453115-113453116 on Build GRCh38
rs963447066 Chr.4: 113453144-113453144 on Build GRCh38
rs188974986 Chr.4: 113453022-113453022 on Build GRCh38
rs140599580 Chr.4: 113453237-113453237 on Build GRCh38
rs746627747 Chr.4: 113453048-113453048 on Build GRCh38
rs775951912 Chr.4: 113453355-113453355 on Build GRCh38
rs572574060 Chr.4: 113453219-113453219 on Build GRCh38
rs745412194 Chr.4: 113453270-113453270 on Build GRCh38
rs544205380 Chr.4: 113453368-113453368 on Build GRCh38
rs577789673 Chr.4: 113452916-113452916 on Build GRCh38
rs151338559 Chr.4: 113453106-113453106 on Build GRCh38
rs1027883669 Chr.4: 113452898-113452898 on Build GRCh38
rs936408955 Chr.4: 113453303-113453303 on Build GRCh38
rs563372975 Chr.4: 113452912-113452921 on Build GRCh38

View Full Product Details