Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 121671413-121671840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA5
Entrez Gene ID
308
Gene Name
annexin A5
Gene Aliases
ANX5, ENX2, HEL-S-7, PP4, RPRGL3
UniGene
Hs.480653
Gene Chromosome Location
Chr.4: 121667997-121696992 on Build GRCh38
Transcripts
NM_001154.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
732801 c.721+1G>T p.? Chr.4: 121671546-121671546 on Build GRCh38
1633435 c.638A>G p.Y213C Chr.4: 121671630-121671630 on Build GRCh38
7448832 c.679C>T p.R227C Chr.4: 121671589-121671589 on Build GRCh38
6446982 c.646A>G p.I216V Chr.4: 121671622-121671622 on Build GRCh38
5526725 c.671C>T p.T224I Chr.4: 121671597-121671597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766305577 Chr.4: 121671502-121671502 on Build GRCh38
rs999333628 Chr.4: 121671800-121671800 on Build GRCh38
rs920048933 Chr.4: 121671607-121671607 on Build GRCh38
rs1046906073 Chr.4: 121671684-121671684 on Build GRCh38
rs1030731504 Chr.4: 121671490-121671490 on Build GRCh38
rs756639275 Chr.4: 121671515-121671515 on Build GRCh38
rs113374737 Chr.4: 121671546-121671546 on Build GRCh38
rs187499448 Chr.4: 121671586-121671586 on Build GRCh38
rs371191338 Chr.4: 121671589-121671589 on Build GRCh38
rs72911843 Chr.4: 121671786-121671786 on Build GRCh38
rs753803969 Chr.4: 121671492-121671492 on Build GRCh38
rs1001021176 Chr.4: 121671809-121671809 on Build GRCh38
rs780760057 Chr.4: 121671675-121671675 on Build GRCh38
rs767698464 Chr.4: 121671618-121671618 on Build GRCh38
rs752790362 Chr.4: 121671622-121671622 on Build GRCh38
rs758672905 Chr.4: 121671644-121671644 on Build GRCh38
rs780380715 Chr.4: 121671658-121671658 on Build GRCh38
rs964993141 Chr.4: 121671541-121671541 on Build GRCh38
rs772855301 Chr.4: 121671689-121671689 on Build GRCh38
rs754631699 Chr.4: 121671670-121671670 on Build GRCh38
rs1013040683 Chr.4: 121671443-121671443 on Build GRCh38
rs779186859 Chr.4: 121671664-121671664 on Build GRCh38
rs767808075 Chr.4: 121671465-121671465 on Build GRCh38
rs774301700 Chr.4: 121671610-121671610 on Build GRCh38
rs776205756 Chr.4: 121671578-121671578 on Build GRCh38
rs769149840 Chr.4: 121671687-121671687 on Build GRCh38
rs1021383832 Chr.4: 121671456-121671456 on Build GRCh38
rs779278469 Chr.4: 121671558-121671558 on Build GRCh38
rs532847147 Chr.4: 121671596-121671596 on Build GRCh38
rs112559998 Chr.4: 121671545-121671545 on Build GRCh38
rs970568643 Chr.4: 121671482-121671482 on Build GRCh38
rs755167298 Chr.4: 121671507-121671507 on Build GRCh38
rs1046326225 Chr.4: 121671797-121671797 on Build GRCh38
rs1057049194 Chr.4: 121671718-121671718 on Build GRCh38
rs757565939 Chr.4: 121671539-121671539 on Build GRCh38
rs987085101 Chr.4: 121671537-121671537 on Build GRCh38
rs2306419 Chr.4: 121671508-121671508 on Build GRCh38
rs1030764304 Chr.4: 121671806-121671806 on Build GRCh38
rs765233478 Chr.4: 121671628-121671628 on Build GRCh38
rs543534792 Chr.4: 121671632-121671632 on Build GRCh38
rs942073009 Chr.4: 121671702-121671702 on Build GRCh38
rs766211395 Chr.4: 121671597-121671597 on Build GRCh38
rs762878936 Chr.4: 121671500-121671500 on Build GRCh38
rs559259816 Chr.4: 121671459-121671459 on Build GRCh38
rs906440270 Chr.4: 121671799-121671799 on Build GRCh38
rs999300790 Chr.4: 121671488-121671488 on Build GRCh38
rs759791586 Chr.4: 121671615-121671615 on Build GRCh38
rs746594543 Chr.4: 121671559-121671559 on Build GRCh38
rs768203545 Chr.4: 121671570-121671570 on Build GRCh38
rs147775952 Chr.4: 121671587-121671587 on Build GRCh38
rs749628486 Chr.4: 121671521-121671521 on Build GRCh38
rs1037708308 Chr.4: 121671709-121671709 on Build GRCh38
rs751780419 Chr.4: 121671504-121671504 on Build GRCh38
rs752777699 Chr.4: 121671512-121671512 on Build GRCh38
rs747861614 Chr.4: 121671679-121671679 on Build GRCh38
rs755371269 Chr.4: 121671770-121671770 on Build GRCh38
rs2306418 Chr.4: 121671667-121671667 on Build GRCh38
rs111877480 Chr.4: 121671617-121671617 on Build GRCh38
rs989176235 Chr.4: 121671499-121671499 on Build GRCh38

View Full Product Details