Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 127982861-127983366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD18
Entrez Gene ID
80167
Gene Name
abhydrolase domain containing 18
Gene Aliases
C4orf29
UniGene
Hs.143995
Gene Chromosome Location
Chr.4: 127965256-128039711 on Build GRCh38
Transcripts
NM_001319305.1 NM_001319306.1 NM_001319307.1 XM_005263238.4 XM_006714318.3 XM_006714320.3 XM_011532272.2 XM_011532273.2 XM_011532274.2 XM_011532276.1 XM_011532278.2 XM_017008631.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1051142 c.31C>T p.R11W Chr.4: 127982986-127982986 on Build GRCh38
4557240 c.71G>A p.G24E Chr.4: 127983026-127983026 on Build GRCh38
1051143 c.31C>T p.R11W Chr.4: 127982986-127982986 on Build GRCh38
4557241 c.71G>A p.G24E Chr.4: 127983026-127983026 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772355594 Chr.4: 127983018-127983019 on Build GRCh38
rs541490315 Chr.4: 127982906-127982906 on Build GRCh38
rs868522262 Chr.4: 127983017-127983017 on Build GRCh38
rs72618809 Chr.4: 127983125-127983125 on Build GRCh38
rs759840611 Chr.4: 127982967-127982967 on Build GRCh38
rs975399286 Chr.4: 127983127-127983127 on Build GRCh38
rs892490789 Chr.4: 127982892-127982892 on Build GRCh38
rs189155907 Chr.4: 127983133-127983133 on Build GRCh38
rs922139287 Chr.4: 127983197-127983197 on Build GRCh38
rs376388796 Chr.4: 127982922-127982922 on Build GRCh38
rs868867503 Chr.4: 127982966-127982966 on Build GRCh38
rs988841407 Chr.4: 127983053-127983053 on Build GRCh38
rs770989199 Chr.4: 127983167-127983167 on Build GRCh38
rs976392158 Chr.4: 127983201-127983201 on Build GRCh38
rs756533181 Chr.4: 127983078-127983078 on Build GRCh38
rs761686345 Chr.4: 127982939-127982939 on Build GRCh38
rs1037399742 Chr.4: 127983300-127983300 on Build GRCh38
rs577614250 Chr.4: 127983214-127983214 on Build GRCh38
rs907646317 Chr.4: 127983265-127983265 on Build GRCh38
rs984465471 Chr.4: 127983116-127983116 on Build GRCh38
rs750671699 Chr.4: 127982986-127982986 on Build GRCh38
rs756533732 Chr.4: 127983221-127983221 on Build GRCh38
rs34611986 Chr.4: 127983173-127983174 on Build GRCh38
rs538493385 Chr.4: 127983055-127983056 on Build GRCh38
rs755502727 Chr.4: 127983023-127983023 on Build GRCh38
rs1024955808 Chr.4: 127982934-127982934 on Build GRCh38
rs758565519 Chr.4: 127982987-127982987 on Build GRCh38
rs527538720 Chr.4: 127982970-127982970 on Build GRCh38
rs780155020 Chr.4: 127982997-127982997 on Build GRCh38
rs148201878 Chr.4: 127983272-127983272 on Build GRCh38
rs373645423 Chr.4: 127983198-127983198 on Build GRCh38
rs143027224 Chr.4: 127983228-127983228 on Build GRCh38
rs746075557 Chr.4: 127982997-127982997 on Build GRCh38
rs767747606 Chr.4: 127983021-127983021 on Build GRCh38
rs370471176 Chr.4: 127983032-127983032 on Build GRCh38
rs764869945 Chr.4: 127982953-127982953 on Build GRCh38
rs149651574 Chr.4: 127983049-127983049 on Build GRCh38
rs751606816 Chr.4: 127983006-127983006 on Build GRCh38
rs546327744 Chr.4: 127983252-127983253 on Build GRCh38
rs753638475 Chr.4: 127982938-127982938 on Build GRCh38

View Full Product Details