Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 143337079-143337582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAB1
Entrez Gene ID
2549
Gene Name
GRB2 associated binding protein 1
Gene Aliases
-
UniGene
Hs.618456
Gene Chromosome Location
Chr.4: 143336830-143474565 on Build GRCh38
Transcripts
NM_002039.3 NM_207123.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1427309 c.44delC p.P17fs*5 Chr.4: 143337232-143337232 on Build GRCh38
4439606 c.10G>A p.G4S Chr.4: 143337198-143337198 on Build GRCh38
1427308 c.43_44insC p.E18fs*34 Chr.4: 143337231-143337232 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774361896 Chr.4: 143337174-143337174 on Build GRCh38
rs988199164 Chr.4: 143337375-143337376 on Build GRCh38
rs368003709 Chr.4: 143337300-143337300 on Build GRCh38
rs550277809 Chr.4: 143337344-143337344 on Build GRCh38
rs748459152 Chr.4: 143337152-143337152 on Build GRCh38
rs755581817 Chr.4: 143337231-143337232 on Build GRCh38
rs756903861 Chr.4: 143337385-143337385 on Build GRCh38
rs775444261 Chr.4: 143337295-143337295 on Build GRCh38
rs986352085 Chr.4: 143337180-143337180 on Build GRCh38
rs891901820 Chr.4: 143337113-143337113 on Build GRCh38
rs763453383 Chr.4: 143337167-143337167 on Build GRCh38
rs891188460 Chr.4: 143337357-143337357 on Build GRCh38
rs1025763571 Chr.4: 143337396-143337396 on Build GRCh38
rs916986591 Chr.4: 143337503-143337503 on Build GRCh38
rs1051192280 Chr.4: 143337346-143337346 on Build GRCh38
rs374071088 Chr.4: 143337283-143337283 on Build GRCh38
rs750183246 Chr.4: 143337306-143337306 on Build GRCh38
rs776463872 Chr.4: 143337145-143337145 on Build GRCh38
rs1024744017 Chr.4: 143337456-143337456 on Build GRCh38
rs564593551 Chr.4: 143337281-143337281 on Build GRCh38
rs777158611 Chr.4: 143337232-143337232 on Build GRCh38
rs1054451001 Chr.4: 143337111-143337111 on Build GRCh38
rs578237934 Chr.4: 143337267-143337267 on Build GRCh38
rs750189834 Chr.4: 143337233-143337233 on Build GRCh38
rs962955046 Chr.4: 143337392-143337392 on Build GRCh38
rs746487196 Chr.4: 143337237-143337237 on Build GRCh38
rs769792292 Chr.4: 143337258-143337258 on Build GRCh38
rs773589380 Chr.4: 143337272-143337272 on Build GRCh38
rs929496362 Chr.4: 143337466-143337466 on Build GRCh38
rs917637334 Chr.4: 143337215-143337215 on Build GRCh38
rs991912931 Chr.4: 143337200-143337200 on Build GRCh38
rs936618699 Chr.4: 143337554-143337554 on Build GRCh38
rs1044892973 Chr.4: 143337268-143337268 on Build GRCh38
rs770252546 Chr.4: 143337157-143337157 on Build GRCh38
rs751261243 Chr.4: 143337325-143337325 on Build GRCh38
rs572661306 Chr.4: 143337471-143337471 on Build GRCh38
rs368281444 Chr.4: 143337274-143337274 on Build GRCh38
rs758369672 Chr.4: 143337236-143337236 on Build GRCh38
rs904192486 Chr.4: 143337513-143337513 on Build GRCh38
rs13137712 Chr.4: 143337421-143337421 on Build GRCh38
rs1030277700 Chr.4: 143337128-143337128 on Build GRCh38
rs759605222 Chr.4: 143337293-143337293 on Build GRCh38
rs1056812701 Chr.4: 143337276-143337276 on Build GRCh38
rs919395283 Chr.4: 143337444-143337444 on Build GRCh38
rs767639491 Chr.4: 143337294-143337294 on Build GRCh38
rs879894314 Chr.4: 143337108-143337108 on Build GRCh38
rs753027403 Chr.4: 143337209-143337209 on Build GRCh38
rs781476200 Chr.4: 143337394-143337394 on Build GRCh38
rs368753740 Chr.4: 143337380-143337381 on Build GRCh38
rs756988560 Chr.4: 143337221-143337221 on Build GRCh38
rs545713738 Chr.4: 143337402-143337402 on Build GRCh38
rs1006974596 Chr.4: 143337148-143337148 on Build GRCh38
rs370794473 Chr.4: 143337277-143337277 on Build GRCh38
rs765022351 Chr.4: 143337232-143337232 on Build GRCh38
rs147010310 Chr.4: 143337239-143337239 on Build GRCh38
rs569753389 Chr.4: 143337122-143337122 on Build GRCh38
rs951085148 Chr.4: 143337407-143337407 on Build GRCh38
rs904424039 Chr.4: 143337309-143337309 on Build GRCh38
rs995768498 Chr.4: 143337393-143337393 on Build GRCh38
rs1031959091 Chr.4: 143337133-143337133 on Build GRCh38
rs543233957 Chr.4: 143337556-143337556 on Build GRCh38
rs759658067 Chr.4: 143337183-143337183 on Build GRCh38
rs962903912 Chr.4: 143337154-143337154 on Build GRCh38
rs767692877 Chr.4: 143337194-143337194 on Build GRCh38
rs1030432683 Chr.4: 143337377-143337377 on Build GRCh38
rs374296210 Chr.4: 143337171-143337171 on Build GRCh38
rs964945648 Chr.4: 143337136-143337136 on Build GRCh38
rs532144688 Chr.4: 143337288-143337288 on Build GRCh38
rs764253430 Chr.4: 143337304-143337304 on Build GRCh38
rs1000075588 Chr.4: 143337548-143337548 on Build GRCh38
rs200986459 Chr.4: 143337259-143337259 on Build GRCh38
rs769338188 Chr.4: 143337262-143337262 on Build GRCh38
rs1010318794 Chr.4: 143337124-143337124 on Build GRCh38
rs768402774 Chr.4: 143337140-143337140 on Build GRCh38
rs761019037 Chr.4: 143337391-143337391 on Build GRCh38
rs929024819 Chr.4: 143337245-143337245 on Build GRCh38
rs1044056848 Chr.4: 143337511-143337511 on Build GRCh38
rs948498906 Chr.4: 143337505-143337505 on Build GRCh38
rs980625868 Chr.4: 143337472-143337472 on Build GRCh38
rs891786659 Chr.4: 143337316-143337316 on Build GRCh38
rs1012551809 Chr.4: 143337298-143337298 on Build GRCh38
rs756590742 Chr.4: 143337545-143337545 on Build GRCh38
rs752462526 Chr.4: 143337156-143337156 on Build GRCh38
rs749679488 Chr.4: 143337273-143337273 on Build GRCh38
rs956802826 Chr.4: 143337156-143337156 on Build GRCh38
rs764039743 Chr.4: 143337369-143337369 on Build GRCh38
rs773465905 Chr.4: 143337158-143337158 on Build GRCh38

View Full Product Details