Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185175896-185176436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP97
Entrez Gene ID
57587
Gene Name
cilia and flagella associated protein 97
Gene Aliases
KIAA1430, hmw
UniGene
Hs.535734
Gene Chromosome Location
Chr.4: 185159662-185210251 on Build GRCh38
Transcripts
NM_001292033.1 NM_020827.2 XM_017008483.1 XM_017008484.1 XM_017008485.1 XM_017008486.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3825601 c.1205C>T p.A402V Chr.4: 185175901-185175901 on Build GRCh38
6694063 c.1096T>A p.F366I Chr.4: 185176010-185176010 on Build GRCh38
4695652 c.1191A>G p.E397E Chr.4: 185175915-185175915 on Build GRCh38
3130402 c.1077delA p.G360fs*12 Chr.4: 185176029-185176029 on Build GRCh38
1053751 c.1193T>C p.L398P Chr.4: 185175913-185175913 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373907107 Chr.4: 185175960-185175960 on Build GRCh38
rs540447166 Chr.4: 185176356-185176356 on Build GRCh38
rs567292410 Chr.4: 185175976-185175976 on Build GRCh38
rs976339005 Chr.4: 185176161-185176161 on Build GRCh38
rs71593605 Chr.4: 185176065-185176066 on Build GRCh38
rs896229587 Chr.4: 185176264-185176264 on Build GRCh38
rs113459393 Chr.4: 185176109-185176110 on Build GRCh38
rs1023277788 Chr.4: 185175959-185175959 on Build GRCh38
rs781506180 Chr.4: 185176135-185176135 on Build GRCh38
rs572936317 Chr.4: 185176197-185176197 on Build GRCh38
rs1016371870 Chr.4: 185176045-185176045 on Build GRCh38
rs978373976 Chr.4: 185176106-185176106 on Build GRCh38
rs936848370 Chr.4: 185176108-185176108 on Build GRCh38
rs767859346 Chr.4: 185176089-185176092 on Build GRCh38
rs368175136 Chr.4: 185176123-185176125 on Build GRCh38
rs570390457 Chr.4: 185176036-185176036 on Build GRCh38
rs780714040 Chr.4: 185175971-185175971 on Build GRCh38
rs373785037 Chr.4: 185176109-185176109 on Build GRCh38
rs200291624 Chr.4: 185176065-185176065 on Build GRCh38
rs778879098 Chr.4: 185176100-185176100 on Build GRCh38
rs77776340 Chr.4: 185176114-185176114 on Build GRCh38
rs1017664806 Chr.4: 185176354-185176354 on Build GRCh38
rs1002735468 Chr.4: 185176199-185176199 on Build GRCh38
rs770234795 Chr.4: 185176062-185176063 on Build GRCh38
rs760820247 Chr.4: 185175940-185175940 on Build GRCh38
rs965059149 Chr.4: 185176072-185176072 on Build GRCh38
rs773654144 Chr.4: 185176044-185176044 on Build GRCh38
rs924102994 Chr.4: 185176107-185176107 on Build GRCh38
rs58565485 Chr.4: 185176348-185176348 on Build GRCh38
rs377617878 Chr.4: 185175948-185175948 on Build GRCh38
rs779474320 Chr.4: 185176057-185176058 on Build GRCh38
rs751174957 Chr.4: 185176062-185176062 on Build GRCh38
rs561532648 Chr.4: 185176060-185176062 on Build GRCh38
rs757083787 Chr.4: 185176099-185176099 on Build GRCh38
rs762163186 Chr.4: 185176028-185176028 on Build GRCh38
rs777242941 Chr.4: 185175944-185175944 on Build GRCh38
rs370181930 Chr.4: 185176075-185176076 on Build GRCh38
rs752526312 Chr.4: 185175973-185175973 on Build GRCh38
rs747272096 Chr.4: 185175993-185175993 on Build GRCh38
rs772343101 Chr.4: 185175915-185175915 on Build GRCh38
rs907096753 Chr.4: 185176198-185176198 on Build GRCh38
rs386402610 Chr.4: 185176072-185176073 on Build GRCh38
rs368666785 Chr.4: 185176006-185176006 on Build GRCh38
rs368034368 Chr.4: 185176122-185176123 on Build GRCh38
rs201430524 Chr.4: 185176111-185176112 on Build GRCh38
rs1054600187 Chr.4: 185176119-185176119 on Build GRCh38
rs774067634 Chr.4: 185176044-185176054 on Build GRCh38
rs1027902776 Chr.4: 185176322-185176322 on Build GRCh38
rs887744322 Chr.4: 185176333-185176333 on Build GRCh38
rs1054851018 Chr.4: 185176263-185176263 on Build GRCh38
rs114641019 Chr.4: 185176123-185176123 on Build GRCh38
rs372105448 Chr.4: 185176103-185176103 on Build GRCh38
rs763355108 Chr.4: 185176047-185176047 on Build GRCh38
rs745592538 Chr.4: 185176028-185176029 on Build GRCh38
rs937709613 Chr.4: 185176247-185176247 on Build GRCh38
rs759778149 Chr.4: 185176067-185176068 on Build GRCh38
rs149817619 Chr.4: 185176285-185176285 on Build GRCh38
rs199637576 Chr.4: 185176012-185176012 on Build GRCh38
rs377061979 Chr.4: 185175963-185175963 on Build GRCh38
rs1030344340 Chr.4: 185176361-185176361 on Build GRCh38
rs763192015 Chr.4: 185176066-185176066 on Build GRCh38
rs755645483 Chr.4: 185176075-185176075 on Build GRCh38
rs913789836 Chr.4: 185176188-185176188 on Build GRCh38
rs978321493 Chr.4: 185176385-185176385 on Build GRCh38
rs556874914 Chr.4: 185175967-185175967 on Build GRCh38
rs979365661 Chr.4: 185176215-185176215 on Build GRCh38
rs370176540 Chr.4: 185175978-185175978 on Build GRCh38
rs200071745 Chr.4: 185176113-185176114 on Build GRCh38
rs766770816 Chr.4: 185176057-185176057 on Build GRCh38
rs765465709 Chr.4: 185175947-185175947 on Build GRCh38
rs869140723 Chr.4: 185176076-185176077 on Build GRCh38
rs375689229 Chr.4: 185175987-185175987 on Build GRCh38
rs752254911 Chr.4: 185176074-185176074 on Build GRCh38
rs558979650 Chr.4: 185176162-185176162 on Build GRCh38
rs1045889752 Chr.4: 185176288-185176288 on Build GRCh38
rs536319321 Chr.4: 185176068-185176068 on Build GRCh38
rs8180220 Chr.4: 185176130-185176130 on Build GRCh38
rs575857431 Chr.4: 185176241-185176241 on Build GRCh38
rs954733577 Chr.4: 185176378-185176378 on Build GRCh38
rs778399544 Chr.4: 185176011-185176012 on Build GRCh38
rs749104945 Chr.4: 185175986-185175986 on Build GRCh38
rs74754445 Chr.4: 185175945-185175945 on Build GRCh38
rs116235300 Chr.4: 185175957-185175957 on Build GRCh38
rs898026801 Chr.4: 185176299-185176299 on Build GRCh38
rs758760446 Chr.4: 185175955-185175955 on Build GRCh38
rs114119491 Chr.4: 185176126-185176126 on Build GRCh38
rs945259781 Chr.4: 185176191-185176191 on Build GRCh38
rs964841708 Chr.4: 185176350-185176350 on Build GRCh38
rs534539197 Chr.4: 185175922-185175922 on Build GRCh38
rs553140338 Chr.4: 185176121-185176121 on Build GRCh38
rs924219150 Chr.4: 185176413-185176413 on Build GRCh38
rs993693540 Chr.4: 185176311-185176311 on Build GRCh38
rs369749223 Chr.4: 185175941-185175941 on Build GRCh38
rs753764336 Chr.4: 185176095-185176095 on Build GRCh38
rs140276553 Chr.4: 185176076-185176076 on Build GRCh38
rs1007974512 Chr.4: 185176349-185176349 on Build GRCh38
rs1044073415 Chr.4: 185176164-185176164 on Build GRCh38
rs376848813 Chr.4: 185176125-185176125 on Build GRCh38
rs560915183 Chr.4: 185176287-185176287 on Build GRCh38
rs770333893 Chr.4: 185176037-185176037 on Build GRCh38

View Full Product Details