Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 24504920-24505439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH10
Entrez Gene ID
1008
Gene Name
cadherin 10
Gene Aliases
-
UniGene
Hs.92489
Gene Chromosome Location
Chr.5: 24487100-24644978 on Build GRCh38
Transcripts
NM_001317222.1 NM_001317224.1 NM_006727.4 XM_011513923.2 XM_017008915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5647406 c.1257-1G>C p.? Chr.5: 24505249-24505249 on Build GRCh38
387532 c.1283T>A p.L428H Chr.5: 24505222-24505222 on Build GRCh38
1311081 c.1293C>G p.I431M Chr.5: 24505212-24505212 on Build GRCh38
6624583 c.1316G>A p.G439E Chr.5: 24505189-24505189 on Build GRCh38
4997650 c.1283T>G p.L428R Chr.5: 24505222-24505222 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893124389 Chr.5: 24505398-24505398 on Build GRCh38
rs963425811 Chr.5: 24505350-24505350 on Build GRCh38
rs1026246145 Chr.5: 24505366-24505366 on Build GRCh38
rs919547706 Chr.5: 24504994-24504994 on Build GRCh38
rs745687203 Chr.5: 24505296-24505296 on Build GRCh38
rs13177908 Chr.5: 24504962-24504962 on Build GRCh38
rs938435391 Chr.5: 24505120-24505120 on Build GRCh38
rs760235178 Chr.5: 24505214-24505214 on Build GRCh38
rs952407748 Chr.5: 24505325-24505325 on Build GRCh38
rs951055436 Chr.5: 24505367-24505367 on Build GRCh38
rs868847730 Chr.5: 24505313-24505313 on Build GRCh38
rs973366216 Chr.5: 24504961-24504961 on Build GRCh38
rs149199529 Chr.5: 24505152-24505152 on Build GRCh38
rs746340617 Chr.5: 24505122-24505122 on Build GRCh38
rs376724275 Chr.5: 24505289-24505289 on Build GRCh38
rs770913066 Chr.5: 24505168-24505168 on Build GRCh38
rs555555015 Chr.5: 24505241-24505241 on Build GRCh38
rs1026598320 Chr.5: 24505308-24505308 on Build GRCh38
rs537790264 Chr.5: 24505405-24505405 on Build GRCh38
rs566574417 Chr.5: 24505297-24505297 on Build GRCh38
rs13178070 Chr.5: 24505025-24505025 on Build GRCh38
rs910768272 Chr.5: 24505353-24505353 on Build GRCh38
rs144680689 Chr.5: 24505262-24505262 on Build GRCh38
rs184607550 Chr.5: 24505015-24505015 on Build GRCh38
rs755023658 Chr.5: 24505133-24505133 on Build GRCh38
rs372226944 Chr.5: 24505208-24505208 on Build GRCh38
rs947351313 Chr.5: 24505399-24505399 on Build GRCh38
rs567310947 Chr.5: 24505383-24505383 on Build GRCh38
rs751846268 Chr.5: 24505121-24505121 on Build GRCh38
rs767814386 Chr.5: 24505273-24505273 on Build GRCh38
rs1023327185 Chr.5: 24505106-24505106 on Build GRCh38
rs746488894 Chr.5: 24505299-24505299 on Build GRCh38
rs187087282 Chr.5: 24505018-24505018 on Build GRCh38
rs373822384 Chr.5: 24505288-24505288 on Build GRCh38
rs894221809 Chr.5: 24505130-24505130 on Build GRCh38
rs763303674 Chr.5: 24505258-24505258 on Build GRCh38
rs774088289 Chr.5: 24505390-24505390 on Build GRCh38
rs766576483 Chr.5: 24505119-24505119 on Build GRCh38
rs778672806 Chr.5: 24505169-24505169 on Build GRCh38
rs866850339 Chr.5: 24505202-24505202 on Build GRCh38
rs763322190 Chr.5: 24505094-24505094 on Build GRCh38
rs934726433 Chr.5: 24505386-24505386 on Build GRCh38
rs903128657 Chr.5: 24505223-24505223 on Build GRCh38
rs745564475 Chr.5: 24505185-24505185 on Build GRCh38
rs985039983 Chr.5: 24505348-24505348 on Build GRCh38
rs1053108250 Chr.5: 24505005-24505005 on Build GRCh38
rs375518042 Chr.5: 24505151-24505151 on Build GRCh38
rs901896192 Chr.5: 24504954-24504954 on Build GRCh38
rs780952857 Chr.5: 24505266-24505267 on Build GRCh38
rs13178076 Chr.5: 24505049-24505049 on Build GRCh38
rs572587062 Chr.5: 24504998-24504998 on Build GRCh38
rs13174039 Chr.5: 24505129-24505129 on Build GRCh38
rs376630462 Chr.5: 24505248-24505248 on Build GRCh38
rs1017675736 Chr.5: 24505317-24505317 on Build GRCh38
rs13177916 Chr.5: 24504968-24504968 on Build GRCh38
rs554217401 Chr.5: 24504971-24504971 on Build GRCh38
rs973397296 Chr.5: 24505358-24505358 on Build GRCh38
rs963312394 Chr.5: 24504942-24504942 on Build GRCh38
rs752659067 Chr.5: 24505276-24505276 on Build GRCh38
rs943644397 Chr.5: 24505365-24505365 on Build GRCh38
rs544116954 Chr.5: 24505146-24505146 on Build GRCh38
rs551532614 Chr.5: 24505283-24505283 on Build GRCh38
rs771873502 Chr.5: 24505196-24505196 on Build GRCh38
rs762762281 Chr.5: 24505360-24505360 on Build GRCh38
rs13178071 Chr.5: 24505034-24505034 on Build GRCh38
rs111297824 Chr.5: 24505292-24505292 on Build GRCh38
rs770143615 Chr.5: 24505234-24505234 on Build GRCh38
rs202215608 Chr.5: 24505298-24505298 on Build GRCh38
rs982702392 Chr.5: 24505044-24505044 on Build GRCh38
rs765803427 Chr.5: 24505086-24505086 on Build GRCh38
rs756273162 Chr.5: 24505282-24505282 on Build GRCh38
rs13178067 Chr.5: 24505020-24505020 on Build GRCh38
rs377199667 Chr.5: 24505068-24505068 on Build GRCh38
rs13173886 Chr.5: 24505052-24505052 on Build GRCh38
rs1042728194 Chr.5: 24505180-24505180 on Build GRCh38
rs138563115 Chr.5: 24505381-24505381 on Build GRCh38
rs749021068 Chr.5: 24505159-24505159 on Build GRCh38
rs777777744 Chr.5: 24505158-24505158 on Build GRCh38
rs112158914 Chr.5: 24505411-24505411 on Build GRCh38
rs762177341 Chr.5: 24505078-24505078 on Build GRCh38
rs759709217 Chr.5: 24505269-24505269 on Build GRCh38
rs1040562024 Chr.5: 24504951-24504951 on Build GRCh38
rs990984493 Chr.5: 24505403-24505403 on Build GRCh38

View Full Product Details