Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 36680496-36681006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC1A3
Entrez Gene ID
6507
Gene Name
solute carrier family 1 member 3
Gene Aliases
EA6, EAAT1, GLAST, GLAST1
UniGene
Hs.481918
Gene Chromosome Location
Chr.5: 36606355-36688334 on Build GRCh38
Transcripts
NM_001166695.2 NM_001289939.1 NM_001289940.1 NM_004172.4 XM_005248342.2 XM_011514084.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6512975 c.1196T>A p.M399K Chr.5: 36680496-36680496 on Build GRCh38
6170975 c.1227T>A p.A409A Chr.5: 36680527-36680527 on Build GRCh38
1437252 c.1269C>T p.F423F Chr.5: 36680569-36680569 on Build GRCh38
6546156 c.1226C>T p.A409V Chr.5: 36680526-36680526 on Build GRCh38
1695611 c.1204A>G p.T402A Chr.5: 36680504-36680504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927005907 Chr.5: 36680666-36680666 on Build GRCh38
rs971289403 Chr.5: 36680880-36680880 on Build GRCh38
rs763237927 Chr.5: 36680583-36680583 on Build GRCh38
rs777262480 Chr.5: 36680762-36680762 on Build GRCh38
rs528412871 Chr.5: 36680594-36680594 on Build GRCh38
rs971774674 Chr.5: 36680979-36680979 on Build GRCh38
rs373832946 Chr.5: 36680614-36680614 on Build GRCh38
rs889162943 Chr.5: 36680791-36680791 on Build GRCh38
rs796729954 Chr.5: 36680915-36680916 on Build GRCh38
rs937477956 Chr.5: 36680932-36680932 on Build GRCh38
rs374310289 Chr.5: 36680637-36680637 on Build GRCh38
rs1036115593 Chr.5: 36680718-36680718 on Build GRCh38
rs374779736 Chr.5: 36680788-36680788 on Build GRCh38
rs534252145 Chr.5: 36680781-36680781 on Build GRCh38
rs557272227 Chr.5: 36680761-36680761 on Build GRCh38
rs573479293 Chr.5: 36680760-36680760 on Build GRCh38
rs752225936 Chr.5: 36680613-36680613 on Build GRCh38
rs958184688 Chr.5: 36680921-36680921 on Build GRCh38
rs940535955 Chr.5: 36680711-36680711 on Build GRCh38
rs10710837 Chr.5: 36680931-36680931 on Build GRCh38
rs929629975 Chr.5: 36680642-36680642 on Build GRCh38
rs745942837 Chr.5: 36680559-36680559 on Build GRCh38
rs535738667 Chr.5: 36680689-36680689 on Build GRCh38
rs1009759049 Chr.5: 36680852-36680852 on Build GRCh38
rs550678656 Chr.5: 36680656-36680656 on Build GRCh38
rs3822470 Chr.5: 36680915-36680915 on Build GRCh38
rs780717613 Chr.5: 36680688-36680688 on Build GRCh38
rs563374163 Chr.5: 36680893-36680893 on Build GRCh38
rs1032437824 Chr.5: 36680842-36680842 on Build GRCh38
rs751041640 Chr.5: 36680604-36680604 on Build GRCh38
rs769918067 Chr.5: 36680561-36680561 on Build GRCh38
rs1046474343 Chr.5: 36680696-36680696 on Build GRCh38
rs901770975 Chr.5: 36680754-36680754 on Build GRCh38
rs1000862577 Chr.5: 36680826-36680826 on Build GRCh38
rs371379106 Chr.5: 36680915-36680915 on Build GRCh38
rs7716057 Chr.5: 36680629-36680629 on Build GRCh38
rs577580501 Chr.5: 36680843-36680843 on Build GRCh38
rs376105109 Chr.5: 36680632-36680632 on Build GRCh38
rs781147237 Chr.5: 36680539-36680539 on Build GRCh38
rs200947079 Chr.5: 36680584-36680584 on Build GRCh38
rs554801551 Chr.5: 36680681-36680681 on Build GRCh38
rs370028507 Chr.5: 36680597-36680597 on Build GRCh38
rs190941111 Chr.5: 36680855-36680855 on Build GRCh38
rs918177606 Chr.5: 36680646-36680646 on Build GRCh38
rs185488774 Chr.5: 36680792-36680792 on Build GRCh38
rs940420077 Chr.5: 36680957-36680957 on Build GRCh38
rs569081853 Chr.5: 36680663-36680663 on Build GRCh38
rs759106070 Chr.5: 36680609-36680609 on Build GRCh38
rs970978604 Chr.5: 36680658-36680658 on Build GRCh38
rs796678747 Chr.5: 36680916-36680916 on Build GRCh38
rs758802146 Chr.5: 36680569-36680569 on Build GRCh38
rs376479451 Chr.5: 36680596-36680596 on Build GRCh38
rs373010365 Chr.5: 36680781-36680782 on Build GRCh38
rs869295514 Chr.5: 36680915-36680916 on Build GRCh38
rs958397636 Chr.5: 36680917-36680917 on Build GRCh38
rs113193520 Chr.5: 36680894-36680894 on Build GRCh38
rs3733811 Chr.5: 36680611-36680611 on Build GRCh38
rs528367885 Chr.5: 36680977-36680977 on Build GRCh38
rs768110692 Chr.5: 36680602-36680602 on Build GRCh38
rs999502125 Chr.5: 36680815-36680815 on Build GRCh38
rs200788327 Chr.5: 36680916-36680916 on Build GRCh38
rs536452217 Chr.5: 36680675-36680675 on Build GRCh38
rs187427195 Chr.5: 36680946-36680946 on Build GRCh38
rs1032193433 Chr.5: 36680911-36680911 on Build GRCh38
rs62356362 Chr.5: 36680949-36680949 on Build GRCh38
rs908957503 Chr.5: 36680698-36680698 on Build GRCh38

View Full Product Details