Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 38940793-38941296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSMR
Entrez Gene ID
9180
Gene Name
oncostatin M receptor
Gene Aliases
IL-31R-beta, IL-31RB, OSMRB, PLCA1
UniGene
Hs.120658
Gene Chromosome Location
Chr.5: 38845858-38945579 on Build GRCh38
Transcripts
XM_011514161.2 XM_017010019.1 NM_001285439.1 NM_001285440.1 NM_152756.4 XM_006714463.3 XM_011514005.2 XM_011514006.2 XM_017009311.1 XM_017009312.1 XM_017009313.1 XM_017009314.1 XM_017009315.1

Gene Symbol
RICTOR
Entrez Gene ID
253260
Gene Name
RPTOR independent companion of MTOR complex 2
Gene Aliases
AVO3, PIA, hAVO3
UniGene
Hs.407926
Gene Chromosome Location
Chr.5: 38937920-39074421 on Build GRCh38
Transcripts
XM_011514161.2 XM_017010019.1 NM_001285439.1 NM_001285440.1 NM_152756.4 XM_006714463.3 XM_011514005.2 XM_011514006.2 XM_017009311.1 XM_017009312.1 XM_017009313.1 XM_017009314.1 XM_017009315.1

SNP Detail

SNP ID Chromosome Location
rs990512251 Chr.5: 38941143-38941143 on Build GRCh38
rs1031923547 Chr.5: 38941117-38941117 on Build GRCh38
rs930501693 Chr.5: 38941198-38941198 on Build GRCh38
rs45601539 Chr.5: 38941176-38941176 on Build GRCh38
rs544164662 Chr.5: 38941123-38941123 on Build GRCh38
rs956485613 Chr.5: 38941120-38941120 on Build GRCh38
rs994244178 Chr.5: 38940975-38940975 on Build GRCh38
rs979841534 Chr.5: 38941191-38941191 on Build GRCh38
rs928386528 Chr.5: 38941183-38941183 on Build GRCh38
rs948325082 Chr.5: 38941238-38941238 on Build GRCh38
rs944607890 Chr.5: 38941219-38941219 on Build GRCh38
rs188773716 Chr.5: 38941212-38941212 on Build GRCh38
rs996916109 Chr.5: 38940976-38940981 on Build GRCh38
rs1041882071 Chr.5: 38940900-38940900 on Build GRCh38
rs983652313 Chr.5: 38941205-38941205 on Build GRCh38
rs140589719 Chr.5: 38941221-38941221 on Build GRCh38
rs1018115597 Chr.5: 38941116-38941116 on Build GRCh38
rs115757729 Chr.5: 38941124-38941124 on Build GRCh38
rs898527912 Chr.5: 38940956-38940956 on Build GRCh38
rs914225079 Chr.5: 38941145-38941145 on Build GRCh38
rs1012627253 Chr.5: 38940946-38940946 on Build GRCh38
rs904703144 Chr.5: 38941091-38941091 on Build GRCh38
rs895402352 Chr.5: 38940935-38940935 on Build GRCh38
rs753646537 Chr.5: 38940970-38940970 on Build GRCh38
rs540699269 Chr.5: 38941260-38941260 on Build GRCh38
rs1041057396 Chr.5: 38940934-38940934 on Build GRCh38
rs1028388312 Chr.5: 38941007-38941007 on Build GRCh38
rs184649170 Chr.5: 38940883-38940883 on Build GRCh38
rs571835787 Chr.5: 38941118-38941118 on Build GRCh38
rs528177561 Chr.5: 38940890-38940890 on Build GRCh38
rs986601154 Chr.5: 38941087-38941087 on Build GRCh38
rs554533555 Chr.5: 38941217-38941217 on Build GRCh38
rs895337855 Chr.5: 38941216-38941216 on Build GRCh38
rs1016926418 Chr.5: 38940947-38940947 on Build GRCh38
rs1025711585 Chr.5: 38940996-38940996 on Build GRCh38
rs902671483 Chr.5: 38940905-38940905 on Build GRCh38
rs401259 Chr.5: 38941207-38941207 on Build GRCh38
rs577263429 Chr.5: 38940915-38940915 on Build GRCh38
rs945732760 Chr.5: 38941167-38941167 on Build GRCh38
rs955187088 Chr.5: 38941077-38941077 on Build GRCh38
rs35737255 Chr.5: 38941004-38941004 on Build GRCh38
rs1004221224 Chr.5: 38940931-38940931 on Build GRCh38
rs1023913937 Chr.5: 38941151-38941152 on Build GRCh38
rs377245251 Chr.5: 38940971-38940971 on Build GRCh38
rs1044239324 Chr.5: 38940850-38940851 on Build GRCh38
rs769629957 Chr.5: 38941197-38941197 on Build GRCh38
rs779875867 Chr.5: 38940920-38940920 on Build GRCh38

View Full Product Details