Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55767698-55768110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX4
Entrez Gene ID
54514
Gene Name
DEAD-box helicase 4
Gene Aliases
VASA
UniGene
Hs.223581
Gene Chromosome Location
Chr.5: 55737994-55817146 on Build GRCh38
Transcripts
NM_001142549.1 NM_001166533.1 NM_001166534.1 NM_024415.2 XM_011543495.2 XM_011543497.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5715759 c.64C>T p.P22S Chr.5: 55767907-55767907 on Build GRCh38
6836492 c.376G>C p.G126R Chr.5: 55767922-55767922 on Build GRCh38
1582658 c.367C>T p.R123W Chr.5: 55767913-55767913 on Build GRCh38
1582657 c.70C>T p.R24W Chr.5: 55767913-55767913 on Build GRCh38
1595409 c.55G>A p.E19K Chr.5: 55767898-55767898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374972771 Chr.5: 55767913-55767913 on Build GRCh38
rs370258736 Chr.5: 55768084-55768086 on Build GRCh38
rs530007635 Chr.5: 55767754-55767754 on Build GRCh38
rs143793571 Chr.5: 55767889-55767889 on Build GRCh38
rs769267053 Chr.5: 55767930-55767930 on Build GRCh38
rs959575020 Chr.5: 55767803-55767803 on Build GRCh38
rs968701737 Chr.5: 55767838-55767838 on Build GRCh38
rs570373422 Chr.5: 55767921-55767921 on Build GRCh38
rs764868412 Chr.5: 55767995-55767995 on Build GRCh38
rs762924791 Chr.5: 55768011-55768011 on Build GRCh38
rs114582735 Chr.5: 55767993-55767993 on Build GRCh38
rs777600910 Chr.5: 55767896-55767896 on Build GRCh38
rs781432280 Chr.5: 55767902-55767902 on Build GRCh38
rs753321139 Chr.5: 55767832-55767832 on Build GRCh38
rs978841789 Chr.5: 55767849-55767849 on Build GRCh38
rs759272098 Chr.5: 55767781-55767781 on Build GRCh38
rs1012151351 Chr.5: 55767818-55767818 on Build GRCh38
rs60640325 Chr.5: 55768015-55768015 on Build GRCh38
rs112518640 Chr.5: 55767772-55767772 on Build GRCh38
rs765099488 Chr.5: 55767852-55767852 on Build GRCh38
rs373279272 Chr.5: 55767958-55767958 on Build GRCh38
rs770585245 Chr.5: 55767723-55767726 on Build GRCh38
rs751383476 Chr.5: 55768033-55768033 on Build GRCh38
rs141132406 Chr.5: 55767910-55767910 on Build GRCh38
rs956252606 Chr.5: 55767901-55767901 on Build GRCh38
rs888106116 Chr.5: 55767729-55767729 on Build GRCh38
rs537763609 Chr.5: 55767984-55767984 on Build GRCh38
rs756155094 Chr.5: 55767912-55767912 on Build GRCh38
rs148169621 Chr.5: 55767897-55767897 on Build GRCh38
rs774515015 Chr.5: 55767831-55767838 on Build GRCh38
rs774966145 Chr.5: 55768004-55768004 on Build GRCh38
rs1023004405 Chr.5: 55767819-55767819 on Build GRCh38
rs548510543 Chr.5: 55767798-55767798 on Build GRCh38
rs776289883 Chr.5: 55767961-55767961 on Build GRCh38
rs928507586 Chr.5: 55768064-55768064 on Build GRCh38
rs369208045 Chr.5: 55767957-55767957 on Build GRCh38
rs202158802 Chr.5: 55767865-55767865 on Build GRCh38
rs533503015 Chr.5: 55768083-55768085 on Build GRCh38
rs759576360 Chr.5: 55767863-55767863 on Build GRCh38
rs758326078 Chr.5: 55767894-55767894 on Build GRCh38
rs748507209 Chr.5: 55767939-55767939 on Build GRCh38
rs79005593 Chr.5: 55767943-55767943 on Build GRCh38
rs922782259 Chr.5: 55767923-55767923 on Build GRCh38
rs759037569 Chr.5: 55767834-55767834 on Build GRCh38
rs767557836 Chr.5: 55767970-55767971 on Build GRCh38
rs757458025 Chr.5: 55767898-55767898 on Build GRCh38
rs757024582 Chr.5: 55768034-55768034 on Build GRCh38
rs896886345 Chr.5: 55768047-55768047 on Build GRCh38
rs749870232 Chr.5: 55767914-55767914 on Build GRCh38
rs1005297959 Chr.5: 55767731-55767731 on Build GRCh38
rs766025305 Chr.5: 55767830-55767830 on Build GRCh38
rs74473274 Chr.5: 55767932-55767932 on Build GRCh38
rs1008492282 Chr.5: 55767785-55767787 on Build GRCh38
rs765190927 Chr.5: 55768016-55768016 on Build GRCh38
rs897000845 Chr.5: 55767771-55767771 on Build GRCh38

View Full Product Details