Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75663316-75663823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKDD1B
Entrez Gene ID
728780
Gene Name
ankyrin repeat and death domain containing 1B
Gene Aliases
-
UniGene
Hs.370455
Gene Chromosome Location
Chr.5: 75611289-75671846 on Build GRCh38
Transcripts
NM_001276713.1 XM_011543618.2 XM_011543623.2 XM_017009814.1 XM_017009815.1 XM_017009816.1 XM_017009817.1

SNP Detail

SNP ID Chromosome Location
rs948785636 Chr.5: 75663628-75663628 on Build GRCh38
rs1041824330 Chr.5: 75663397-75663397 on Build GRCh38
rs889079836 Chr.5: 75663736-75663736 on Build GRCh38
rs977768421 Chr.5: 75663780-75663780 on Build GRCh38
rs1047049564 Chr.5: 75663673-75663673 on Build GRCh38
rs1036476020 Chr.5: 75663703-75663703 on Build GRCh38
rs568449542 Chr.5: 75663418-75663418 on Build GRCh38
rs542593419 Chr.5: 75663574-75663574 on Build GRCh38
rs1007335502 Chr.5: 75663718-75663718 on Build GRCh38
rs147988502 Chr.5: 75663653-75663653 on Build GRCh38
rs776665444 Chr.5: 75663507-75663508 on Build GRCh38
rs1017744098 Chr.5: 75663719-75663719 on Build GRCh38
rs1055575696 Chr.5: 75663650-75663650 on Build GRCh38
rs906663417 Chr.5: 75663464-75663464 on Build GRCh38
rs6869286 Chr.5: 75663395-75663395 on Build GRCh38
rs993198552 Chr.5: 75663701-75663701 on Build GRCh38
rs1019254216 Chr.5: 75663762-75663763 on Build GRCh38
rs977299371 Chr.5: 75663640-75663640 on Build GRCh38
rs935627305 Chr.5: 75663415-75663415 on Build GRCh38
rs539814621 Chr.5: 75663728-75663728 on Build GRCh38
rs1037658546 Chr.5: 75663463-75663463 on Build GRCh38
rs574391011 Chr.5: 75663713-75663713 on Build GRCh38
rs893219894 Chr.5: 75663355-75663355 on Build GRCh38
rs907468995 Chr.5: 75663428-75663428 on Build GRCh38
rs887550074 Chr.5: 75663717-75663717 on Build GRCh38
rs1031622246 Chr.5: 75663752-75663752 on Build GRCh38
rs956078414 Chr.5: 75663591-75663591 on Build GRCh38
rs775378137 Chr.5: 75663401-75663401 on Build GRCh38
rs997755186 Chr.5: 75663749-75663749 on Build GRCh38
rs897499820 Chr.5: 75663482-75663482 on Build GRCh38
rs185737795 Chr.5: 75663570-75663570 on Build GRCh38
rs1027447406 Chr.5: 75663520-75663520 on Build GRCh38
rs746896432 Chr.5: 75663506-75663506 on Build GRCh38
rs914687258 Chr.5: 75663609-75663609 on Build GRCh38
rs529179593 Chr.5: 75663556-75663557 on Build GRCh38
rs773939447 Chr.5: 75663531-75663531 on Build GRCh38
rs965907256 Chr.5: 75663746-75663746 on Build GRCh38
rs955943057 Chr.5: 75663768-75663768 on Build GRCh38
rs554714584 Chr.5: 75663412-75663412 on Build GRCh38
rs925694326 Chr.5: 75663413-75663414 on Build GRCh38
rs544941781 Chr.5: 75663704-75663704 on Build GRCh38
rs948922713 Chr.5: 75663387-75663387 on Build GRCh38
rs897398870 Chr.5: 75663698-75663698 on Build GRCh38
rs1055481271 Chr.5: 75663417-75663417 on Build GRCh38
rs938813396 Chr.5: 75663694-75663694 on Build GRCh38
rs188662605 Chr.5: 75663435-75663435 on Build GRCh38
rs554326179 Chr.5: 75663555-75663555 on Build GRCh38
rs1029067632 Chr.5: 75663732-75663732 on Build GRCh38
rs967331688 Chr.5: 75663765-75663765 on Build GRCh38
rs966248394 Chr.5: 75663572-75663572 on Build GRCh38
rs934483146 Chr.5: 75663702-75663716 on Build GRCh38
rs1037291340 Chr.5: 75663696-75663696 on Build GRCh38
rs994911441 Chr.5: 75663549-75663549 on Build GRCh38
rs746116446 Chr.5: 75663594-75663594 on Build GRCh38
rs987786519 Chr.5: 75663561-75663561 on Build GRCh38
rs1007428729 Chr.5: 75663538-75663538 on Build GRCh38
rs1005056159 Chr.5: 75663467-75663467 on Build GRCh38
rs925661246 Chr.5: 75663645-75663645 on Build GRCh38
rs189586752 Chr.5: 75663646-75663646 on Build GRCh38
rs193015492 Chr.5: 75663558-75663558 on Build GRCh38
rs987430305 Chr.5: 75663365-75663365 on Build GRCh38
rs1015401167 Chr.5: 75663510-75663510 on Build GRCh38
rs146955921 Chr.5: 75663584-75663584 on Build GRCh38
rs949756576 Chr.5: 75663659-75663659 on Build GRCh38
rs951770689 Chr.5: 75663533-75663533 on Build GRCh38
rs1012893291 Chr.5: 75663422-75663422 on Build GRCh38

View Full Product Details