Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 112164977-112165482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L4A
Entrez Gene ID
64097
Gene Name
erythrocyte membrane protein band 4.1 like 4A
Gene Aliases
EPB41L4, NBL4
UniGene
Hs.584954
Gene Chromosome Location
Chr.5: 112141829-112419490 on Build GRCh38
Transcripts
NM_022140.3 XM_011543530.1 XM_011543531.2 XM_011543532.1 XM_011543533.1 XM_017009689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
448430 c.2018_2020delCAA p.T673delT Chr.5: 112165031-112165033 on Build GRCh38
4127084 c.2029A>G p.I677V Chr.5: 112165022-112165022 on Build GRCh38
5623528 c.1934G>T p.R645I Chr.5: 112165117-112165117 on Build GRCh38
4127083 c.2046C>T p.L682L Chr.5: 112165005-112165005 on Build GRCh38
5989591 c.1933-2_1933-1insTTA p.? Chr.5: 112165119-112165120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879645698 Chr.5: 112165213-112165213 on Build GRCh38
rs200449454 Chr.5: 112165095-112165095 on Build GRCh38
rs774146701 Chr.5: 112165117-112165117 on Build GRCh38
rs552383923 Chr.5: 112165207-112165207 on Build GRCh38
rs372333096 Chr.5: 112165039-112165039 on Build GRCh38
rs531074634 Chr.5: 112165199-112165199 on Build GRCh38
rs764305656 Chr.5: 112165121-112165122 on Build GRCh38
rs904384563 Chr.5: 112165238-112165238 on Build GRCh38
rs757818794 Chr.5: 112165146-112165146 on Build GRCh38
rs546980844 Chr.5: 112165342-112165342 on Build GRCh38
rs768227870 Chr.5: 112165103-112165103 on Build GRCh38
rs542437030 Chr.5: 112165143-112165143 on Build GRCh38
rs761462030 Chr.5: 112165023-112165023 on Build GRCh38
rs779627577 Chr.5: 112165168-112165168 on Build GRCh38
rs959892240 Chr.5: 112165007-112165007 on Build GRCh38
rs772726022 Chr.5: 112165033-112165034 on Build GRCh38
rs184684511 Chr.5: 112165033-112165033 on Build GRCh38
rs576427101 Chr.5: 112165036-112165036 on Build GRCh38
rs937036521 Chr.5: 112165192-112165192 on Build GRCh38
rs145708081 Chr.5: 112165119-112165120 on Build GRCh38
rs781666158 Chr.5: 112165051-112165051 on Build GRCh38
rs200146895 Chr.5: 112165031-112165031 on Build GRCh38
rs762850623 Chr.5: 112165012-112165012 on Build GRCh38
rs1000093771 Chr.5: 112165239-112165239 on Build GRCh38
rs759417132 Chr.5: 112165112-112165119 on Build GRCh38
rs923317190 Chr.5: 112165058-112165058 on Build GRCh38
rs78027176 Chr.5: 112165127-112165127 on Build GRCh38
rs747295977 Chr.5: 112165001-112165001 on Build GRCh38
rs908065015 Chr.5: 112165102-112165102 on Build GRCh38
rs747562842 Chr.5: 112165041-112165041 on Build GRCh38
rs988912592 Chr.5: 112165201-112165201 on Build GRCh38
rs776458709 Chr.5: 112165079-112165080 on Build GRCh38
rs767512690 Chr.5: 112165206-112165206 on Build GRCh38
rs746268329 Chr.5: 112165156-112165156 on Build GRCh38
rs1551935 Chr.5: 112165054-112165054 on Build GRCh38
rs201809605 Chr.5: 112165009-112165009 on Build GRCh38
rs911400529 Chr.5: 112165255-112165255 on Build GRCh38
rs576117395 Chr.5: 112165002-112165002 on Build GRCh38
rs1056432951 Chr.5: 112165319-112165319 on Build GRCh38
rs955746333 Chr.5: 112165174-112165174 on Build GRCh38
rs143889091 Chr.5: 112165294-112165294 on Build GRCh38
rs896484601 Chr.5: 112165347-112165347 on Build GRCh38
rs566004334 Chr.5: 112165367-112165367 on Build GRCh38
rs746179708 Chr.5: 112165160-112165160 on Build GRCh38
rs200807148 Chr.5: 112165118-112165118 on Build GRCh38
rs3067637 Chr.5: 112165128-112165129 on Build GRCh38
rs752971301 Chr.5: 112165131-112165131 on Build GRCh38
rs1000408599 Chr.5: 112165456-112165456 on Build GRCh38
rs771699308 Chr.5: 112165002-112165003 on Build GRCh38
rs3067634 Chr.5: 112165126-112165127 on Build GRCh38
rs563714723 Chr.5: 112165150-112165150 on Build GRCh38
rs754005178 Chr.5: 112165027-112165027 on Build GRCh38
rs532502547 Chr.5: 112165318-112165319 on Build GRCh38
rs759243348 Chr.5: 112165008-112165008 on Build GRCh38
rs763967074 Chr.5: 112165020-112165020 on Build GRCh38
rs1022797975 Chr.5: 112165386-112165386 on Build GRCh38
rs33929905 Chr.5: 112165127-112165128 on Build GRCh38
rs373187409 Chr.5: 112165125-112165126 on Build GRCh38
rs758492611 Chr.5: 112165132-112165132 on Build GRCh38
rs780144220 Chr.5: 112165394-112165394 on Build GRCh38
rs78887676 Chr.5: 112165286-112165286 on Build GRCh38
rs776087947 Chr.5: 112165005-112165005 on Build GRCh38
rs367564664 Chr.5: 112165010-112165010 on Build GRCh38
rs752431836 Chr.5: 112165167-112165167 on Build GRCh38
rs189618155 Chr.5: 112165383-112165383 on Build GRCh38
rs756499229 Chr.5: 112165162-112165162 on Build GRCh38
rs201659688 Chr.5: 112165129-112165129 on Build GRCh38
rs879099117 Chr.5: 112165048-112165048 on Build GRCh38
rs755673108 Chr.5: 112165309-112165321 on Build GRCh38
rs745486168 Chr.5: 112165053-112165053 on Build GRCh38
rs771185706 Chr.5: 112165050-112165050 on Build GRCh38
rs568177274 Chr.5: 112165043-112165043 on Build GRCh38
rs941485183 Chr.5: 112165268-112165268 on Build GRCh38
rs1030353092 Chr.5: 112165320-112165323 on Build GRCh38
rs754339786 Chr.5: 112165056-112165056 on Build GRCh38
rs764291654 Chr.5: 112165137-112165137 on Build GRCh38
rs751523217 Chr.5: 112165022-112165022 on Build GRCh38
rs895528079 Chr.5: 112165204-112165209 on Build GRCh38
rs371801383 Chr.5: 112165029-112165029 on Build GRCh38
rs1014125487 Chr.5: 112165070-112165070 on Build GRCh38
rs766129916 Chr.5: 112165060-112165063 on Build GRCh38
rs930061529 Chr.5: 112165310-112165310 on Build GRCh38
rs771620451 Chr.5: 112165059-112165061 on Build GRCh38
rs959974023 Chr.5: 112165063-112165063 on Build GRCh38
rs752004934 Chr.5: 112165144-112165144 on Build GRCh38
rs1049995630 Chr.5: 112165220-112165220 on Build GRCh38
rs922243714 Chr.5: 112165211-112165211 on Build GRCh38
rs758226317 Chr.5: 112165346-112165346 on Build GRCh38

View Full Product Details