Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 129748101-129748600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1024L
Entrez Gene ID
100127206
Gene Name
KIAA1024 like
Gene Aliases
-
UniGene
Hs.582534
Gene Chromosome Location
Chr.5: 129748191-129765063 on Build GRCh38
Transcripts
NM_001257308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3393112 c.57C>T p.D19D Chr.5: 129748247-129748247 on Build GRCh38
7290878 c.58G>A p.V20I Chr.5: 129748248-129748248 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs991416950 Chr.5: 129748486-129748486 on Build GRCh38
rs78847720 Chr.5: 129748344-129748344 on Build GRCh38
rs374406945 Chr.5: 129748255-129748255 on Build GRCh38
rs74757867 Chr.5: 129748361-129748361 on Build GRCh38
rs936911086 Chr.5: 129748429-129748429 on Build GRCh38
rs959561166 Chr.5: 129748463-129748463 on Build GRCh38
rs553913502 Chr.5: 129748564-129748564 on Build GRCh38
rs1016167827 Chr.5: 129748343-129748343 on Build GRCh38
rs1046610711 Chr.5: 129748456-129748456 on Build GRCh38
rs538073106 Chr.5: 129748262-129748262 on Build GRCh38
rs954210985 Chr.5: 129748569-129748569 on Build GRCh38
rs145318066 Chr.5: 129748258-129748258 on Build GRCh38
rs745621592 Chr.5: 129748290-129748290 on Build GRCh38
rs746763933 Chr.5: 129748319-129748319 on Build GRCh38
rs552070107 Chr.5: 129748248-129748248 on Build GRCh38
rs574518368 Chr.5: 129748291-129748291 on Build GRCh38
rs115273603 Chr.5: 129748460-129748460 on Build GRCh38
rs886294364 Chr.5: 129748304-129748304 on Build GRCh38
rs945555143 Chr.5: 129748238-129748238 on Build GRCh38
rs991055973 Chr.5: 129748247-129748247 on Build GRCh38
rs115791431 Chr.5: 129748405-129748405 on Build GRCh38
rs1028898185 Chr.5: 129748552-129748552 on Build GRCh38
rs535176789 Chr.5: 129748363-129748363 on Build GRCh38
rs978357384 Chr.5: 129748570-129748570 on Build GRCh38
rs187281858 Chr.5: 129748283-129748283 on Build GRCh38
rs756992670 Chr.5: 129748321-129748321 on Build GRCh38
rs4836471 Chr.5: 129748391-129748391 on Build GRCh38
rs751085596 Chr.5: 129748156-129748156 on Build GRCh38
rs542447478 Chr.5: 129748451-129748451 on Build GRCh38
rs996060965 Chr.5: 129748532-129748532 on Build GRCh38
rs934038791 Chr.5: 129748351-129748351 on Build GRCh38
rs531694356 Chr.5: 129748147-129748147 on Build GRCh38
rs541134864 Chr.5: 129748505-129748505 on Build GRCh38
rs1035173265 Chr.5: 129748171-129748171 on Build GRCh38
rs781212843 Chr.5: 129748288-129748288 on Build GRCh38
rs563222751 Chr.5: 129748423-129748423 on Build GRCh38
rs774937513 Chr.5: 129748362-129748362 on Build GRCh38
rs1054804283 Chr.5: 129748278-129748278 on Build GRCh38
rs538504066 Chr.5: 129748242-129748242 on Build GRCh38
rs1055362388 Chr.5: 129748440-129748440 on Build GRCh38
rs562038647 Chr.5: 129748452-129748452 on Build GRCh38
rs745767810 Chr.5: 129748356-129748356 on Build GRCh38
rs954044201 Chr.5: 129748368-129748368 on Build GRCh38
rs757086810 Chr.5: 129748175-129748175 on Build GRCh38
rs981172183 Chr.5: 129748136-129748136 on Build GRCh38
rs912738278 Chr.5: 129748237-129748237 on Build GRCh38
rs949061550 Chr.5: 129748449-129748449 on Build GRCh38
rs898978785 Chr.5: 129748512-129748512 on Build GRCh38
rs116144969 Chr.5: 129748428-129748428 on Build GRCh38
rs916259968 Chr.5: 129748448-129748448 on Build GRCh38
rs978249372 Chr.5: 129748409-129748409 on Build GRCh38
rs968260026 Chr.5: 129748565-129748565 on Build GRCh38
rs986878268 Chr.5: 129748181-129748181 on Build GRCh38
rs777718991 Chr.5: 129748297-129748297 on Build GRCh38
rs1004553048 Chr.5: 129748478-129748478 on Build GRCh38
rs78028068 Chr.5: 129748382-129748382 on Build GRCh38

View Full Product Details